48ab - Appl/Samples/Products/Resources/Contact
Advanced (scanning) transmission electron microscopy (S)TEM have become a necessary part of all leading-edge wafer-fabrication workflows due to an increasing demand for high-quality, rapid analysis and imaging of complex structures. Semiconductor designers need analytical tools that offer excellent flexibility and stability, as well as high-resolution imaging, in order to properly analyze and optimize device performance. (S)TEM characterization allows manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
Beyond imaging, a range of techniques are available on (S)TEM instrumentation that are critical for semiconductor device characterization, including; electron diffraction, for crystal orientation mapping or strain analysis, energy-dispersive X-ray spectroscopy (EDS), for qualitative or quantitative composition analysis, and electron energy-loss spectroscopy (EELS), for compositional and chemical information. Additionally, advanced imaging techniques, such as electron holography, tomography, Lorentz imaging and (integrated) differential phase-contrast (iDPC/DPC) imaging, are also available and are now routinely used in semiconductor labs.
Thermo Fisher Scientific is the world leader in TEM workflows for metrology, analysis, and pathfinding in semiconductor devices thanks to an uncompromising combination of high-resolution imaging and high analytical performance. Click through to the appropriate product pages below to learn more.
TEM imaging and analysis workflow example
Resources
Applications
Semiconductor-RD_1920x1080
Semiconductor research and development
Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
Semi-Metrology_1920x1080
Semiconductor metrology
Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
Semiconductor Failure Analysis
Semiconductor Failure Analysis
Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
Semi-Materials-Character_1920x1080
Semiconductor materials characterization
Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
Semiconductor power devices
Semiconductor power devices
Novel architectures and materials pose new challenges. Learn how to pinpoint faults and characterize materials, structures, and interfaces.
Display Module Failure Analysis
Semiconductor display technology
Display technologies are evolving to improve display quality and light conversion efficiency. Learn how metrology, failure analysis, and characterization solutions provide insights.
Samples
Semiconductor Materials and Device Characterization
As semiconductor devices shrink and become more complex, new designs and structures are needed. High-productivity 3D analysis workflows can shorten device development time, maximize yield, and ensure that devices meet the future needs of the industry.
Learn more ›
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Products
Style Sheet for Instrument Cards Original
Spectra 300
  • Highest-resolution structural and chemical information at the atomic level
  • Flexible high-tension range from 30-300 kV
  • Three lens condenser system
Talos F200E TEM
  • High-quality (S)TEM imaging of semiconductor and microelectronic devices
  • Precise, high-speed chemical characterization with EDS
  • Dedicated semiconductor-related applications
Metrios AX TEM
  • Automation options to support quality, consistency, metrology, and reduced OPEX
  • Leverages machine learning for superior autofunctions and feature recognition
  • Workflows for both in-situ and ex-situ lamella preparation
AutoTEM 5
  • Fully automated in situ S/TEM sample preparation
  • Support of top-down, planar and inverted geometry
  • Highly configurable workflow
  • Easy to use, intuitive user interface
Contact us
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›