Organism TypeMouseB82 M721 cells are murine fibroblasts devoid of endogenous EGFR. B82M721 cells stably express kinase-deficient human EGFR.
TissuesFibroblast
PhenotypeAdherent
Primaryno
ApplicationStable Cell Transfection
Products
Catalog # Name Size Price (USD) Qty
18324012 Lipofectamine™ Transfection Reagent, 1 mL Each
682.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.