Search Thermo Fisher Scientific
Organism Type | Mouse | ATCC# CLR1642: Cell line established from the lung of a C57BL mouse bearing a tumor resulting from an implantation of primary Lewis lung carcinoma. The cells are resistant to 1,3-bis-(2-chloroethyl)-1-nitrosourea, but are sensitive to methotrexate. The cells are reported to be highly tumorigenic, but weakly metastatic in mice. The cells form multilayers in flasks without actually becoming confluent. Tested and found negative for ectromelia virus (mousepox). |
Tissues | Lung | |
Phenotype | Either | |
Primary | no | |
Application | Stable Cell Transfection,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||