Search Thermo Fisher Scientific
Our broad portfolio of typing products offer low to high resolution detection to identify and match HLA. Assays inlclude those that have revolutionized the workflow by putting the most efficient next-generation sequencing method into the hands of laboratory professionals.
A probe-based hybrid capture assay for eleven HLA and seven non classical loci with the option of ABO genotyping. Universal protocol for whole blood and buccal swabs with reduced hands- on and turnaround time.
Multiplex PCR for genotyping for eleven HLA Class I and Class II loci with a single tube amplification for with short and long-range sequencing options. Streamlined workflow on multiple platforms with minimal hands-on time.
Combining Luminex xMAP technology with reverse sequence-specific oligonucleotide (rSSO) technology, high throughput DNA typing is achieved.
The LinkSēq line of real-time PCR products offers rapid genotyping of HLA and KIR with a simple workflow.
Amplify genomic DNA using SSP to determine HLA Class I and Class II alleles.
Our tissue typing family offers a host of HLA testing reagents optimized for use with our HLA typing trays.
Our exclusive LMT feature premixed monoclonal antibodies and rabbit complement for a fast and dependable method of determining Class I or Class II antigens.
Terasaki HLA Tissue Typing Trays contain well-characterized antisera that produce consistent and accurate HLA typing with complement-dependent cytotoxicity methods.
Not all products are CE marked or have 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.