Search Thermo Fisher Scientific
Explores the versatile applications of the Thermo Scientific iQ Series of instruments encompassing ambient gas monitoring, particulate detection, and emissions gas analysis. Discover how our advanced air monitoring solutions can help you meet stringent regulatory standards, even in the most challenging environments.
The iQ Series of gas monitors for ambient air quality monitoring leverage proven technologies, robust instrument designs and innovative data delivery systems to help ensure worker safety and regulatory compliance. Proactive communications and predictive diagnostics help eliminate unexpected trips to the field, so you can plan maintenance tasks around your schedule. Choose from a wide range of instruments that reliably measure air quality, low and high levels of criteria pollutants, various gases, and toxins according to your needs.
Continuously and reliably monitor particulates in ambient air using advanced air sampling technology. Measure critical regulatory parameters including PM-10 and PM-2.5 mass concentration as it exists in ambient air. Measuring mass concentrations utilizing one of our leading technologies: Tapered Element Oscillating Microbalance (TEOM), Beta Attenuation and Light Scattering Nephelometry.
Our emissions monitoring solutions are designed to measure and analyze emissions directly from industrial processes and point sources. Monitoring CO2, NOx, or other key pollutants, our technology provides accurate, real-time data to help industries meet regulatory requirements, optimize combustion processes, and reduce environmental impact. Engineered for reliability in high-temperature and high-concentration conditions, our systems help deliver the precision and efficiency needed for effective emissions management.
Service and product availability may vary by country and are subjected to varying regulatory requirements. Please contact your local sales representative for availability