Compliance and better air quality for all
As governmental regulations tighten to reduce air pollutants and toxins, our proven gas and particulate analysis technologies offer accurate, reliable air quality data in user-friendly formats. Explore how our solutions can help you efficiently meet regulatory requirements and provide cleaner air for everyone.
iQ Series Gas Analyzers
Discover the non-stop intelligence of the iQ Series of Gas Analyzers
Take control of your data and your day with an instrument designed to get smarter over time. Introducing the Thermo Scientific iQ Series of Gas Analyzers – experience non-stop intelligence that predicts and prevents problems. Featuring a user-friendly touch screen display, proactive communications, and the trusted reliability of an advanced Thermo Scientific gas analyzer.
Explore our wide range of gas analyzer models below:
iQ Series Particulate Monitors
icon for particulate monitoring
Setting new standards in precision, reliability, and ease of use, these monitors are designed to meet the most stringent regulatory standards. The 5014iQ and 5030iQ Monitors deliver real-time data accuracy and feature user-friendly interfaces, making them ideal for diverse environmental monitoring applications.
Browse the models below to discover the best match for your requirements
5014iQ Beta Attenuation Monitor
The Thermo Scientific 5014iQ Beta Attenuation Monitor is an automated ambient particulate measurement utilizing beta attenuation.
photo of Thermo Scientific 5014iQ Beta Attenuation Monitor
5030iQ SHARP Monitor
The Thermo Scientific 5030iQ SHARP Monitor is a real-time particulate monitor that utilizes beta attenuation and light scattering design.
photo of Thermo Scientific 5030iQ SHARP monitor
410iQ Series: Carbon Dioxide (CO₂)
icon for Carbon-dioxide
Human activity such as burning fossil fuels (coal, natural gas, and oil), solid waste, trees, and other materials, as well as some chemical reactions can release large amounts of CO₂ into the air. As such, continuous monitoring of carbon dioxide has become critical to compliance with every increasing government regulation for industry. Our CO₂ analyzers employ reliable Nondispersive Infrared (NDIR) technology for long instrument life and advanced optical filters for accuracy.
410iQ Carbon Dioxide Analyzer
The Thermo Scientific 410iQ Carbon Dioxide Analyzer utilizes advanced Non-Dispersive Infrared (NDIR) optical filter technology to measure concentrations of CO2.
Thermo Scientific 410iQ Carbon Dioxide Analyzer
410iQ High Level Carbon Dioxide Analyzer
The Thermo Scientific 410iQ High Level Carbon Dioxide Analyzer utilizes advanced Non-Dispersive Infrared (NDIR) optical filter technology to measure concentrations of CO2.
410iQ High Level Carbon Dioxide Analyzer
48iQ Series: Carbon Monoxide (CO)
icon for Carbon-monoxide
CO is a colorless, odorless gas that is naturally occurring but can cause sickness and death when present in large concentrations in ambient air. The gas is most commonly released as a by-product of fossil fuel combustion in vehicles and machinery and is a highly regulated pollutant. Our CO analyzers use advanced gas filter correlation technology to detect and measure CO gas concentrations in ambient air.
48iQ Carbon Monoxide Analyzer
The Thermo Scientific 48iQ Carbon Monoxide Analyzer utilizes gas filter correlation technology to measure the amount of carbon monoxide in the air.
photo of  Thermo Scientific 48iQ Carbon Monoxide Analyzer
48iQ Trace Level Carbon Monoxide Analyzer
The Thermo Scientific 48iQ Trace Level Carbon Monoxide Analyzer utilizes gas filter correlation technology to measure the amount of carbon monoxide in the air. 
photo of Thermo Scientific 48iQ Trace Level Carbon Monoxide Analyzer
450iQ Hydrogen Sulfide and Sulfur Dioxide (SO₂) Analyzer
icon for Hydrogen-sulfide-sulfer-dioxide
Sulfur dioxide (SO₂) is produced in significant quantity through the burning of fossil fuels during the production of energy and other industrial processes, including industrial transportation.  Sulfur dioxide can harm the human respiratory system and contribute to environmental pollution. Our sulfur dioxide analyzers measure up to 100ppm in ambient air using pulsed fluorescence technology.
450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer
The Thermo Scientific 450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer utilizes pulsed fluorescence technology. 
photo of  Thermo Scientific 450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer
42iQ Series: Nitrogen (N₂) and Nitrous Oxide Species (NO, NO₂, NOₓ NOᵧ)
symbol icon for nitrogen
Various nitrogen and nitrous oxide compounds (NO, NO2, NOY, NOx ) are emitted during agricultural, land use, industrial activities, fossil fuels combustion, solid waste management and the treatment of wastewater.  Our analyzers utilize chemiluminescence technology to measure the amount of nitrogen, and nitrous oxides in the air from sub-ppb levels up to 1000ppb.
42iQ NO-NO2-NOx Analyzer
The Thermo Scientific 42iQ NO-NO2-NOx Analyzer utilizes chemiluminescence technology to measure the amount of nitrogen oxides in the air from sub-ppb levels up to 100ppm.
photo of  Thermo Scientific 42iQ NO-NO2-NOx Analyzer
42iQ Low Source NO-NO2-NOx Analyzer
The Thermo Scientific 42iQ Low Source NO-NO2-NOx Analyzer utilizes chemiluminescence technology to measure the amount of nitrogen oxides in the air from ppb levels up to 500ppm.
photo of  Thermo Scientific 42iQ Low Source NO-NO2-NOx Analyzer
42iQ High Level NO-NO2-NOx Analyzer
The Thermo Scientific 42iQ High Level NO-NO2-NOx Analyzer utilizes chemiluminescence technology to measure the amount of nitrogen oxides in the air from 10 ppm up to 5000 ppm.
photo of  Thermo Scientific 42iQ High Level NO-NO2-NOx Analyzer
42iQ Trace Level NO-NO2-NOx Analyzer
The Thermo Scientific 42iQ Trace Level NO-NO2-NOx Analyzer utilizes chemiluminescence technology to measure the amount of nitrogen oxides in the air from sub-ppb levels up to 1000ppb. 
photo of  Thermo Scientific 42iQ Trace Level NO-NO2-NOx Analyzer
49iQ Series: Ozone (O3)
Ozone icon
Ozone, also known as trioxygen, is an inorganic molecule visible as a pale blue gas.  At high altitude, a layer of this gas provides protection against UV radiation from the sun.  Terrestrially, it is used as a powerful industrial oxidant and, even at very low concentrations, is harmful to animals and humans. Our ozone analyzers utilize UV Photometric technology to reliably detect and monitor Ozone.
49iQ Ozone Analyzer
The Thermo Scientific 49iQ Ozone Analyzer utilizes UV Photometric technology to measure the amount of ozone in the air from ppb levels up to 200ppm.
photo of  Thermo Scientific 49iQ Ozone Analyzer
49iQ Ozone Primary Standard
The Thermo Scientific 49iQ Ozone Primary Standard utilizes UV Photometric technology to produce ozone at a rate of up to 5000 ppb.
photo of  Thermo Scientific 49iQ Ozone Primary Standard
43iQ Series: Sulfur Dioxide (SO₂)
icon for sulfer dioxide
Sulfur dioxide (SO₂) is produced in significant quantity through the burning of fossil fuels during the production of energy and other industrial processes, including industrial transportation.  Sulfur dioxide can harm the human respiratory system and contribute to environmental pollution. Our sulfur dioxide analyzers measure up to 100ppm in ambient air using pulsed fluorescence technology.
43iQ Sulfur Dioxide Analyzer
The Thermo Scientific 43iQ Sulfur Dioxide Analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air.
photo of Thermo Scientific 43iQ Sulfur Dioxide Analyzer
43iQ High Level SO2 Analyzer
The Thermo Scientific 43iQ High Level SO2 Analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air to 1000ppm.
photo of Thermo Scientific 43iQ High Level SO2 Analyzer
43iQ Trace Level SO2 Analyzer
The Thermo Scientific 43iQ Trace Level SO2 Analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air to .05ppb. 
photo of Thermo Scientific 43iQ Trace Level SO2 Analyzer
111iQ Zero Air Supply
icon for Zero-air
Zero air, or air that has been purified by removing pollutants such as NO, NO₂, O3, SO₂, CO, and hydrocarbons, is used to test and calibrate spanning ambient air analyzers during routine maintenance.  Other analyzers can be calibrated using a multi-gas calibration device that performs zero, precision, Level 1 span checks, audits, and multi-point routines in accordance with US EPA requirements. We offer zero air supply and multi-gas calibrators to keep your systems running at peak performance.
111iQ Zero Air Supply
The Thermo Scientific 111iQ Zero Air Supply is designed for any application where pollutant free levels of NO, NO2, O3, SO2, CO and hydrocarbons are required, with flows up to 20 liters per minute at pressure of 30 PSI.
photo of  Thermo Scientific 111iQ Zero Air Supply
146iQ Multi-Gas Calibrator
icon for gas calibration
Calibrate ambient air and other gas monitoring instrumentation with our 146iQ Multi-Gas Calibrator, which generates gas standards. The output is used to calibrate instruments that perform zero, precision, Level 1 span checks, audits, and multi-point routines in accordance with USEPA requirements.
146iQ Multi-Gas Calibrator
The Thermo Scientific 146iQ Multi-Gas Calibrator supplies precise levels of ozone, carbon monoxide, non-methane hydrocarbons, sulfur dioxide, nitric oxide and nitrogen dioxide or other gases that a user may require.
photo of  Thermo Scientific 146iQ Multi-Gas Calibrator
Service and product availability may vary by country and are subjected to varying regulatory requirements. Please contact your local sales representative for availability