Search Thermo Fisher Scientific
High Resolution Inductively Coupled Plasma Mass Spectrometry is a technique used for elemental analysis of solutions and solid samples via laser ablation. The superior ionization of inductively coupled plasma source is combined with a magnetic sector multicollector mass spectrometer resulting in ultimate sensitivity.
The high mass resolution potential of a sector-field mass spectrometer can physically distinguish and separate an analyte from interferences by exploiting small differences in mass-to-charge. This means that almost all elements in the periodic table can be analyzed without the requirement for complicated reaction gases regardless of sample matrix.
The Thermo Scientific Element HR-ICP-MS has a large dynamic range of more than 12 orders of magnitude. As such, it can be used to measure matrix elements (%), traces (ppm) and ultra-traces (ppb). There are a wide range of applications that use HR-ICP-MS from geoscience research, environmental testing, process chemicals and semiconductor applications.
The workhorse for semiconductor applications. The high performance, double-focusing magnetic sector field Element 2 HR-ICP-MS is optimized for ease-of-use, stability and productivity. Get access to a powerful technique for solving specific problems with excellent sensitivity and signal-to-noise ratio.
Learn more about the Element 2 HR-ICP-MS ›
The gold standard for full elemental analysis of laser ablation and solution samples. The 12 orders of magnitude linear dynamic range of the Element XR HR-ICP-MS enables analysis of any matrix element straight without further dilution, ideal for laser ablation analysis or applications with extreme isotope ratios.
Learn more about the Element XR HR-ICP-MS ›
The Thermo Scientific Jet Interface, in combination with a desolvating nebulizer system, increases sensitivity by 10 to 20 times compared to standard wet plasma.
Learn more about Jet Interface option ›
Application notes:
From battery through to medical research, the Element HR-ICP-MS is the instrument of choice for high precision elemental analysis. Learn more the applications of HR-ICP-MS below and dive deeper by visiting our Isotope Fingerprinting and Isotope Research pages.
Learn how the Element HR-ICP-MS is used to monitor contamination in workers in nuclear power plants and military personnel.
The low detection limits of the Element HR-ICP-MS are ideal for analysis of high-purity chemicals.
A high mass resolution and large dynamic range makes the Element HR-ICP-MS the workhorse for elemental mapping of geological samples.
Coupling chromatography and HR-ICP-MS allows researchers to quantify unknown reaction products occurring as a result of aging within the electrolyte.
Environmental samples can be differentiated based on geographical region, botanical origin, and mineralization processes.
Natural variations in the relative abundances of isotopes and elements can explain geological systems on Earth and in the cosmos.
Our HR-ICP-MS have low detection limits ideal to monitoring ultratrace levels of elemental impurities while maintaining a high sample throughput.
Learn more about semiconductor and high purity metal analysis ›
Improve performance, convenience and flexibility with inductively coupled plasma mass spectrometer consumables and accessories designed specifically for your instrument.
Performing multi-element determination of unknown samples containing major, minor and trace elements across a wide range of sample types can be easy.
JKC detects metal elements of lowest concentrations. Challenges of interferences are overcome with HR-ICP-MS.
HR-ICP-MS at the MEET Battery Research Center
Speed up your lab, with High Resolution ICP-MS. Now you can perform quantitative multi-element analyses at trace level, with the highest sensitivity.
* Required field