Glycan Analysis Information
Glycan and Glycoprotein analysis
Glycans serve a variety of structural and functional roles in membrane and secreted proteins, with the majority of proteins undergoing some degree of glycosylation during their synthesis. Regulatory agencies worldwide, including the FDA and EMA, are increasing the demands placed upon manufacturers to comprehensively analyze the glycosylation of their therapeutics, and also to demonstrate how process can affect glycan composition.
Changes in the glycosylation pattern of protein biotherapeutics have been shown to impact their half-life, stability, safety and efficacy. In general, glycans are divided into two main groups; O-linked and N-linked glycans. O-linked glycosylation involves the attachment of oligosaccharides to serine or threonine amino acid residues through an oxygen atom, and N-linked glycosylation involves the attachment of oligosaccharides to asparagine amino acid residues through a nitrogen atom.
More than 60% of therapeutic proteins are post translationally modified following biosynthesis by the addition of N- or O-linked glycans. Glycosylation of biotherapeutics can be influenced by a multitude of process related factors, such as pH, carbon source, dissolved oxygen, temperature during manufacture, and by the choice of expression system.
Whilst glycosylation is the most common post-translational modification of proteins, it is also the most demanding from an analytical point of view. In order to maintain consistent biotherapeutic glycosylation patterns, efficient manufacturing processes and effective glycan characterization are required. The complete analysis of a glycoprotein provides information on the primary structure of the oligosaccharides, as well as their variation at individual glycosylation sites being achieved through a number of different analytical approaches.
To view the products and workflows related to glycan analysis please visit our Glycan Analysis product page.
ICH (Q6B) recommends 6 test approaches for characterization and confirmation of biological products: "For glycoproteins, the carbohydrate content and structure (neutral sugars, amino sugars, and sialic acids) is determined."
 Explore the workflows
What is my glycosylation pattern? Intact glycoprotein analysis
general-intact-mass-and-intact-glycopretein-workflow-740x220
Intact glycoprotein profiling is used to ascertain the pattern and degree of glycosylation. Due to the heterogeneity of the attached glycan moieties, intact glycoprotein profiling is best performed using high resolution accurate mass (HRAM) mass spectrometry (MS), together with chromatographic separation to gain full insight into the various glycoforms present on a protein or biotherapeutic.
Where do my glycans reside? Glycopeptide profiling
general-glycopeptide-workflow-740x220
Glycan analysis can also be performed at the peptide level, with the goal of obtaining both glycan composition and peptide sequence at the site of glycosylation.
Which glycans are in my drug and what is their concentration? Glycan structure determination
general-labeled-glycans-workflow-740x220
Monitoring of specific glycan species or determination of relative quantities of a particular set of glycans provides important information for the development of biotherapeutics. Due to the complexity of glycan structures, quantitation and identification is performed upon release of glycans from the protein. N-linked glycans are released by enzymatic treatment, whereas O-linked glycans need to be released by chemical methods, as no enzyme exists for this purpose. Glycans have no chromophore and therefore have a poor response with conventional LC-UV detection; as such, they are commonly labeled with fluorescent tags prior to high sensitivity analysis by LC-fluorescence detection, the most common label being 2-aminobenzamide (2-AB). MS has emerged as one of the most powerful tools for glycan structure elucidation. However, as most glycans do not ionize efficiently 2-AB labeling can also be performed to improve sensitivity.
Labeled glycan protocols in AppsLab Library of Methods
Get instant access to over a dozen IC and UHPLC methods for 2-AA, 2-AB labeled N-glycans from therapeutic proteins. Or access our latest featured application from NIBRT Ireland on fast profiling of the N-glycan population in biotherapeutic antibodies by UHPLC-FLD with MS confirmation.
 Labeled N-glycan AppsLab methods
 Featured NIBRT application
What is the composition of sugars in my drug entity? Monosaccharide and sialic acid analysis
general-monosaccharides-and-sialic-acids-workflow-740x220
Monosaccharide composition, namely fucose, galactosamine, glucosamine, galactose, glucose and mannose, is routinely determined as the number and composition of sugar units bound to the protein and can impact the efficacy of biotherapeutics. Monosaccharides are weak acids and can be separated using anion-exchange chromatography under basic conditions. Samples are acid hydrolyzed to release monosaccharides and analyzed by high-performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD) after chromatographic separation.
Glycosylation plays an important part in protein structure and function. Factors such as cell age, culture conditions, and purification affect the nature of protein glycosylation.
 Learn more
Featured glycan analysis information learning content
Applications compendium
HPAE-PAD is a well-established method for carbohydrate analysis, separating carbohydrates through specific interactions between the hydroxyl and carboxyl groups of carbohydrates based on charge, size, composition, isomerism, and linkage.
White paper
Carbohydrate analysis, also known as glycosylation analysis, glycan analysis, or sometimes simply as sugar analysis, is of growing importance to sciences as diverse as pharmaceutical drug development, cancer research, stem cell research and biofuels development.
 Download the white paper
Featured videos
Ion Chromatography - the SGS workhorse for therapeutic glycoprotein analysis
Characterization and monitoring of the glycan profile of therapeutic proteins is critical for drug efficacy and safety. Ion chromatography (IC) is a workhorse technique for this application used by SGS in West Chester PA.
Video Player is loading.
Current Time 0:00
Duration 4:18
Loaded: 0%
Stream Type LIVE
Remaining Time 4:18
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextIon Chromatography - the SGS workhorse for therapeutic glycoprotein analysis
        Characterization and monitoring of the glycan profile of therapeutic proteins is critical for drug efficacy and safety. Ion chromatography (IC) is a workhorse technique for this application used by SGS in West Chester PA.
      2. Now Playing
        Up NextBioPharMoore Episode 9: Modern IC, the USP and me
        Discover how modern ion chromatography solutions are perfect for the pharmaceutical laboratory and aligned with pharmacopeial monograph modernization
      3. Now Playing
        Up NextThe Sweet Revolution in Glycan and Antibody Separations
        This webinar discusses the difficulties and also the current technologies used to maximize glycan separation capabilities and structural elucidation.
      4. Now Playing
        Up NextOrbitrap Hits the Sweet Spot: Thermo Fisher Scientific Solutions For Glycan Analysis
        Mass spectrometry has emerged as one of the most powerful tools for the structural elucidation of glycans. Due to its sensitivity of detection and its ability to analyze complex mixtures of glycans, MS based methods is the maximization of structural
    Search all glycan analysis information resources
    TypeTitleProduct focusYear
    Application CompendiumMonosaccharide and Sialic Acid Determinations in Biosimilars Using HPAE-PADIon Chromatography2015
    Application NoteHILIC – an alternative separation technique for glycopeptidesHPLC / UHPLC2017
    Application NoteAutomated Glycan Structural Isomer Differentiation Using Bioinformatics ToolLife Science Mass Spec2011
    Application NoteComprehensive protein glycosylation comparison of an innovator monoclonal antibody to a candidate biosimilar by HILIC UHPLC analysisHPLC / UHPLC2017
    Application NoteEvaluating Protein Glycosylation in Limited-Quantity Samples by HPAE-PADIon Chromatography2015
    Application NoteFast profiling of the N-glycan population in biotherapeutic antibodies by UHPLC-FLD with MS confirmationHPLC / UHPLC2017
    Application NoteHigh Performance Anion-Exchange Chromatography with Pulsed Amperometric Detection (HPAE-PAD) Analysis of Mannose-6-PhosphateIon Chromatography2010
    Application NoteIntegrated LC/MS Workflow for the Analysis of Labeled and Native N-Glycans from Proteins Using a Novel Mixed-Mode Column and a Q Exactive Mass SpectrometerWorkflow2014
    Application NoteLabel-Free Analysis by UHPLC with Charged Aerosol Detection of Glycans Separated by Charge, Size, and Isomeric StructureHPLC / UHPLC2015
    Application NoteSeparation of 2AA-Labeled N-Linked Glycans from Glycoproteins on a High Resolution Mixed-Mode ColumnChemistries and Consumables2013
    Application NoteSeparation of 2AA-Labeled N-Linked Glycans from Human IgG on a High Resolution Mixed-Mode ColumnChemistries and Consumables2013
    Application NoteSeparation of 2AB-Labeled N-Linked Glycans from Bovine Fetuin on a Novel Ultra High Resolution Mixed-Mode ColumnChemistries and Consumables2013
    Application NoteAnalysis of Gentamicin Using a pH Stable Specialty Column for Aminoglycoside Antibiotics SeparationChemistries and Consumables2015
    Application NoteAnalysis of Spectinomycin Using a pH Stable Specialty Column for Aminoglycoside Antibiotics SeparationChemistries and Consumables2015
    Application NoteThe Determination of Carbohydrates, Alcohols, and Glycols in Fermentation BrothsIon Chromatography2013
    Application NoteAn ultrafast, batch-to-batch comparison of monoclonal antibody glycosylationHPLC / UHPLC2017
    Application NoteComparison of Therapeutic Antibody Originator and Biosimilar Glycosylation Using an Integrated Glycan Labeling SolutionHPLC / UHPLC2018
    Application NotebookHigh-Performance Anion-Exchange Chromatography with Pulsed Amperometric Detection (HPAE-PAD) - Carbohydrates Analysis Application NotebookIon Chromatography2014
    Application UpdateImproved Separation of Aminobenzamide (2-AB)-Labeled N-glycans from Human α1 Acid-Glycoprotein for Analysis by HPAE-FLDIon Chromatography2016
    Application UpdateRapid Screening of Sialic Acids in Glycoproteins by HPAE-PADIon Chromatography2011
    AppsLabIn-depth protein glycosylation comparison of a biosimilar candidate by hydrophilic interaction UHPLCChemistries and Consumables2016
    AppsLabA fast separation of 2-AB labelled N-Glycans from polyclonal human serum IgGChemistries and Consumables2016
    AppsLabA high resolution separation of 2-AA derivatized N-Glycans from a commercial chimeric IgG1 monoclonal antibody (Infliximab)Chemistries and Consumables2016
    AppsLabUltrafast hydrophilic interaction UHPLC analysis of 2-AA labelled N-glycans from monoclonal antibodies (mAbs)Chemistries and Consumables2016
    ArticleGlycobiology ToolsLife Science Mass Spec2016
    BrochureCarbohydrate analysis with HPAE-PADIon Chromatography2013
    BrochureThermo Scientific Glycan Analysis for Biotherapeutics: Join the Sweet Revolution in BiopharmaceuticalsWorkflow2015
    eBookGlycan Analysis Workflows for BiotherapeuticsWorkflow2015
    EditorialGlycopioneerOther2015
    PosterMiddle-down Approach for Monitoring Monoclonal Antibody Variants and DeglycosylationChemistries and Consumables2016
    Poster NoteOrbitrap Based Mass Spectrometric Characterization of Antibody Drug Conjugates Engineered through Antibody GlycansLife Science Mass Spec2015
    Poster Note2-Amino Benzamide Labeling of Oligosaccharides: How Much Sialic Acid Is Lost?Ion Chromatography2014
    Poster NoteAn Ultra High Resolution Column and Mass Spectrometer for isomeric Separation and the Structural Identification of Labeled N-linked GlycansChemistries and Consumables2014
    Poster NoteDirect Measurement of Sialic Acids Released From Glycoproteins, by High Performance Liquid Chromatography and Charged Aerosol DetectionHPLC / UHPLC2016
    Poster NoteHPAE-FLD Method for Separation of Human α1 Acid-Glycoprotein and Bovine Fetuin 2-Aminobenzamide (2AB)-Labeled OligosaccharidesIon Chromatography2016
    Poster NoteLabel-Free Analysis by UHPLC with Charged Aerosol Detection of Glycans Separated by Charge, Size, and Isomeric StructureHPLC / UHPLC2015
    Poster NoteLabel-Free Profiling of O-linked Glycans by UHPLC with Charged Aerosol DetectionHPLC / UHPLC2016
    PresentationAnalytical Strategies for Studying Glycosylation of BiopharmaceuticalsWorkflow2016
    PresentationSix analytical strategies for studying glycosylation of biopharmaceuticalsWorkflow2016
    Technical NoteGlycoprotein Monosaccharide Analysis Using HPAE-PAD with Eluent GenerationIon Chromatography2016
    Technical NoteGlycoprotein monosaccharide analysis using HPAE-PAD with manually prepared eluentIon Chromatography2017
    Technical NoteHPAE-PAD N-linked oligosaccharide profiling of IgGIon Chromatography2017
    Technical NoteReview of peer-reviewed HPAE-PAD glycoprotein oligosaccharide analysis scientific literature evaluating electrochemical response.Ion Chromatography2013
    Technical NoteUHPLC Analysis of 2-AB-labeled Dextran Ladder and Assignment of Glucose Units to Unknown GlycansChromatography Data Software2014
    Technical NoteUHPLC Analysis of 2-Aminobenzamide-Labeled Glycans with the Vanquish Flex SystemHPLC / UHPLC2015
    WebinarIon Chromatography2013
    WebinarLife Science Mass Spec2015
    WebinarLife Science Mass Spec2015
    WebinarWorkflow2015
    WebinarWorkflow2015
    White PaperHPAE-PAD for the Analysis of CarbohydratesIon Chromatography2013
    White PaperOrbitrap Fusion MS for Glycan and Glycopeptide AnalysisLife Science Mass Spec2015
    Application NoteSubunit Analysis Approach for the Determination of Fucosylation Levels in Monoclonal AntibodiesHPLC / UHPLC2018
    Application NoteA Quick and Accurate Comparison of Biosimilar and Originator BiotherapeuticsHPLC / UHPLC2018
    Application NoteHPAE-PAD method for determination of Hib capsular polysaccharide contentHPLC / UHPLC2021
    Application NoteAn Improved Method for Galactosyl Oligosaccharide CharacterizationHPLC / UHPLC2022
    Share