Antibiotics Analysis Information
Ensuring the safety of antibiotics
Antibiotics, or antibacterials, are antimicrobial drugs used to treat or prevent bacterial infections. Antibiotics are heralded as the medicinal heroes of the 20th century. However, their effectiveness and accessibility has led to overuse and, in recent years, consequent antibiotic resistance.
Antibiotics are divided into classes  based upon their method of production or bactericidal method of action. They may be produced biologically (fermentation), biologically and chemically (semi-synthetic), or by chemical synthesis alone.
Analysis of biological/natural product antibiotics
E.g., aminoglycosides, macrolides, some tetracylines, lipopeptides and glycopeptides
Antibiotics produced through fermentation processes are less predictable, less controllable and more complex than synthetic antibiotics. For this reason the variability in products derived from fermentation is often greater than products derived by chemical synthesis. The impurity profile of a fermentation product may also be more complex and less predictable than that of a synthetic product.
One of the most common antibiotic groups which is biologically synthesized are the aminoglycosides. An aminoglycoside is a molecule composed of a sugar group and an amino group.
GentamicinR1R2R3
C1aHHH
C2HCH3H
C2bCH3HH
C2aHHCH3
C1CH3HH
Streptomycin was the first aminoglycoside antibiotic discovered and used in clinical therapy. These antibiotics are now widely used as clinical and veterinary medicines to treat bacterial infections because of their protein synthesis inhibition capability, leading to cell death. However, these antibiotics can have serious side effects and cause varying degrees of toxicity. It is important to develop sensitive and reliable analytical methods to characterize and quantify drug purity and detect minor degradants or impurities. Based on the nature of their production (fermentation), there are often mixtures of related components (congeners, isomers) and fractions which must be monitored and controlled.
Reversed-phase HPLC separation of gentamicin on the Thermo Scientific Acclaim AmG C18 column with charged aerosol detection (CAD) demonstrates speed and resolution advantages with optimised solvent composition.
Reversed-phase HPLC separation of gentamicin on the Acclaim AmG C18 LC column with Dionex Corona Veo RS (rapid separation) charged aerosol detection (CAD) demonstrates speed and resolution advantages with optimized solvent composition.
ColumnAcclaim AmG C18, 3 µm
Dimensions4.6 x 150 mm
Mobile Phase A100 mM TFA
Mobile Phase BAcetonitrile
Flow Rate1 mL/min
Inj. Volume5 µL
Temperature30 °C
DetectionCorona Veo RS (Filter = 5.0s; Evaporation Temp = 35 °C; Data Rate = 5 Hz; Power Function = 1.00)
SampleGentamicin (1 mg/mL)
Peaks
  1. C1a
  2. C2
  3. C2b
  4. C2a
  5. C1
As well as being structurally-related, many aminoglycoside antibiotics are actually synthesised from one another. For example, sisomicin is a broad spectrum aminoglycoside isolated from the fermentation broth of Micromonospora. Netilmicin is a semi-synthetic aminoglycoside antibiotic prepared from sisomicin. Both sisomicin and netilmicin are mainly used in the treatment of severe infections, particularly those resistant to gentamicin. Etimicin is semi-synthesized from gentamicin C1a, and so on.
Analysis of aminoglycosides and their related impurities is often achieved by ion-pairing reversed-phase (RP) high performance liquid chromatography (HPLC), based on their hydrophilic and positively charged nature. However, due to the lack of a suitable chromophore, aminoglycosides cannot be detected by ultraviolet (UV). Corona charged aerosol detectors (CAD), evaporative light scattering detectors (ELSD), mass spectrometers (MS), and electrochemical detectors are generally used to detect these compounds without prior derivatization.
Separation of gentamicin in various substances
Separation of gentamicin in A) topical cream extract, B) ophthalmic ointment extract, C) topical ointment extract, and D) diluted ophthalmic solution (25% signal offset) on the Acclaim RSLC PA2 LC column with a HFBA/TFA gradient. The column temperature was maintained at 15 °C, which allowed greater separation between gentamicin C2b and gentamicin C2.
ColumnAcclaim RSLC PA2, 2.2 µm Analytical (2.1 x 100 mm)
Mobile Phase A0.025:95:5 HFBA:DI water:acetonitrile
Mobile Phase B0.3:95:5 TFA:DI water:acetonitrile
Gradient
  • From 0 to 3 min: 1-10% mobile phase B (99-90% mobile phase A)
  • From 3 to 8 min: 10-100% mobile phase B (90-0% mobile phase A)
  • From 8 to 11 min: 100% mobile phase B
  • 4 min of equilibration at 99% mobile phase A before injection
Flow Rate0.45 mL/min
Inj. Volume1.0 µL
Temperature15 °C
DetectionCAD (Corona ultra RS, nebulizer temperature 15 °C, low filter, 60 Hz data collection rate)
Sample
  • A) Gentamicin sulfate cream
  • B) Gentamicin sulfate ophthalmic ointment
  • C) Gentamicin sulfate ointment
  • D) Gentamicin sulfate ophthalmic solution
Peaks
  1. Unretained ions from matrix
  2. Garamine-like compound
  3. Sisomicin
  4. Gentamicin C1a
  5. Gentamicin C2
  6. Gentamicin C2b
  7. Gentamicin C2a
  8. Gentamicin C1
The USP monographs for some aminoglycoside drug substances (and drug products made from them) often involve high pressure anion exchange (HPAE) ion chromatography (IC) assays with integrated pulsed amperometric detection (IPAD).
Determination of kanamycin A, kanamycin B and tobramycin on a Thermo Scientific Dionex CarboPac PA1 anion exchange column by HPAE-IPAD.
Determination of kanamycin A, kanamycin B and tobramycin on a Dionex CarboPac PA1 anion exchange column by HPAE-IPAD.
ColumnDionex CarboPac PA1 guard, 4 x 50 mm
Dionex CarboPac PA1 guard, 4 x 250 mm
Eluent2 mM KDH
Eluent SourceDionex EGC-500 KOH cartridge, with CR-ATC 600 trap column, Dionex high pressure degasser
Flow Rate0.5 mL/min
Column Temp.30 °C
Detector Compart.30 °C
Inj. Volume20 µL
DetectioniPAD, AAA-Direct Au disposable electrode, 0.002" thick gasket
Reference ElectrodepH/Ag/AgC1, pH mode
WaveformAAA-Direct, versus pH, 1.67 Hz
Peaks
  1. Void volume
  2. System peak
  3. Kanamycin A
  4. Kanamycin B
  5. Tobramycin
  6. Oxygen dip
Analysis of synthetic antibiotics
E.g., sulphonamides, quinolones, oxazolidinones
Chemically synthesised antibiotics often involve a number of intermediate compounds in their preparation, and these may remain as impurities in the final product. Chemical antibiotics are usually assayed by HPLC with UV detection, but where a chromophore is absent in either the active pharmaceutical ingredient (API) or impurities, IC with suppressed conductivity detection is considered the best alternative for selective determination.
Overlay of seven chromatograms of 0.1 mg/mL linezolid spiked with 10 µg/L morpholine, separated on a Thermo Scientific Dionex IonPac CS19 column set.
Overlay of seven chromatograms of 0.1 mg/mL linezolid spiked with 10 µg/L morpholine, separated on a Dionex IonPac CS19 column set.
ColumnIonPac CG19 Guard, 2 x 50 mm
IonPac CS19 Analytical, 2 x 250 mm
Eluent7.5mM MSA
Eluent SourceEGC-500 MSA with Dionex CR-CTC 500
Flow Rate0.25 mL/min
Inj. Volume100 µL
Conc ColumnDionex IonPac TCC-ULP1
DetectionSuppressed conductivity, Dionex CSRS 300, 2 mm, 7 mA, recycle mode
Peaks
  1. Sodium
  2. Ammonium
  3. Potassium
  4. Morpholine 10
  5. Magnesium
  6. Calcium
Analysis of semi-synthetic antibiotics
E.g., beta-lactams (penicillins, cephalosporins, penems, carbapenems, and monobactams), some tetracylines and glycopeptides
Semi-synthetic antibiotics generally have fewer impurities than their biological counterparts. Impurities may include fermented starting material with related impurities, synthesis by-products, synthesis intermediates, and degradation products.
Beta-lactam antibiotics are a class of broad-spectrum antibiotics, consisting of all antibiotic agents that contain a beta-lactam ring in their molecular structures. These antibiotics function through the inhibition of bacterial cell wall synthesis. Following extensive use, some bacterial populations have shown ability to develop resistance to beta-lactams and become more virulent.
Cephalosporins are a members of the β-Lactam antibiotics class, first discovered and isolated from the fungus Cephalosporum Acremonium. The core structure of these antiobitics features a number of hydrogen bond donors and acceptor groups, as well as an acid function. Together these groups contribute towards the polarity of this class of compounds.
Cephalosporins are a members of the β-Lactam antibiotics class, first discovered and isolated from the fungus Cephalosporum acremonium. The core structure of these antiobitics features a number of hydrogen bond donors and acceptor groups, as well as an acid function. Together these groups contribute towards the polarity of this class of compounds.
While beta-lactam antibiotics are similar to one another in many ways, they may differ in pharmacokinetics, antibacterial activity, and potential to cause serious allergic reactions. Some beta-lactam intermediate compounds and derivatives (from fermentation and/or synthesis) also possess similar sensitization and cross–reactivity properties. Beta-lactam intermediate compounds, such as β-lactam antibiotic API precursors, can undergo molecular changes or purification before use in manufacture. As a result of these changes, the intermediate compounds may develop antigenic characteristics that can produce allergic reactions. Drug manufacturers are required to take steps to control for the risk of cross-contamination and impurities for all beta-lactam products.
Separation of a mixture of cephalosporins on a Thermo Scientific Syncronis aQ 3 µm particle column. Good separation was achieved in a total run time of six minutes, with an elution window of 1.8 minutes for all four analytes. A controlled interaction mechanism and selectivity of the Syncronis aQ polar endcapping group is demonstrated.
Separation of a mixture of cephalosporins on a Syncronis aQ LC column with 3 µm particle size. Good separation was achieved in a total run time of six minutes, with an elution window of 1.8 minutes for all four analytes. A controlled interaction mechanism and selectivity of the Syncronis aQ polar endcapping group is demonstrated.
Using the Thermo Scientific Chromeleon Chromatography Data System and simultaneous detection with UV and MS detectors provides complementary information for impurity profiling. The MS component channel with the extracted ion chromatograms of seven antibiotic standards was overlaid to the UV trace. The two traces could be perfectly aligned by taking into account the delay time between the two detectors. This simple comparison reveals the presence of an UV active impurity.
Using the Chromeleon Chromatography Data System and simultaneous detection with UV and MS detectors provides complementary information for impurity profiling. The MS component channel with the extracted ion chromatograms of seven antibiotic standards was overlaid to the UV trace. The two traces could be perfectly aligned by taking into account the delay time between the two detectors. This simple comparison reveals the presence of an UV active impurity.
Often beta-lactamase inhibitors, such as clavulanate (clauvulanic acid) and sulbactam are co-formulated with beta-lactam antibiotics to increase their effectiveness through the counteraction of bacterial resistance. Impurities associated with the inhibitor as well as the antibiotic therefore need to be controlled and monitored.
Chromatographic comparison of clavulanate on the Thermo Scientific Dionex IonPac AS11 column A) without and B) with 4 µg/mL (0.8%) 2-ethylhexanoic acid. This column has high capacity, low surface hydrophobicity and is able to separate a wide range of inorganic and organic anions in complex matrices.
Chromatographic comparison of clavulanate on the Dionex IonPac AS11 column A) without and B) with 4 µg/mL (0.8 %) 2-ethylhexanoic acid. This column has high capacity, low surface hydrophobicity and is able to separate a wide range of inorganic and organic anions in complex matrices.
ColumnIonPac AG11, AS11, 2 mm
Eluent3 mM KOH from 0 to 10 min,
3 to 60 mM KOH from 10 to 10.1 min,
60 mM KOH from 10.1 to 20.1 min
Eluent SourceEGC II KOH with CR-ATC
Flow Rate0.25 mL/min
Inj. Volume5 µL
Temperature30 °C
DetectionSuppressed conductivity, ASRS 300 2 mm, recycle mode,
2 mA suppressor current during 3 mM KOH,
switch to 38 mA at 10.1 min
SamplesClavulanate with and without 4 µg/mL (0.8 %)
2-Ethylhexanoic acid
Peaks2-Ethylhexanoic acid (4 µg/mL – 0.8 %)
Antibiotics in bioprocess of biopharmaceuticals
Antibiotics are heavily used in the production of biopharmaceuticals. Mammalian cell lines that express biotherapeutic proteins, such as antibodies, must be maintained over several weeks. They are fed with culture media supplemented with various vitamins, growth factors, and antibiotics to avoid contamination and growth failure. Testing of residual antibiotics in the product is required to ensure patient safety.
Analysis of a sample of cell lysate after tigecycline treatment (A) and the same sample spiked with 0.05 μg/mL tigecycline (B) using an on-line SPE-HPLC-UV method. This is a convenient method to determine trace amounts of tigecycline in tigecycline-treated cells which cannot be determined using a routine HPLC-UV detection method.
Analysis of a sample of cell lysate after tigecycline treatment (A) and the same sample spiked with 0.05 μg/mL tigecycline (B) using an on-line SPE-HPLC-UV method. This is a convenient method to determine trace amounts of tigecycline in tigecycline-treated cells which cannot be determined using a routine HPLC-UV detection method.
Conditions
On-Line SPE
ColumnAcclaim PolarAdvantage II (PA2) Guard Cartridge, 5 µm, 4.6 x 10 mm (P/N 069699)
Mobile PhaseDI water
Flow Rate1.0 mL/min
Inj. Volume1500 µL on the on-line SPE cartridge
Separation
ColumnAcclaim 120, C18, 3 µm Analytical, 3.0 x 150 mm (P/N 063691)
Mobile PhasePhosphate buffer/CH3CN (85:15. v/v)
Flow Rate0.6 mL/min
Autosampler Temp.4 °C
Column Temp.30 °C
DetectionUV absorbance at 247 nm
Featured learning content
Brochure
Ion Chromatography in Pharma & BioPharma Brochure
Discover the unique benefits of modern ion chromatography systems for the analysis of pharmaceuticals and biopharmaceuticals.
Applications notebook
Pharmaceutical Applications Notebook—Antibiotics
Learn more in this applications notebook, which has been compiled to help the pharmaceutical scientist by providing a wide range of application examples relevant to the analysis of antibiotics.
Application
Determination of tobramycin using HPAE-IPAD on a compact ion chromatography system
Discover a HPAE-IPAD method for the determination of kanamycin A, kanamycin B, and tobramycin using the high-pressure-capable Dionex Integrion HPIC system.
Application
Analysis of Gentamicin Using a pH Stable Specialty Column for Aminoglycoside Antibiotics Separation
This study describes a high-performance separation of gentamicin congeners and their related compounds using ion-pairing reversed-phase liquid chromatography (IP-RPLC).
Featured video
Discover how modern ion chromatography solutions are perfect for the pharmaceutical laboratory and aligned with pharmacopeial monograph modernization.
Search our antibiotics analysis literature library
Analyte
(click title for methods)
ReferencesTechnique
 AmikacinHPLC, IC
 AmoxicillinIC
 AmpicillinIC
ApramycinHPLC
ArbekacinHPLC
 Cefaclor (Cephaclor)HPLC
 Cefadroxil (Cefadroxyl, Cephadroxyl, Cephadroxil)HPLC, IC
 Cefalexin (Cephalexin)HPLC, IC
 Cefaloridine (Cephaloridine)HPLC, IC
Cefalothin (Cephalothin)IC
 Cefamandole (Cephamandole)HPLC
Cefapirin (Cephapirin)IC
 Cefazolin (Cephazolin, Cefazoline, Cephazoline)HPLC, IC
 CefdinirNo referencesHPLC
 Cefepime (Cephapime)HPLC, IC
 CefiximeNo referencesHPLC
 Cefotaxim (Cefotaxime, Cephotaxime, Cephotaxim)HPLC, IC
 Cefradine (Cephradine)HPLC, IC
 Ceftazidime (Cephtazidime)No referencesHPLC
 CeftriaxoneNo referencesHPLC
 ChloramphenicolHPLC
Cinchophen (Cinchofen)HPLC
 CinoxacinHPLC
 CiprofloxacinHPLC
 Clavulanate (Clavulanic acid; beta lactamase inhibitor)IC
 CloxacillinHPLC, IC
 DanofloxacinHPLC
 DihydrostreptomycinHPLC, IC
 DoxorubicinNo referencesHPLC
 ErythromycinHPLC
EtimicinHPLC
 FlucloxacillinNo referencesHPLC
 FosfomycinNo referencesHPLC
GentamicinHPLC
 KanamycinHPLC, IC
 Kanamycin AIC
 Kanamycin B (Bekanamycin)IC
 LincomycinHPLC, IC
 LinezolidHPLC, IC
 LomefloxacinHPLC
 MetronidazoleNo referencesHPLC
 MinocyclineNo referencesHPLC
 MoxifloxacinNo referencesHPLC
 Nalidixic AcidHPLC
 Neomycin (Neamine, Paromamine)HPLC, IC
NetilmicinHPLC
 NitrofurantoinHPLC
 NorfloxacinNo referencesHPLC
 OfloxacinHPLC
 Oxolinic AcidHPLC
 OxytetracyclineHPLC
 ParomomycinHPLC, IC
 Penicillin GHPLC, IC
 Penicillin VHPLC, IC
 PolymyxcinNo referencesHPLC
RibostamycinHPLC
SisomicinHPLC, IC
 SpectinomycinHPLC
 StreptomycinHPLC, IC
 StreptomycinHPLC, IC
 StreptomycinHPLC, IC
 StreptomycinHPLC, IC
 SulbactamNo referencesHPLC
 SulfachloropyridazineHPLC
 SulfadiazineNo referencesHPLC
 SulfadimethoxineHPLC
 SulfamerazineNo referencesHPLC
 SulfamethazineHPLC
 SulfamethizoleHPLC
 SulfanilamideHPLC, IC
 SulfapyridineNo referencesHPLC
 SulfaquinoxzlineNo referencesHPLC
 SulfathiazoleHPLC
 TetracyclineHPLC
 ThiamphenicolNo referencesHPLC
TigecyclineHPLC
 TobramycinHPLC, IC
 TrimethoprimHPLC, IC
 TylosinHPLC