Search Thermo Fisher Scientific
A drug product container-closure system should not release chemicals that can accumulate in the drug product in quantities sufficient to present a risk of toxicity, or affect its stability or efficacy.
We are unique in offering comprehensive analytical workflows including ICP-MS, GC-MS, LC-MS and advanced information to enable identification and quantification of unknown organic and elemental impurities.
In May 1999, the U.S. FDA published a guidance document on Container Closure Systems for Packaging Human Drugs and Biologics. The FDA mandated that pharmaceutical manufacturers demonstrate the safety of materials used in production systems, container closure systems or drug delivery devices. Failure to demonstrate material safety could result in failure to receive FDA approval for a product. Everything from glass and plastic bottles to foil pouches and the ink used in labels and packaging materials has the potential to leach unwanted contaminants into a drug product.
In order to study the risk of materials extraction studies are performed; generally using aggressive solvent conditions including acidic, basic, organic and aqueous solvents and sohxlet or accelerated solvent extraction. It is important that the extraction process does not deform the material of interest. Compounds present in the resulting extracts are called "extractables". Extractables studies are typically performed to create a "worst case scenario" and assist in materials selection and early risk assessment.
Leachables are chemical species that make their way into the product under normal product, application or storage conditions. There is generally an overlap such that the leachables involved may be classified as a subset of the extractables. Forced or accelerated leachable studies can be performed to assess leachables that migrate under simulated environmental conditions by analyzing the drug formulation after exposure to elevated temperature. However, if the leachable interacts with the drug product or packaging materials, new components can be present. These are termed secondary leachables. Drug formulations often contain buffers, surfactants, fillers and other excipients. This complexity means that there are sometimes secondary leachables that can only be identified after long stability studies over the typical drug shelf life.
Extractable and leachable studies identify key components using a range of analytical technologies:
Quantification of these compounds is performed using relative reference standards, in which a qualified toxicologist calculates the analytical evaluation threshold (AET), above which an extractable or leachable needs to be reported for toxicological assessment. Analytical data for these impurity classes is meant to provide information to help customers determine what further studies are needed to mitigate risks associated with the final product.
There are currently no single regulated method prescribed for analyzing extractables and leachables. Generally, the Product Quality Research Institute (PQRI) recommendation document in the table below is accepted by the industry as a primary recommendation to follow as it was drafted with U.S. FDA involvement. However, to date it has not been formally accepted. There are several methods in draft currently being considered by global regulators and guidance documents from industry bodies, such as BioPhorum Operations Group (BPOG) and BioProcess Systems Alliance (BPSA):
Author | Focus | Title | Year |
---|---|---|---|
PQRI | OINDP Drugs | Best Practices for Extractables and Leachables in Orally Inhaled and Nasal Drug Products: An Overview of the PQRI Recommendations | 2008 |
Join industry expert Dr. Jason Creasy from GlaxoSmithKline as he talks with The Medicine Maker magazine about the history, developments and future of extractable impurities testing for pharmaceuticals.
Join this 3-part webinar series to learn extractables and leachables testing for pharmaceutical container-closure systems and food contact materials. Hear from experts about regulations and the latest technologies and workflows to deliver E&L testing.
The National Institute for Bioprocessing Research and Training (NIBRT) is publishing new research investigating extractable & leachable impurities in the manufacture of biopharmaceuticals. Get their groundbreaking research report first.
Get this interesting and informative editorial from The Medicine Maker magazine, which talks with experts in extractables and leachables testing.
Learn how the most advanced GC-MS technology available can be used to give absolute confidence in unknown identification and quantification of extractable and leachable impurities.
Join applications scientist Keeley Murphy as he presents this extractables and leachables poster at the American Association of Pharmaceutical Scientists (AAPS) conference.
In this study, comprehensive extractables analysis of USP class VI O-rings was conducted using LC-MS and ICP-MS. Organic compounds were identified using mzCloud high-resolution MS/MS libraries.
E&L challenges & Thermo Scientifics differentiated solutions to increase sample throughput, streamline workflows, and increase confidence in results.
E&L analysis overview of differentiated solutions to increase sample throughput, streamline workflows, and increase confidence in results.
Global pharmaceutical CRO chooses Thermo Scientific Dionex ASE 350 Accelerated Solvent Extractor for automated, compliant sample preparation for the study of extractable impurities from drug packaging and production systems
A range of impurities are made together with any medicine. Learn how and why a global CRO chose Thermo Scientific's Q Exactive Plus and Compound Discover to identify and profile extractable and leachable impurities.
Elemental analysis and trace metals testing are important for the commercial release of pharmaceutical products, stability studies, extractable and leachable studies, and clinical trial support.
An anonymized video showcasing Thermo Fisher's Exactive GC instruments conducting impurity analysis.
Keeley Murphy describes a comprehensive extractables analysis of USP class VI O-rings using the Q Exactive Plus MS, iCAP Q ICP-MS for trace element analysis, and the software packages SIEVE, Mass Frontier, and mzCloud spectral databases.
Kyle D'Silva, Pharma & Biopharma Vertical Marketing Manger, gives a preview of his AAPS 2015 seminar, No More Unknowns - A Complete Analytical Workflow for Extractable and Leachable Impurities.
Kyle D'Silva's AAPS presentation that covered impurity analysis (extractables and leachables).
Hear about the latest workflows for analysis of elemental and organic leachable container-closure system pharmaceutical impurities.
Extractables and leachable compounds are a complex and ever growing list of pharmaceutical impurities. Stephen Guilfoyle discusses developments in the regulatory landscape and a broad portfolio of tools to meet these challenges.
Jason Creasey (Glaxo Smith Kline) discusses the history of extractable and leachable testing and how the industry has evolved due to growing knowledge of contact-closure materials provided as new analytical tools became available.
Kyle D’Silva discusses new developments in GC-MS technology for Extractables and Leachables testing, including the new Q Exactive™ GC hybrid quadrupole-Orbitrap GC-MS/MS and mzCloud.org online mass spectral library.
Andrew Feilden (Smithers Rapra) discusses how the E&L Conference is advancing the knowledge of container closure systems for the pharmaceutical industry.
Kate Comstock discusses her long experience with extractables testing in pharmaceutical companies, and how the discipline has evolved through the latest technological developments in mass spectrometry.
Recent webinar discussing the screening of targeted and non-targeted E&L compounds
Get insight into extractable/leachable study designs following regulatory guidelines for both elemental and organic impurities.
Recent webinar discussing the basics of Extractables and Leachables with respect to Pharmaceutical QA.
Learn the latest instrument technology for unknown impurity identification and follow a leachable study for both GC and LC amenable organic impurities.