extractables and leachables
Advanced techniques for extractables testing
Extractable and leachable (E&L) testing methods, such as draft pharmacopeial chapters USP 1663 and USP 1664, demand carefully controlled extraction. Traditionally, routine extractables testing has been performed using soxhlet and reflux extraction, which have lengthy extraction times (over 24 hours), variability in manual handling, and require disposal of potentially flammable and hazardous organic solvents.
Accelerated solvent extraction is a proven technology with significant improvements over traditional extraction techniques. You can use less bench space, extract up to 10 times faster, and use less solvent.
Contact us
Extractables extraction workflow
extractables workflow
Once extracted the samples follow one of four analytical workflows:
Featured extractables and leachables categories
Accelerated extraction for fast material selection studies
Automate extraction, in-cell cleanup and evaporation of compounds from solid and semi-solid samples using the Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor. The EXTREVA ASE system can perform four parallel sample extractions, substantially increasing throughput and efficiency. Because the entire sample preparation workflow is automated in one seamless operation, no manual transfer of samples between steps is required. Get reproducible and efficient parallel extractions that cut manual sample preparation steps from hours to minutes, increase productivity, minimize errors, and dramatically reduce solvent use to meet laboratory sustainability and cost goals.
The Thermo Scientific Orbitrap IQ-X Tribrid Mass Spectrometer combines industry-leading mass analyzer technology dedicated for small molecule analysis with the Thermo Scientific AcquireX intelligent MSn data acquisition strategy, to collect more meaningful data.
Featured videos
Extractables and Leachables: Challenges and Solutions
E&L challenges & Thermo Scientifics differentiated solutions to increase sample throughput, streamline workflows, and increase confidence in results.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextExtractables and Leachables: Challenges and Solutions
          E&L challenges & Thermo Scientifics differentiated solutions to increase sample throughput, streamline workflows, and increase confidence in results.
        2. Now Playing
          Up NextExtractables & Leachables: Overview
          E&L analysis overview of differentiated solutions to increase sample throughput, streamline workflows, and increase confidence in results.
        3. Now Playing
          Up NextExtractable & Leachable CRO ASE 350 Testimonial
          Global pharmaceutical CRO chooses Thermo Scientific Dionex ASE 350 Accelerated Solvent Extractor for automated, compliant sample preparation for the study of extractable impurities from drug packaging and production systems
        4. Now Playing
          Up NextGlobal pharmaceutical CRO chooses Thermo Scientific Q Exactive Plus and Compund Discover for identification of Extractable & Leachable impurities
          A range of impurities are made together with any medicine. Learn how and why a global CRO chose Thermo Scientific's Q Exactive Plus and Compound Discover to identify and profile extractable and leachable impurities.
        5. Now Playing
          Up NextGlobal pharmaceutical CRO chooses iCAP RQ & Qtegra ISDS software for identification and quantification of elemental Extractable & Leachable impurities
          Elemental analysis and trace metals testing are important for the commercial release of pharmaceutical products, stability studies, extractable and leachable studies, and clinical trial support.
        6. Now Playing
          Up NextExactive GC Testimonial Video with Leading GMP Testing Service Provider
          An anonymized video showcasing Thermo Fisher's Exactive GC instruments conducting impurity analysis.
        7. Now Playing
          Up NextComprehensive Extractables Analysis of Medical Grade O-Ring
          Keeley Murphy describes a comprehensive extractables analysis of USP class VI O-rings using the Q Exactive Plus MS, iCAP Q ICP-MS for trace element analysis, and the software packages SIEVE, Mass Frontier, and mzCloud spectral databases.
        8. Now Playing
          Up NextNo More Unknowns – Workflows for Extractable and Leachable Impurities
          Kyle D'Silva, Pharma & Biopharma Vertical Marketing Manger, gives a preview of his AAPS 2015 seminar, No More Unknowns - A Complete Analytical Workflow for Extractable and Leachable Impurities.
        9. Now Playing
          Up NextAAPS 2015 - Extractable and Leachable Impurity Analysis
          Kyle D'Silva's AAPS presentation that covered impurity analysis (extractables and leachables).
        10. Now Playing
          Up NextNo More Unknowns – Analytical Workflow for Extractable and Leachable Impurities
          Hear about the latest workflows for analysis of elemental and organic leachable container-closure system pharmaceutical impurities.
        11. Now Playing
          Up NextSolving the Challenges of Leachable Pharmaceutical Impurities Using a Full Analytical Portfolio
          Extractables and leachable compounds are a complex and ever growing list of pharmaceutical impurities. Stephen Guilfoyle discusses developments in the regulatory landscape and a broad portfolio of tools to meet these challenges.
        12. Now Playing
          Up NextTalking Extractables and Leachables – Glaxo Smith Kline
          Jason Creasey (Glaxo Smith Kline) discusses the history of extractable and leachable testing and how the industry has evolved due to growing knowledge of contact-closure materials provided as new analytical tools became available.
        13. Now Playing
          Up NextTalking Extractables and Leachables – No More Unknowns?
          Kyle D’Silva discusses new developments in GC-MS technology for Extractables and Leachables testing, including the new Q Exactive™ GC hybrid quadrupole-Orbitrap GC-MS/MS and mzCloud.org online mass spectral library.
        14. Now Playing
          Up NextTalking Extractables and Leachables – Smithers Rapra
          Andrew Feilden (Smithers Rapra) discusses how the E&L Conference is advancing the knowledge of container closure systems for the pharmaceutical industry.
        15. Now Playing
          Up NextTalking Extractables and Leachables – Testing Using Orbitrap-based Mass Spectrometry
          Kate Comstock discusses her long experience with extractables testing in pharmaceutical companies, and how the discipline has evolved through the latest technological developments in mass spectrometry.
        16. Now Playing
          Up NextExtractable & Leachable Compounds: Combining Targeted & Non-targeted Screening to Improve Unique Identifications
          Recent webinar discussing the screening of targeted and non-targeted E&L compounds
        17. Now Playing
          Up NextTesting for extractables & leachables in pharmaceutical contact closure materials - A Complete ICP-MS, GC-MS & LC-MS Workflow
          Get insight into extractable/leachable study designs following regulatory guidelines for both elemental and organic impurities.
        18. Now Playing
          Up NextUnderstanding the Challenges of E&L Contaminant Analysis for Pharmaceutical Quality Assurance
          Recent webinar discussing the basics of Extractables and Leachables with respect to Pharmaceutical QA.
        19. Now Playing
          Up NextUsing GC & LC Orbitrap mass spectrometry to confidently identify leached packaging and process impurities
          Learn the latest instrument technology for unknown impurity identification and follow a leachable study for both GC and LC amenable organic impurities.
      Enhance your capabilities
      Unknown impurity identification and quantification workflows for:
      • Pharmaceutical packaging and contact-closure materials
      • Medical devices
      • Food contact materials
      Resources