Counter Ion Analysis
Effectively assay pharmaceutical anions and cations
Accelerate the determination and quantification of inorganic anions and cations in pharmaceutical formulations. Approximately half of all drugs are formulated and administered as salts. A broad selection of inorganic and organic ions can be used as drug substance counterions, with different ions being used to alter the biological and physicochemical properties such as solubility, stability, and dissolution rate. Screening and assaying of a broad variety of counterions and active pharmaceutical ingredients (APIs) is crucial for quality control (QC) of pharmaceutical salts to confirm identity of salt form and mass balance of the API.
Contact us
Featured counterion analysis categories
Versatile UHPLC for counterion determination
Detect and quantify counterions with increased separation speed and resolution using the Thermo Scientific Vanquish UHPLC system with near-universal detection capabilities of the Vanquish Charged Aerosol Detector (CAD). With enhanced linear dynamic range and accurate, consistent response, this detector can measure low-level impurities, counterions and APIs simultaneously, regardless of chemical structure. One can measure virtually any non-volatile and most semi-volatile analytes with sub-nanogram sensitivity. The charged aerosol detector expands the capabilities for discovery and is a powerful addition to any chromatography lab.
Ultimate performance and flexibility
The Dionex ICS-6000 Hybrid HPIC System is a top-of-the-line ion chromatography (IC) system capable of addressing a full range of counterion assays. This high-performance Reagent-Free IC (RFIC) system can operate at pressures up to 5000 psi. This high-pressure capability gives you the flexibility to use small-particle columns in standard lengths for higher resolution without sacrificing speed, or to use shorter (150mm) columns for higher flow rates and high throughput, to meet the demands of the pharmaceutical laboratory.
An ion chromatograph worth getting to know
Designed for those running the same method for an extended time, the Thermo Scientific Dionex Integrion HPIC System has the reliability you need at the lowest ongoing cost. Equipped with whole system monitoring and a simplified plumbing layout, even those who are new to IC can quickly become seasoned users. You'll also save time and improve reproducibility with precise, automatic eluent generation and sample preparation.
Compliant and intuitive chromatography data system
Thermo Scientific Chromeleon 7.3 CDS software unifies workflows for chromatography and routine quantitative analysis, providing full integration of Thermo Scientific instruments in a 21 CFR Part 11 compliant GMP or GLP enterprise environment. Quickly and easily process and report your counterion assay data in one application.