Gibco Collagen I from bovine and rat
Collagen is the most widely used extracellular matrix (ECM) protein for cell culture, facilitating cell attachment, growth, differentiation, migration, and tissue morphogenesis.
Why is collagen used in cell culture?
Collagen I is used in both 2D and 3D cell culture to make the in vitro cellular environment more in vivo-like. A thin coating of diluted collagen solution supports cell attachment and growth of some primary cells, including hepatocytes. When collagen solution is neutralized, it forms a gel that can support 3D cell cultures. 
Collagen for cell culture in convenient liquid form
We offer Collagen I, rat tail (3 mg/ml) and Collagen I, bovine (5 mg/mL), providing the flexibility to use for both coating and gel-forming applications.
Figure 1 shows Gibco liquid collagen for cell culture and collagen from two other suppliers. These were prepared at the concentrations stated and allowed to solidify in individual wells of a 24-well plate. Gibco Collagen I forms a firm, clearer gel matrix.
Figure 1. Gibco Collagen I compared to collagen from other suppliers. Gibco liquid collagen for cell culture, Supplier A collagen, and Supplier B collagen were plated into a 24-well plate at 3 mg and 6 mg concentrations. After the collagen solidified, Gibco Collagen I (A) appears more solid and transparent versus other suppliers.
Research applications for collagen I
Collagen consists of three α-chains, which form a triple helix, providing tensile strength for the ECM. Collagen I is the most common fibrillar collagen for cell culture and is found in skin, bone, tendons, and other connective tissues.
Collagen I plays key roles in cell biology and biomedical applications: 
  • Promotes attachment, proliferation, and differentiation in breast cancer cells [1]
  • Forms gland-like circular structures from embryonic stem cells [2]
  • Allows better representation of endometrial physiology and morphology in vitro, allowing for in vivo studies of endometrial cancer cell invasion [3]
  • Helps human microvascular endothelial cells (HMVECs) adopt an in vitro spindle-shaped morphology and form solid cord-like assemblies [4]
  • Used for in vitro angiogenesis assays, as a barrier in cell invasion assays, and to enable cell adhesion studies [5–6]
Collagen for cell culture ordering information
Catalog # Name Size Price (USD) Qty
A1048301 Collagen I, rat tail Each
337.00
A1142801 Collagen I, Coated Plate, 6 well Each
128.65
Online Exclusive
140.00
Save 11.35 (8%)
A1142802 Collagen I, Coated Plate, 24 well Each
128.65
Online Exclusive
145.00
Save 16.35 (11%)
A1142803 96-well, Collagen Type I-Coated, Flat-Bottom Microplates Each
150.00
References: