Product box and its contents: 21 tubes of One Shot Mach1 T1R Chemically Competent E. coli, 1 tube of pUC19 DNA, and one tube of SOC medium
DH5α competent E. coli is one of the most common laboratory strains. We offer several versatile strains for general cloning applications within a variety of transformation efficiencies and formats to suit your workflow and budget.
Why is DH5α good for cloning?
DH5α is a popular E. coli strain for routine cloning due to several key features:
  • Blue/white screening with lacZ∆M15
  • High insert stability due to recA1 mutation
  • High yield and quality of DNA due to endA mutation
  • High transformation efficiency (Figure 1)
Figure 1. Comparing transformation efficiencies of DH5α competent E. coli with NEB 5-alpha 5α and Zymo 5a in similar product configurations. Transformation efficiency was measured by performing triplicate transformations, using 10 pg pUC19 DNA, according to manufacturer’s recommended protocol. Each transformation was plated in duplicate.
E. coli DH5α competent cell strains
DH5α competent cells for routine cloning
DH5α is one of the most common laboratory E. coli strains utilized for small plasmid propagation and is selected for its ability to achieve high transformation efficiency. Usually, this strain is transformed using the calcium chloride (CaCl2) method; specially designed strain derivatives like DH5α-E can be transformed using electroporation.
 Subcloning Efficiency™ DH5α Competent CellsLibrary Efficiency DH5α Competent CellsMAX Efficiency DH5α Competent Cells
Product box and its contents: 4 tubes of Subcloning Efficiency DH5α Competent Cells and 1 vial of pUC19 DNA
Product box and its contents: 5 tubes of Library Efficiency DH5α Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Product box and its contents: 5 tubes of Max Efficiency DH5α Competent Cells, 1 vial of pUC19 DNA, 2 tubes of SOC medium
Best for
  • General, everyday use
  • Cloning difficult constructs
  • Applications using limiting amounts of DNA
  • Increase insert stability
  • Improve the quality of plasmid DNA prepared from minipreps
Type of transformationChemicalChemicalChemical
Transformation efficiency
>1 x 106 cfu/µg>1 x 108 cfu/µg>1 x 109 cfu/µg
Available formats
4 x 500 µL/tube
5 x 200 µL/tube
5 x 200 µL/tube
Check out other strains for routine cloning
E. coli DH5α-T1R competent cell strains
DH5α-T1R cells have the benefit of the tonA (fhuA) genotype for T1, T5, and φ80 phage resistance, with the same genetic markers and growth properties as the original strain.
 MAX Efficiency DH5α-T1R Competent Cells MultiShot DH5α T1R Competent Cells
Product box and its contents: 21 tubes of One Shot MAX Efficiency DH5α-T1R Competent Cells, 1 vial of pUC19 DNA, 1 bottle of SOC medium
Product box and its contents: 1 96-well plate of MultiShot StripWell DH5α T1R Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Best for
  • Library construction
  • Cloning small amount of DNA
  • High throughput applications
Type of transformationChemicalChemical
Transformation efficiency
>1 × 109 cfu/µg>1 x 108 cfu/µg
Available formats
DH5α competent cells for specialized cloning applications
 MAX Efficiency DH5αF´IQ Competent CellsElectroMAX DH5α-E Competent Cells
Product box and its contents: 5 tubes of MAX Efficiency DH5αF'IQ Competent Cells, 2 tubes of F'IQ Lawn Cell Assay, 1 vial of M13mp18 RF DNA, and 2 tubes of SOC medium
Product box and its contents. 5 tubes of ElectroMAX DH5α-E Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Best for
  • M13 cloning
  • Protein expression in non-BL21 strain
  • Generation of cDNA libraries
  • Transformations with limited input DNA
Type of transformationChemicalElectroporation
Transformation efficiency
>3 x 108 cfu/µg>1 x 1010 cfu/µg
Available formats
5 x 200 µL/tube
5 x 100 µL/tube
Shop all Invitrogen DH5α competent cells
DH5α competent cells for routine cloning
DH5α is one of the most common laboratory E. coli strains utilized for small plasmid propagation and is selected for its ability to achieve high transformation efficiency. Usually, this strain is transformed using the calcium chloride (CaCl2) method; specially designed strain derivatives like DH5α-E can be transformed using electroporation.
 Subcloning Efficiency™ DH5α Competent CellsLibrary Efficiency DH5α Competent CellsMAX Efficiency DH5α Competent Cells
Product box and its contents: 4 tubes of Subcloning Efficiency DH5α Competent Cells and 1 vial of pUC19 DNA
Product box and its contents: 5 tubes of Library Efficiency DH5α Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Product box and its contents: 5 tubes of Max Efficiency DH5α Competent Cells, 1 vial of pUC19 DNA, 2 tubes of SOC medium
Best for
  • General, everyday use
  • Cloning difficult constructs
  • Applications using limiting amounts of DNA
  • Increase insert stability
  • Improve the quality of plasmid DNA prepared from minipreps
Type of transformationChemicalChemicalChemical
Transformation efficiency
>1 x 106 cfu/µg>1 x 108 cfu/µg>1 x 109 cfu/µg
Available formats
4 x 500 µL/tube
5 x 200 µL/tube
5 x 200 µL/tube
Check out other strains for routine cloning
E. coli DH5α-T1R competent cell strains
DH5α-T1R cells have the benefit of the tonA (fhuA) genotype for T1, T5, and φ80 phage resistance, with the same genetic markers and growth properties as the original strain.
 MAX Efficiency DH5α-T1R Competent Cells MultiShot DH5α T1R Competent Cells
Product box and its contents: 21 tubes of One Shot MAX Efficiency DH5α-T1R Competent Cells, 1 vial of pUC19 DNA, 1 bottle of SOC medium
Product box and its contents: 1 96-well plate of MultiShot StripWell DH5α T1R Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Best for
  • Library construction
  • Cloning small amount of DNA
  • High throughput applications
Type of transformationChemicalChemical
Transformation efficiency
>1 × 109 cfu/µg>1 x 108 cfu/µg
Available formats
DH5α competent cells for specialized cloning applications
 MAX Efficiency DH5αF´IQ Competent CellsElectroMAX DH5α-E Competent Cells
Product box and its contents: 5 tubes of MAX Efficiency DH5αF'IQ Competent Cells, 2 tubes of F'IQ Lawn Cell Assay, 1 vial of M13mp18 RF DNA, and 2 tubes of SOC medium
Product box and its contents. 5 tubes of ElectroMAX DH5α-E Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Best for
  • M13 cloning
  • Protein expression in non-BL21 strain
  • Generation of cDNA libraries
  • Transformations with limited input DNA
Type of transformationChemicalElectroporation
Transformation efficiency
>3 x 108 cfu/µg>1 x 1010 cfu/µg
Available formats
5 x 200 µL/tube
5 x 100 µL/tube
Shop all Invitrogen DH5α competent cells
Competent cells: Featured products
In addition to chemically competent cells and electrocompetent cells, we offer a broad suite of products for the cloning workflow.
Microbial
media
Explore types of bacterial growth media to use for cloning, plasmid DNA preparation, and protein expression.
Browse media
Electroporation
cuvettes
Learn more about universal-fit electroporation cuvettes for use with competent cells.
View electroporation cuvettes
TOPO PCR
cloning kits
Learn more about one-step PCR cloning that combines traditional restriction and ligation cloning.
View TOPO kits
Gibson Assembly
cloning kits
Gibson Assembly cloning kits provide highly efficient, seamless cloning, enabling the assembly of multiple DNA fragments of varying lengths into any vector.
Explore Gibson Assembly cloning
Resources
Have any questions on competent cells or transformation? Click on the resources listed below to access overviews, videos, genotype guides, and educational resources.
Icon with monitor with speech bubble onscreen.
Stay up-to-date on industry trends in molecular cloning with on-demand webinars.
Icon with two sheets of paper
Short articles and digital handbook for cloning applications
Icon of mouse inside a circle
Get support for your cloning experiments
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments