What are electrocompetent cells?
Electroporation-competent cells are engineered bacteria that have highly permeable cell membranes for efficient uptake of foreign DNA. Electroporation involves application of electrical pulses across the bacterial membrane, which creates pores and allows genetic material to permeate the membrane. We offer a variety of Invitrogen electrocompetent E. coli cells to reliably clone your target DNA with high efficiency. These competent cells come in convenient packaging options to fulfill your needs for transforming bacteria by electroporation. Standard cuvettes for electroporation can be acquired separately.
Learn about the differences between chemically competent and electrocompetent cells
Electrocompetent E. coli: Selection guide
DH10B electrocompetent cells
DH10B strains are suitable for highest transformation efficiency. Competent cells are prepared using proprietary methods to deliver best-in-class performance. 
 ElectroMAX DH10B Cells ElectroMAX DH10B T1 Phage-Resistant Competent Cells MegaX DH10B T1R Electrocomp™ Cells
Product box and its contents: 5 tubes of ElectroMAX DH10B Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Product box and its contents: 5 tubes of ElectroMAX DH10B T1 Phage-Resistant Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Product box and its contents: 5 tubes of MegaX DH10B T1R Electrocomp Cells, 1 vial of pUC19 DNA, and 3 bottles of recovery media
Best for
  • High transformation efficiency
  • Library construction and other challenging applications
  • High transformation efficiency
  • Library construction and other challenging applications
  • T1, T5, and φ80 phage resistance
  • Highest efficiency
  • Highly demanding cloning
  • Library construction applications
  • T1, T5, and φ80 phage resistance
Type of transformationElectroporationElectroporationElectroporation
Transformation efficiency >1 x 1010 cfu/µg>1 x 1010 cfu/µg>3 x 1010 cfu/µg
Available formats
5 x 100 µL/tube
5 x 100 µL/tube
5 x 100 µL/tube
TOP10 electrocompetent cells
The TOP10 strains are genetically similar to the DH10B strain and are suitable for general cloning applications.
 TOP10 Electrocomp™ Kit One Shot TOP10 Electrocomp™ E. coliTOP10F´ Electrocomp™ Kit
Product box and content: 5 tubes of TOP10 Electrocomp E. coli, 1 vial of pUC19 vector, and 1 bottle of SOC medium
Product box and content: 11 tubes of One Shot TOP10 Electrocomp E. coli, 1 vial of pUC19 DNA, and 1 bottle of SOC medium
Product box and content: 10 tubes of TOP10F' Electrocomp E. coli, 1 vial of pUC19 plasmid, and 2 bottles of SOC medium
Best for
  • High transformation efficiency required (e.g., small DNA amount)
  • High transformation efficiency required (e.g., small DNA amount)
  • Single-use format
  • Carries F´ episome for ssDNA isolation
Type of transformationElectroporationElectroporationElectroporation
Transformation efficiency
>1 x 1010 cfu/µg>1 x 1010 cfu/µg>1 x 1010 cfu/µg
Available formats
Competent cells for specialty applications
 ElectroMAX DH5α-E Competent CellsElectroMAX DH12S Cells ElectroMAX Stbl4 Competent Cells
Photo of product box and its contents: 5 tubes of ElectroMAX DH5α-E Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Photo of product box and its contents: 5 tubes of ElectroMAX DH12S Cells, 1 vial of pUC19 DNA, and 2 tubes of M13KO7 Helper Phage
Photo of product box and contents: 5 tubes of ElectroMAX Stbl4 Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Best for
  • Generation of cDNA libraries
  • Transformations with limited input DNA
  • Production of exceptionally clean ssDNA using M13KO7 helper phage (supplied)
  • Genomic library construction
  • Cloning unstable DNA: stabilizes direct repeat and retroviral sequences
  • Cloning of methylated genomic DNA
Type of transformation
Electroporation
Electroporation
Electroporation
Transformation efficiency
>1 x 1010 cfu/µg>1 x 1010 cfu/µg>5 x 109 cfu/µg
Available formats
5 x 100 µL/tube
5 x 100 µL/tube
5 x 100 µL/tube
DH10B electrocompetent cells
DH10B strains are suitable for highest transformation efficiency. Competent cells are prepared using proprietary methods to deliver best-in-class performance. 
 ElectroMAX DH10B Cells ElectroMAX DH10B T1 Phage-Resistant Competent Cells MegaX DH10B T1R Electrocomp™ Cells
Product box and its contents: 5 tubes of ElectroMAX DH10B Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Product box and its contents: 5 tubes of ElectroMAX DH10B T1 Phage-Resistant Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Product box and its contents: 5 tubes of MegaX DH10B T1R Electrocomp Cells, 1 vial of pUC19 DNA, and 3 bottles of recovery media
Best for
  • High transformation efficiency
  • Library construction and other challenging applications
  • High transformation efficiency
  • Library construction and other challenging applications
  • T1, T5, and φ80 phage resistance
  • Highest efficiency
  • Highly demanding cloning
  • Library construction applications
  • T1, T5, and φ80 phage resistance
Type of transformationElectroporationElectroporationElectroporation
Transformation efficiency >1 x 1010 cfu/µg>1 x 1010 cfu/µg>3 x 1010 cfu/µg
Available formats
5 x 100 µL/tube
5 x 100 µL/tube
5 x 100 µL/tube
TOP10 electrocompetent cells
The TOP10 strains are genetically similar to the DH10B strain and are suitable for general cloning applications.
 TOP10 Electrocomp™ Kit One Shot TOP10 Electrocomp™ E. coliTOP10F´ Electrocomp™ Kit
Product box and content: 5 tubes of TOP10 Electrocomp E. coli, 1 vial of pUC19 vector, and 1 bottle of SOC medium
Product box and content: 11 tubes of One Shot TOP10 Electrocomp E. coli, 1 vial of pUC19 DNA, and 1 bottle of SOC medium
Product box and content: 10 tubes of TOP10F' Electrocomp E. coli, 1 vial of pUC19 plasmid, and 2 bottles of SOC medium
Best for
  • High transformation efficiency required (e.g., small DNA amount)
  • High transformation efficiency required (e.g., small DNA amount)
  • Single-use format
  • Carries F´ episome for ssDNA isolation
Type of transformationElectroporationElectroporationElectroporation
Transformation efficiency
>1 x 1010 cfu/µg>1 x 1010 cfu/µg>1 x 1010 cfu/µg
Available formats
Competent cells for specialty applications
 ElectroMAX DH5α-E Competent CellsElectroMAX DH12S Cells ElectroMAX Stbl4 Competent Cells
Photo of product box and its contents: 5 tubes of ElectroMAX DH5α-E Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Photo of product box and its contents: 5 tubes of ElectroMAX DH12S Cells, 1 vial of pUC19 DNA, and 2 tubes of M13KO7 Helper Phage
Photo of product box and contents: 5 tubes of ElectroMAX Stbl4 Competent Cells, 1 vial of pUC19 DNA, and 2 bottles of SOC medium
Best for
  • Generation of cDNA libraries
  • Transformations with limited input DNA
  • Production of exceptionally clean ssDNA using M13KO7 helper phage (supplied)
  • Genomic library construction
  • Cloning unstable DNA: stabilizes direct repeat and retroviral sequences
  • Cloning of methylated genomic DNA
Type of transformation
Electroporation
Electroporation
Electroporation
Transformation efficiency
>1 x 1010 cfu/µg>1 x 1010 cfu/µg>5 x 109 cfu/µg
Available formats
5 x 100 µL/tube
5 x 100 µL/tube
5 x 100 µL/tube
Advantages of highest efficiency electrocompetent cells
MegaX DH10B T1R Electrocompetent Cells are the highest efficiency electrocompetent cells available (Figure 1). The advantages of MegaX DH10 T1R electrocompetent cells include:
  • Highest transformation efficiency achieved from proprietary manufacturing procedures (>3 x 1010 cfu/μg of control DNA)
  • Ideal for highly demanding cloning and library construction applications
  • tonA (fhuA) genotype confers resistance to T1, T5, and φ80 lytic phage infection and safeguards clones and libraries
Figure 1. Transformation efficiency comparison of MegaX DH10B T1R Electrocompetent Cells to E. cloni® 10G Supreme Electrocompetent Cells (Lucigen) and Electro Ten-Blue Electroporation-Competent Cells (Agilent) using manufacturer’s recommended protocols. MegaX DH10B T1R consistently outperformed other electrocompetent cells.
Three electroporation cuvettes with red caps
Choose the right electroporation cuvette for your strain.
Learn more about electroporation cuvettes
Competent cells: Featured products
In addition to chemically competent cells and electrocompetent cells, we offer a broad suite of products for the cloning workflow.
Microbial
media
Explore types of bacterial growth media to use for cloning, plasmid DNA preparation, and protein expression.
Browse media
Electroporation
cuvettes
Learn more about universal-fit electroporation cuvettes for use with competent cells.
View electroporation cuvettes
TOPO PCR
cloning kits
Learn more about one-step PCR cloning that combines traditional restriction and ligation cloning.
View TOPO kits
Gibson Assembly
cloning kits
Gibson Assembly cloning kits provide highly efficient, seamless cloning, enabling the assembly of multiple DNA fragments of varying lengths into any vector.
Explore Gibson Assembly cloning
Resources
Have any questions on competent cells or transformation? Click on the resources listed below to access overviews, videos, genotype guides, and educational resources.
Icon with monitor with speech bubble onscreen.
Stay up-to-date on industry trends in molecular cloning with on-demand webinars.
Icon with two sheets of paper
Short articles and digital handbook for cloning applications
Icon of mouse inside a circle
Get support for your cloning experiments
Stylesheet for Classic Wide Template adjustments