Search Thermo Fisher Scientific
Each project has its own unique requirements. To meet these different demands, many of Invitrogen’s competent cells are offered in a variety of packaging formats. Custom formats are also available.
Format / Volume | Chemically competent | Electrocompetent | Throughput | Advantage |
---|---|---|---|---|
One Shot 50 μL | ● | ● | Single-use | Transformation and recovery in the same tube |
MultiShot StripWell 50 μL per well | ● | Medium-throughput | 12 strips of 8 tubes; do as many or as few reactions as needed | |
MultiShot FlexPlate 20 μL per well | ● | High-throughput | 96-well segmented plate: Use multiples of 8-well segments or entire plate | |
MultiShot 15 μL per well | ● | High-throughput | 96-well plates fit automated format | |
Standard 100 µL, 200 µL, or 500 µL | ● | ● | Multi-use for throughput | Economical option |
The unique format provides 50 μL aliquots of chemically competent cells in single-use tubes. Transformation is performed directly in the tube, saving you time and effort. There are no extra pipetting steps, no extra tubes, and no loss of efficiency due to repeated freeze-thaw cycles. These kits come with SOC medium and pUC19 positive control.
The MultiShot StripWell format provides strips of tubes in a 96-tube rack format, each well containing 50 μL of chemically competent cells This allows you to use as many or as few reactions as you want, without wasting reagents. Each kit includes 12 strips of 8 tubes, SOC medium, and pUC19 positive control.
The MultiShot FlexPlate format provides a segmented 96-well plate with 20 μL of chemically competent cells in each well. The plate can be separated into 8-well segments allowing you to choose manual or automated applications. Kits are supplied with one 96-well plate, SOC medium, and pUC19 positive control.
The MultiShot format provides 15 μL aliquots of chemically competent cells in each well of a 96-well plate. Kits are supplied with five 96-well plates, SOC medium, and pUC19 positive control.
Standard kits include competent cells in a bulk format. They are designed for scientists who perform more than one transformation at a time and for general use in a busy lab. Kits are available in 500 μL (5 × 100 μL), 1 mL (5 × 200 μL), and 2 mL (4 × 500 μL) quantities. These kits include SOC medium and pUC19 positive control.
Custom kits with any competent cell type can be packaged in any of the above formats to meet your particular needs. Custom-designed strains can also be made available. Contact your local account manager or send an email to customorders@thermofisher.com to learn more.
Find tips, troubleshooting help, and resources for your
cloning applications
Have any questions on competent cells or transformation? Click below to access overviews, videos, genotype guides and educational resources.
For Research Use Only. Not for use in diagnostic procedures.