Arcturus® PicoPure® Frozen RNA Isolation Kit
The Arcturus® PicoPure® RNA Isolation Kit is designed to recover
high-quality total RNA from fewer than 10 cells―even from a single cell. This kit is designed to provide consistent, efficient RNA recovery without sacrificing quality. Low-volume elution allows you to maximize RNA recovery from small numbers of cells for gene expression analysis.
Key Product Features
  • Greater sensitivity—isolate RNA from even a single cell
  • Superior efficiency—maximize RNA recovery from samples
  • Streamlined concentration—elute with low volumes
  • Flexible applications—compatible with many tube sizes
  • Integrated contamination prevention—column lids help ensure that samples remain RNase-free
Ordering Information
Catalog # Name Size Price (USD) Qty
KIT0214 PicoPure™ RNA Isolation Kit, 200 samples Each
2,510.00
KIT0204 PicoPure™ RNA Isolation Kit, 40 samples Each
578.00
Isolate RNA From Even a Single Cell
The PicoPure® RNA Isolation Kit is engineered to recover high-quality total RNA from fewer than 10 cells. The high recovery rate is important not only for small samples containing picograms of RNA, but also for samples containing up to 100 µg of RNA. Since the RNA elutes in as little as 10 µL of buffer, it is ready for gene expression analysis without any vacuum concentration. This kit is available in two sizes: enough for 40 or 200 isolations (Figure 1).
Isolate RNA from a single cell
 Figure 1. Isolate RNA from a single cell. Quantitative real-time PCR (qRT-PCR) of RNA isolated from 1 to 1,000 laser capture microdissected (LCM) cells. Cultured mouse Leydig (TM3) cells (ATCC) were spun down onto a microscope slide, fixed, and dehydrated. Replicate samples of 1, 10, 100, and 1,000 cells were microdissected using the PixCell® IIe LCM Instrument. RNA was isolated using the PicoPure® RNA Isolation Kit, reverse-transcribed, and subjected to a qRT-PCR assay for GAPDH. Quantifiable message can be detected from a single cell, and the fluorescence signal is proportional to the cell number for all samples studied.
Get Superior RNA Recovery
Small, valuable samples often cannot be replaced. Performing gene expression studies on these precious samples requires getting maximum RNA recovery. Comparative studies show that, with small, RNA-poor cell or tissue samples, the PicoPure® kit dramatically outperforms other commercial total RNA isolation kits in efficiency.
Eliminate Time-Consuming RNA Concentration
The simple isolation protocol consists of extracting cellular RNA, then loading the extract onto the specially designed MiraCol™ Purification Column to bind the RNA. After washing away impurities, the RNA elutes in only 10 µL of buffer, ready for use without vacuum concentration or transfer into a new reaction tube.
High-Yield Purification Columns
The PicoPure® RNA Isolation Kit features unique, high-recovery MiraCol™ Purification Columns, engineered to maximize recovery from microscopic samples. The columns have convenient integrated lids to help ensure that samples remain RNase-free. In addition, the kit includes all necessary buffers, an easy-to-follow user guide, optimized protocols for handling laser capture microdissection (LCM) samples, and protocols for isolating RNA from larger samples (Figure 2).
MiraCol™ Purification Columns
 Figure 2. MiraCol™ Purification Columns
Deliver RNA Ready for Amplification & Microarray Analysis
RNA isolated with the PicoPure® RNA Kit is ready, without vacuum concentration, for amplification with the RiboAmp® RNA Amplification Kit. The two kits together enable highly reproducible gene expression results.
Retain Low-Abundance mRNA & Maintain High RNA Quality
The PicoPure® RNA Isolation Kit preserves mRNA in all abundance classes through the isolation process, minimizing the loss of important genes of interest. Low, medium, and high-abundance genes are recovered and available for expression analysis (Figure 3).
The PicoPure® RNA isolation process allows for high-quality RNA. Total cellular RNA prepared from LCM samples remains intact after isolation, increasing confidence in subsequent molecular analysis (Figure 4).
Superior optics for subcellular microdissection
 Figure 3. RT-PCR of low, medium, and high-abundance genes in RNA isolated using the PicoPure® Kit. RT-PCR was performed on RNA from 500 cells captured from mouse liver and kidney. Equal quantities of cDNA were analyzed with three mouse primer sets: Elongation Factor 1-a (EF-1a, high-abundance gene, ~3,000 copies/cell, 187 bp), Glyceraldehyde-3-phosphate Dehydrogenase (GAPDH, medium-abundance gene, 300–3,000 copies/cell, 357 bp), and Protein Phosphatase 1 (PP1, low-abundance gene, <300 copies/cell, 498 bp). Samples were run on an acrylamide gel. M: molecular weight markers. Lane 1: kidney EF-1a. Lane 2: liver EF-1a. Lane 3: kidney GAPDH. Lane 4: liver GAPDH. Lane 5: kidney PP1. Lane 6: liver PP1. Lane 7: liver EF-1a no RT control. Lane 8: no RT template control.
Figure 4. The PicoPure® Kit maintains RNA integrity. Total RNA from 15,000 mouse kidney cells was isolated from two samples with the PicoPure® RNA Isolation Kit.RNA samples were run using an Agilent Bioanalyzer 2100. Lane 1: Total RNA from non-LCM cells. Lane 2: Total RNA from LCM-captured cells. Comparable 18S and 28S ribosomal RNA band intensities are observed between RNA isolated from both the intact tissue and the LCM sample, demonstrating high quality of RNA obtained.
The Arcturus® PicoPure® RNA Isolation Kit is for research use only, and is not intended for animal or human therapeutic or diagnostic use.