Designed with the performance and features to precisely support your application.
Our portfolio of fit for purpose laboratory and medical freezers facilitates reliable, secure storage with thoughtfully applied features and capabilities to support your specific application needs. From dependable daily, general lab use to the exacting requirements of critical samples and life saving therapeutics, our freezers are precisely configured for performance and reliability.
Proven dependability
DSC0975
Thermo Scientific lab freezers offer measurably dependable performance. Our freezers achieve consistently excellent performance on key metrics like temperature fluctuations in the cabinet over time (peak variability), the difference of those measurements across the cabinet (uniformity) and temperature variations in specific locations inside of the cabinet (stability). We test our freezers using sensors throughout the cabinet under rigorous, real-world testing environments so that you can gain assurance that your samples are consistently, securely and dependably stored.
Figure illustrates temperature performance of a high-performance general purpose 30 cu. ft. freezer  at -30°C with recovery times after an adaptive defrost cycle utilizing 24 open air temperature sensors.
30 cuft Upright freezer at -30c cycle
Fast door opening recovery
Lab freezers door open
Frequent door openings or samples being added to a freezer can create a substantial heat load burden, and having the cooling capacity to quickly recover is critical to performance. Fan-forced air circulation promotes temperature uniformity and faster temperature recovery from out-of-range temperatures. This capability is especially important for busy labs and where biological samples with a low tolerance for temperature fluctuations are stored.
Leading warranty
Thermo Scientific stands behind its freezers, offering comprehensive warranty for up to 2 years on parts and labor, and up to 10 years on our V-Drive compressors.
Quality construction
Every detail of our lab and medical freezers is considered and designed with high quality materials to meet our exacting standards for cold storage performance and long-term durability. This includes scratch-resistant, heavy-gauge, cold-rolled steel interior and exterior with smooth powder coat and epoxy finishes that resist physical and environmental wear.  One-piece seamless polypropylene liner interiors or galveneel options allow for interior wall strength and ease of cleaning.  Closed-cell gaskets facilitate a positive door seal. These seemingly minor elements add up to finely crafted freezer that’s able to meet quality, safety and regulatory standards in accordance with UL, NFPA, AABB, NSF 456 and OSHA, as well as your demands for durability.
World class service and support
Thermo Scientific customers enjoy access to the largest network of lab equipment service professionals on the globe, with fast response times, remote service and problem resolution and a track record of service excellence over 50 years long.  Dependable compliance and preventative maintenance programs are designed to make ownership truly care-free.  If there ever is a problem with your freezer, you can be confident it will be resolved quickly, with minimal downtime.
Environmental sustainability
Our freezers are engineered for environmental sustainability without sacrificing performance.  We address sustainability through a focus on energy efficiency, advancements in cooling technology and the use of environmentally-friendly manufacturing techniques and construction materials.
ACT labeled
Thermo Scientific TSX lab freezers have completed the ACT Environmental Impact Factor Label labeling processes, which is designed to address the need of both scientists and procurement specialists for clear, third-party verified information about the environmental impact of laboratory products. The ACT label process entails third-party verification of the sustainable impacts of a product, its operations, and its end of life. 
Lab freezers ACT Label
ENERGY STAR certified
Our Thermo Scientific environmentally friendly freezers are ENERGY STAR certified to reduce energy usage without compromising performance. With advanced compressor and cooling systems, our high performance freezers consume as little as 4kW-hr/day, a reduction of 55% compared to conventional freezers. Additionally, reduced heat output can provide further energy savings by reducing the HVAC load in your lab or clinical environment.  All TSX Series products are EPA ENERGY STAR certified.
Energy Star certified
SNAP compliant
Thermo Scientific TSX Series high-performance freezers are designed with features that support sustainability objectives in the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), the European Union’s F-Gas compliance, and other standards worldwide. These features include use of non-CFC and HFC-blend natural R290 environmentally friendly refrigerants, and high-density, 2 in. (5.1cm) thick water-blown CFC-free urethane foam insulation.  You can be confident that your Thermo Scientific freezer is a greener choice while maintaining the same confidence in your sample protection.
SNAP
Innovative compressor technology
The Variable Drive (V-Drive) compressor, available on TSX Series freezers, delivers a leap forward in performance and energy efficiency.  Where standard compressors operate in a binary, full-power or off cycle, the innovative V-Drive runs at variable speeds as required to achieve both optimal cooling capacity and energy efficiency.  Combined with an automated tuning control, the V-Drive compressors adjust cooling power to precisely meet the needs required by temperature conditions inside and outside of the freezer.  The result is better temperature uniformity, longer life of the unit and exceptional energy efficiency.
Variable speed technology
User friendly
Pharmacy freezers
Seemingly small details can make big difference when you consider how often and how long you’ll be working with your freezer in healthcare settings and the lab.  With more than 70 years of freezer manufacturing experience and collaboration with our customers, we’ve implemented design and engineering features that set our freezers apart as highly-valued additions to your facility. 
Intuitive controls
Intuitive to operate, and easy to access and read digital displays remove the guess work and help prevent operational mishaps.  The adaptive controller system utilizes mutli-sensor detection to allow precise temperature control to meet current and impending standards from the CDC, WHO, NSF, and other global agencies.
Intuitive controls
Whisper-quiet operation
Thermo Scientific TSX Series high-performance freezers offer whisper-quiet operation to pemit a quiet and comfortable work environment.  These models are up to 10x quieter than previous generations of freezers. Innovative V-Drive technology limits the sound output to as low as 45.5 dBA, comparable to a home refrigerator.
whisper quiet noise
Ergonomic features
Our freezers are designed with the user in mind as well as the samples. Features like vacuum relief ports permit easier door openings with magnetic door gaskets for easy and secure closings.  Ergonomically designed smooth full-length and reversible door handles feature a soft-touch grip.  Comfortable easy-access shelf level placement helps increase user efficiency and lessen physical strain, while reducing the need for floor-level loading of racks. Bright LED interior lighting assists in easy sample access and identification.
Ergonomic features
Optimized, convenient storage design
Our lab freezers can be purpose built for your application.  That includes a well thought out interior design and a range of options for shelving and storage container configurations that are suited to the particular types of samples you store and how often they are accessed.  These small details make a big difference in the usability of your freezer.
Lab freezers purpose-built
Worry-free sample protection
Worry-free sample protection
Factory pre-set and key-operated setpoint security locks in temperature and alarm setpoints to minimize error and help prevent tampering.  Alarm systems with built-in audible/visual warnings for high/low temperature, door ajar, or power failure alert personnel should out-of-range temperature levels occur.  Alarm setting features include ringback, auto-reset, alarm silence, push-button test options, and remote alarm contacts to give you peace of mind. Door locks enable an additional level of security for your precious sample storage.
Connected solutions
Wireless monitoring solutions, included with TSX models and available with all of our cold storage products, facilitate cold chain compliance and sample security through the network or cloud.  Check stored temperature readings anytime, anywhere with your smart phone, receive remote alarms and better manage not only your samples, but your fleet of equipment.
Exacting performance
Thoughtful feature configurations that support the demands of your sample type and lab needs define our extensive portfolio of lab freezers. Our freezers are offered in three general product lines that offer features and technology ranges according to your lab’s needs. 
TSX Series TSG Series
TSV Series
High-performance, advanced freezers for temperature-sensitive critical lab samples and a need for the highest levels of sample security.  
Reliable freezers for busy lab environments needing dependability for semi-critical storage, precise temperature control and excellent operational features. 
Dependable, economical and simple to operate plug-and-play freezers for non-critical samples and basic lab needs.  Great performance with lower cost. 
Performance-oriented refrigeration technology
Exceptional-
V-drive adaptive compressor
Fully adaptive autodefrost
Fully adaptive manual defrost
Precision-
Single speed compressor
Adaptive autodefrost
Reliable-
Single speed compressor
Auto defrost
Manual defrost
Typical applications
Vaccines, enzymes, chemotherapy and pharmaceutical compounds, medical and laboratory-grade products requiring a precisely controlled storage environment.
Reagents, media, proteins, and sample storage with confidence.
Basic biomedical application requirements.
Application-specific configurations
Each of our product lines (TSX, TSG and TSV) include application-specific configurations which include key features and certifications suited to the type of samples you’re working with. These models support an exacting fit to your performance requirements.  
Vaccine storage
Many vaccines require highly specific temperatures to remain effective. Any deviation from these can have a dramatic impact on a vaccine’s viability and efficacy.  Our high-performance, purpose-built vaccine freezers are designed to meet the recommendation and standards from the CDC, WHO, NSF and other global agencies for vaccine storage to maintain cold chain requirements and product quality. Our portfolio includes freezers certified to the ANSI/NSF 456 Vaccine Storage Standard, facilitating the performance and quality you need, in addition to the low sound, low energy and exceptional user experience you deserve.
Plasma freezers
Designed to meet AABB, ANRC, FDA, and EU MDD standards for safety, quality and performance. Our plasma freezers are ideal for demanding blood bank applications where enabling fast heat removal and temperature recovery are key to maintaining product viability.  Our preset, high-performance –30°C plasma freezers with automatic defrost are approved products under the AABB Standards-Compliant Product Program (SCPP) for the storage of plasma.  These freezers include 2” casters for mobility, standard built-in chart recorders and self-closing doors with 90° stop to assist with inventory loads.
Plasma freezers
Flammable materials and explosion-proof freezers
Our flammable material and explosion-proof freezers are engineered to safely store flammable and volatile materials by employing spark-free interiors with no internal electrical components.  They are designed to meet stringent US Pharmacopeia standards for drug storage, NFPA 45 and 99 requirements and OSHA standards. These freezers are available in under-counter, full-height and freestanding models.
Flammable materials and explosion-proof freezers
Manual defrost (-20°C) freezers
Our manual defrost, high-performance -20°C freezers are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated.  Ideal for samples such as enzymes, reagents, pharmaceuticals, biologicals, and other commonly used laboratory materials such as primers, siRNA molecules and fetal bovine serum.  These precision freezers permit cold-wall convection cooling and excellent temperature uniformity.  Our enzyme freezers incorporate specially designed enzyme bins for optimized storage and sample management and are available in variety of capacities for demanding, space challenged environments and are Compatible with GMP Clean Room Class A and ISO 6 (ISO EN 14644-1) standards with appropriate pre-installation
Manual defrost HP -20 freezers
Auto defrost (-30°C) freezers
Reliable, flexible-use workhorses of the lab, our auto defrost -30° freezers use forced air circulation for uniform temperature throughout the cabinet and fast door opening recovery with a non-invasive, adaptive, time and temperature guided defrost cycle that eliminates the need to remove samples during defrost.  These freezers are available in a variety of capacities for samples and applications ranging from routine to complex. 
Auto defrost (-30c) freezers