Perform your dissolved oxygen measurements with confidence
Achieve precise and reliable dissolved oxygen measurement with Orion bench and portable dissolved oxygen meters. Ideal for various industries, including environmental monitoring, wastewater treatment, aquaculture and more, Orion dissolved oxygen meters help you achieve regulatory compliance. Combining advanced technology, user-friendly interfaces, and robust construction, these meters ensure accurate data collection. Suitable for both lab and field use, they measure using polarographic and optical technology, with flexible calibration options. Explore Orion dissolved oxygen meters for precision and reliability tailored to your needs. Browse our complete portfolio or let one of our experts help you make a selection.
Select your dissolved oxygen bench or portable meter
Premium
Advanced
Basic
 Orion Versa Star Pro
Dissolved Oxygen Bench Meter
(Versa Star Pro series)
Orion Pro Star DO213
Dissolved Oxygen Bench Meter
(Pro Star series)
Orion Lab Star DO113
Dissolved Oxygen Bench Meter
(Lab Star series)
 Premium Orion benchtop  dissolved oxygen meters
 Versatile, high-performance laboratory metersEasy to use, dependable
laboratory meters
Economical, simplified
laboratory meters
Ideal for
applications in:
Pharmaceuticals,
biotechnology, industrial
Environmental, water/ wastewater, food & beverage, industrialAcademic,
environmental, power
Compatible probes Optical, polarographicOptical, polarographic Optical, polarographic
DO range (polarographic) 0 to 90 mg/L, 0 to 600% 0.01 to 90.0 mg/L or ppm,
0.0 to 600.0% saturation
0 to 30 mg/L, 0 to 300% 
DO range (RDO optical) 0 to 50 mg/L, 0 to 500% 0.01 to 90.0 mg/L or ppm,
0.0 to 600.0% saturation
DO calibration modes Water-saturated air or air saturated water 100% and 0% saturation, custom concentration value (Winkler)Water-saturated air or air saturated water 100% and 0% saturation, custom concentration value (Winkler)100% and 0% saturation;
custom concentration value
Barometric pressure compensation Automatic with built-in barometer Automatic with built-in barometerAutomatic with manual value input 
Salinity correction Automatic with conductivity module and probe or manual input Automatic with built-in barometer or manual entry   Automatic with manual factor input 
Temperature range-5 to 105°C
23 to 221°F
-30 to 130°C
-22 to 266°F
-5 to 105°C
23 to 221°F
Number of channels4, customizable with modules11
Stirrer probe inputs21
Display5.3-inch, backlit color display7-inch, high-resolution,
backlit color display
5-inch, backlit segmented display
TouchscreenNoYesNo
Onscreen text promptsStep-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Basic calibration and
setup instructions
Identification optionsElectrode ID, sample ID, user IDElectrode ID, sample ID, user ID
Data log2000 with date/time10000 with date/time500 with date/time
Calibration log30 per parameter20 per parameter
Data export optionsComputer, printerComputer, printer, USB flash driveComputer, printer
Power supplyUniversal, 100–240 VAC,
50-60 Hz (Cat. No. STARA-PWR)
Universal, 100-240 VAC,
50-60Hz (Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal, 100-240 VAC,
50-60Hz (Cat. No. LSTAR-PWR),
9 DC adapter, 1.3A
Premium
Advanced
Basic
 Orion Star A323 Dissolved Oxygen
(STARA3230 series)
Orion Star A223 Dissolved Oxygen
(STARA2230 series)
Orion Star A123 Dissolved Oxygen
(STARA1230 series)
 Premium portable Dissolved Oxygen MetersAdvanced portable Dissolved Oxygen MetersBasic portable Dissolved Oxygen Meters
Compatible probesRDO, polarographicRDO, polarographicPolarographic
DO range (polarographic)0 to 600% saturation0 to 600% saturation0 to 200% Saturation
DO range (RDO optical)0 to 50 mg/L0  to 50 mg/L---
DO calibration modesWater-saturated air, air saturated water, manual (Winkler), and zero pointWater-saturated air, air saturated water, manual (Winkler), and zero pointWater-saturated air, manual (Winkler)
Barometric pressure compensation400 to 850 mmHg,
automatic or manual
400 to 850 mmHg,
automatic or manual
---
Salinity Correction0 to 45 ppt0 to 45 ppt0 to 45
Backlit DisplayYesYes---
Number of channels111
Data log5000 with time and date stamp1000 with time and date stamp50
Data export optionsRS232,USBRS232, USBRS232, USB
AC AdapterOptional—universal,
100-240 VAC 
Optional—universal,
100-240 VAC
Optional—universal,
100-240 VAC
Battery powerIncluded—4AA'sIncluded—4AA'sIncluded—4AA's
  Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
Advanced
Basic
 Eutech DO2700 Dissolved Oxygen Bench MeterEutech 1730 Dissolved Oxygen Bench Meter
 Economical, routine laboratory metersBudget-friendly, simple laboratory meters
Compatible probesPolarographicGalvanic
DO range (Polarographic)0.00 to 50.00 mg/L; 0 to 600.0 % saturation---
DO range (Galvanic)---0 to 30 mg/L; 0 to 300.0 % saturation
DO calibration modesWater-saturated air, zero point, manual (Winkler)100% and 0% saturation, manual concentration value (Winkler)  
Barometric pressure compensationAutomatic with built-in barometerAutomatic with manual value entry
Salinity correctionAutomatic with manual value entryAutomatic with manual value entry
Range (temperature)0.0 to 50.0 °C, 32.0 to 122.0 °F-5.0 to 105.0 ºC
Backlit displayYesYes
Number of channels11
Stirrer probe inputsYes---
Data log500 with date/time500 with date/time
Calibration logMost recent viewable on meter and stored in meter memoryActive dissolved oxygen, temperature with time and date
Data export optionsRS232Computer or printe
Power supplyUniversal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401Universal, 100–240VAC, 50–60Hz (Cat. No. EB1700PWR), 9 DC adapter, 1.3A
Video Player is loading.
Current Time 0:00
Duration 1:20
Loaded: 12.16%
Stream Type LIVE
Remaining Time 1:20
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Watch and learn more:
    Features & Benefits of the Eutech 1730 Dissolved Oxygen Bench Meter
    Premium
    Advanced
    Basic
     Eutech DO 450 Portable Dissolved Oxygen Meter
    Eutech DO 600 Portable Dissolved Oxygen Meter
    Eutech DO 6+ Portable Dissolved Oxygen Meter
    Compatible probes RDOPolorographicPolorographic
    DO range (polarographic) --- 0 to 90.00 mg/L or ppm0.00 to 19.99 mg/L or ppm
    DO range (RDO optical) 0 to 200 %; 0 to 20 ppm/mg/L--- --- 
    DO calibration modes Zero and saturation2-point (0 %, 100 %),
    1-point (mg/L)
    2-point (0 %, 100 %),
    1-point (mg/L)
    Barometric pressure compensation Manual450 to 825 mmHg /
    59.9 to 109.9 kPa
    500 to 1499 mmHg /
    66.6 to 199.9 kPa (Manual Input)
    Salinity correction Manual0.0 to 50.0 ppt (Manual Input)0.0 to 50.0 ppt (Manual Input)
    Backlit display  Yes Yes --- 
    Number of channels111
    Data log500 with time and date stamp500 with time and date stamp---
    Data export optionsUSB or RS232IrDA, RS232C (via LED)---
    AC adapter9 V DC adapter, 1.3 A
    (100/240 VAC, SMPS) (optional)
    9 V DC adapter---
    Battery powerTwo AA batteries (included)Included—4AAs4 x AAA batteries
    Premium
    Advanced
    Basic
     Orion Versa Star Pro
    Dissolved Oxygen Bench Meter
    (Versa Star Pro series)
    Orion Pro Star DO213
    Dissolved Oxygen Bench Meter
    (Pro Star series)
    Orion Lab Star DO113
    Dissolved Oxygen Bench Meter
    (Lab Star series)
     Premium Orion benchtop  dissolved oxygen meters
     Versatile, high-performance laboratory metersEasy to use, dependable
    laboratory meters
    Economical, simplified
    laboratory meters
    Ideal for
    applications in:
    Pharmaceuticals,
    biotechnology, industrial
    Environmental, water/ wastewater, food & beverage, industrialAcademic,
    environmental, power
    Compatible probes Optical, polarographicOptical, polarographic Optical, polarographic
    DO range (polarographic) 0 to 90 mg/L, 0 to 600% 0.01 to 90.0 mg/L or ppm,
    0.0 to 600.0% saturation
    0 to 30 mg/L, 0 to 300% 
    DO range (RDO optical) 0 to 50 mg/L, 0 to 500% 0.01 to 90.0 mg/L or ppm,
    0.0 to 600.0% saturation
    DO calibration modes Water-saturated air or air saturated water 100% and 0% saturation, custom concentration value (Winkler)Water-saturated air or air saturated water 100% and 0% saturation, custom concentration value (Winkler)100% and 0% saturation;
    custom concentration value
    Barometric pressure compensation Automatic with built-in barometer Automatic with built-in barometerAutomatic with manual value input 
    Salinity correction Automatic with conductivity module and probe or manual input Automatic with built-in barometer or manual entry   Automatic with manual factor input 
    Temperature range-5 to 105°C
    23 to 221°F
    -30 to 130°C
    -22 to 266°F
    -5 to 105°C
    23 to 221°F
    Number of channels4, customizable with modules11
    Stirrer probe inputs21
    Display5.3-inch, backlit color display7-inch, high-resolution,
    backlit color display
    5-inch, backlit segmented display
    TouchscreenNoYesNo
    Onscreen text promptsStep-by-step calibration and
    guided setup instructions
    Step-by-step calibration and
    guided setup instructions
    Basic calibration and
    setup instructions
    Identification optionsElectrode ID, sample ID, user IDElectrode ID, sample ID, user ID
    Data log2000 with date/time10000 with date/time500 with date/time
    Calibration log30 per parameter20 per parameter
    Data export optionsComputer, printerComputer, printer, USB flash driveComputer, printer
    Power supplyUniversal, 100–240 VAC,
    50-60 Hz (Cat. No. STARA-PWR)
    Universal, 100-240 VAC,
    50-60Hz (Cat. No. PSTAR-PWR),
    9 DC adapter, 2.0A
    Universal, 100-240 VAC,
    50-60Hz (Cat. No. LSTAR-PWR),
    9 DC adapter, 1.3A
    Premium
    Advanced
    Basic
     Orion Star A323 Dissolved Oxygen
    (STARA3230 series)
    Orion Star A223 Dissolved Oxygen
    (STARA2230 series)
    Orion Star A123 Dissolved Oxygen
    (STARA1230 series)
     Premium portable Dissolved Oxygen MetersAdvanced portable Dissolved Oxygen MetersBasic portable Dissolved Oxygen Meters
    Compatible probesRDO, polarographicRDO, polarographicPolarographic
    DO range (polarographic)0 to 600% saturation0 to 600% saturation0 to 200% Saturation
    DO range (RDO optical)0 to 50 mg/L0  to 50 mg/L---
    DO calibration modesWater-saturated air, air saturated water, manual (Winkler), and zero pointWater-saturated air, air saturated water, manual (Winkler), and zero pointWater-saturated air, manual (Winkler)
    Barometric pressure compensation400 to 850 mmHg,
    automatic or manual
    400 to 850 mmHg,
    automatic or manual
    ---
    Salinity Correction0 to 45 ppt0 to 45 ppt0 to 45
    Backlit DisplayYesYes---
    Number of channels111
    Data log5000 with time and date stamp1000 with time and date stamp50
    Data export optionsRS232,USBRS232, USBRS232, USB
    AC AdapterOptional—universal,
    100-240 VAC 
    Optional—universal,
    100-240 VAC
    Optional—universal,
    100-240 VAC
    Battery powerIncluded—4AA'sIncluded—4AA'sIncluded—4AA's
      Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
    Advanced
    Basic
     Eutech DO2700 Dissolved Oxygen Bench MeterEutech 1730 Dissolved Oxygen Bench Meter
     Economical, routine laboratory metersBudget-friendly, simple laboratory meters
    Compatible probesPolarographicGalvanic
    DO range (Polarographic)0.00 to 50.00 mg/L; 0 to 600.0 % saturation---
    DO range (Galvanic)---0 to 30 mg/L; 0 to 300.0 % saturation
    DO calibration modesWater-saturated air, zero point, manual (Winkler)100% and 0% saturation, manual concentration value (Winkler)  
    Barometric pressure compensationAutomatic with built-in barometerAutomatic with manual value entry
    Salinity correctionAutomatic with manual value entryAutomatic with manual value entry
    Range (temperature)0.0 to 50.0 °C, 32.0 to 122.0 °F-5.0 to 105.0 ºC
    Backlit displayYesYes
    Number of channels11
    Stirrer probe inputsYes---
    Data log500 with date/time500 with date/time
    Calibration logMost recent viewable on meter and stored in meter memoryActive dissolved oxygen, temperature with time and date
    Data export optionsRS232Computer or printe
    Power supplyUniversal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401Universal, 100–240VAC, 50–60Hz (Cat. No. EB1700PWR), 9 DC adapter, 1.3A
    Video Player is loading.
    Current Time 0:00
    Duration 1:20
    Loaded: 12.16%
    Stream Type LIVE
    Remaining Time 1:20
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Watch and learn more:
      Features & Benefits of the Eutech 1730 Dissolved Oxygen Bench Meter
      Premium
      Advanced
      Basic
       Eutech DO 450 Portable Dissolved Oxygen Meter
      Eutech DO 600 Portable Dissolved Oxygen Meter
      Eutech DO 6+ Portable Dissolved Oxygen Meter
      Compatible probes RDOPolorographicPolorographic
      DO range (polarographic) --- 0 to 90.00 mg/L or ppm0.00 to 19.99 mg/L or ppm
      DO range (RDO optical) 0 to 200 %; 0 to 20 ppm/mg/L--- --- 
      DO calibration modes Zero and saturation2-point (0 %, 100 %),
      1-point (mg/L)
      2-point (0 %, 100 %),
      1-point (mg/L)
      Barometric pressure compensation Manual450 to 825 mmHg /
      59.9 to 109.9 kPa
      500 to 1499 mmHg /
      66.6 to 199.9 kPa (Manual Input)
      Salinity correction Manual0.0 to 50.0 ppt (Manual Input)0.0 to 50.0 ppt (Manual Input)
      Backlit display  Yes Yes --- 
      Number of channels111
      Data log500 with time and date stamp500 with time and date stamp---
      Data export optionsUSB or RS232IrDA, RS232C (via LED)---
      AC adapter9 V DC adapter, 1.3 A
      (100/240 VAC, SMPS) (optional)
      9 V DC adapter---
      Battery powerTwo AA batteries (included)Included—4AAs4 x AAA batteries
      Browse additional dissolved oxygen products
      Meter Accessories
      find the right benchtop meter for your pH and ORP measurement needs
      Shop our comprehensive range of meter accessories designed to boost productivity, safeguard your investment and ensure optimal performance. Our assortment of accessories for bench and portable meters helps streamline your workflow while enhancing your user experience.
      Electrodes and Probes
      Electrodes, probes, and sensors for use with conductivity meters
      Obtain accurate results with electrodes and probes developed to support the requirements of your sample testing. Choose from a wide variety of parameters, body materials, body styles, fill types, reference systems, junctions and more to meet the needs of your application.
      Standards and Solutions
      Orion standards and solutions
      Ensure the reliability of your measurements with our premium standards and solutions. Our high-quality calibration and maintenance solutions are essential for precise and consistent results. Shop a variety of standards, electrode filling solutions, electrode storage solutions and more.
      Orion Lab Star DO113 DO Bench Meter
      The Thermo Scientific Orion Lab Star DO113 Dissolved Oxygen Bench Meter is ideally suited for laboratories with users of varying levels of technical expertise and who require a simple-to-calibrate and use meter for routine dissolved oxygen testing. Watch this video to explore the features available with the Orion Lab Star DO Bench Meter.  
      Video Player is loading.
      Current Time 0:00
      Duration 1:17
      Loaded: 12.63%
      Stream Type LIVE
      Remaining Time 1:17
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Explore Multiparameter Meters
        Measure pH, ion concentration, conductivity, dissolved oxygen, and temperature with a single, powerful meter. Our bench and portable multiparameter meters are designed to increase your productivity in the lab and in the field.
        Learn more
        Request Consultation
        Get answers to your top water analysis and water purification questions and see products in action. Learn about:
        • pH & Multiparameter Benchtop & Portable Meters
        • Electrodes (pH Electrodes, ISE, Conductivity, DO)
        • Potentiometric Titrators
        • Spectrophotometers
        • Water Purification Systems
        Submit request
        Support
        Validation and qualification (IQ/OQ) services for Thermo Scientific Orion bench and portable meters are currently available through Unity Lab Services in the United States ›
        Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
        Orion Meters Software and Upgrades
        Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
        Find your software