Perform your ion concentration measurements with confidence
Achieve precise and reliable ion concentration measurements with Orion bench and portable ion concentration meters. Ideal for various applications, including pharmaceuticals, environmental monitoring, agriculture, food and beverage production and more, Orion ion concentration meters help you meet industry standards and achieve regulatory compliance. Combining advanced technology, user-friendly interfaces, and robust construction, these meters ensure accurate results. Suitable for both lab and field use, they measure using ion-selective electrode technology. Explore Orion ion concentration meters for precision and reliability. Browse our complete portfolio or let one of our experts assist you in making the right selection.
Select your ion concentration bench or portable meter
Premium
Advanced
 Orion Versa Star Pro
pH/Ion Bench Meter
(Versa Star Pro series)
Orion Pro Star PI217 Dual Channel pH/Ion Bench Meter
(Pro Star series)
Orion Pro Star PI214
pH/Ion Bench Meter
(Pro Star series)
 Premium benchtop Ion Specific (ISE) Meters
 Versatile, high-performance laboratory metersEasy to use, dependable
laboratory meters
Easy to use, dependable
laboratory meters
Ideal applicationsPharmaceuticals,
biotechnology, industrial
Environmental, water/ wastewater, food & beverage, industrialEnvironmental, water/ wastewater, food & beverage, industrial
pH range-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000
pH resolution0.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.1 
pH calibration pointsUp to 6Up to 5Up to 5
mV, RmV, ORP modesYesYesYes
Automatic buffer setsUSA/NIST, DIN, customUSA/NIST, DIN, pure water, standard, customUSA/NIST, DIN, pure water, standard, custom
Ion concentration range0.0001 to 19990 0.0001 to 19990 0.0001 to 19990 
Incremental techniquesSingle known addition or subtraction, double known addition or subtraction, analyte (sample) addition or subtractionSingle known addition or subtraction, double known addition or subtraction, analyte (sample) addition or subtractionSingle known addition or subtraction, double known addition or subtraction, analyte (sample) addition or subtraction
Temperature range-5 to 105°C
23 to 221°F
-30 to 130°C
-22 to 266°F
-30 to 130°C
-22 to 266°F
Number of channels4, customizable with modules21
Stirrer probe inputs221
Display5.3-inch, backlit color display7-inch, high-resolution,
backlit color display
7-inch, high-resolution,
backlit color display
TouchscreenNoYesYes
Onscreen text promptsStep-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Identification optionsElectrode ID, sample ID, user IDElectrode ID, sample ID, user IDElectrode ID, sample ID, user ID
Data log2000 with date/time10000 with date/time10000 with date/time
Calibration log30 per parameter and channel20 per parameter and channel20 per parameter
Data export optionsComputer, printerComputer, printer, USB flash driveComputer, printer, USB flash drive
Power supplyUniversal, 100–240 VAC, 50-60 Hz (Cat. No. STARA-PWR)Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Premium
Advanced
 Orion Star A329 pH/ISE
(STARA3290 series)
Orion Star A324 pH/ISE
(STARA3240 series)
 Premium portable Ion specific ISE metersAdvanced portable Ion specific ISE meters
 Versatile, powerful, multiparameter meterRugged and dependable multi-parameter meter
Ideal for
applications in:
Industrial—waste water, drinking water, food & beverage; Environmental—lakes, pondsIndustrial—waste water, cooling tanks; Environmental—lakes, ponds
pH range–2.000 to 20.000–2.000 to 20.000
pH resolution0.1/0.01/0.0010.1/0.01/0.001
pH relative accuracy±0.002±0.002
pH calibration pointsUp to 5Up to 5
pH calibration editingYesYes
Range (mV)±2000.0 mV±2000.0 mV
Resolution (mV)0.1 mV0.1 mV
Relative accuracy±0.2 mV or ±0.05% of reading, whichever is greater±0.2 mV or ±0.05% of reading, whichever is greater
EH ORP modeYesYes
Range (ISE)0.0001 to 19,9000.0001 to 19,900
Resolution (ISE)0.0001 minimum, auto ranging, up to 3 significant digits (user selectable)0.0001 minimum, auto ranging, up to 3 significant digits (user selectable)
Relative accuracy (ISE)±0.2 mV or ±0.05% of reading, whichever is greater±0.2 mV or ±0.05% of reading, whichever is greater
Unitsppm, M, mg/L, %, ppb, noneppm, M, mg/L, %, ppb, none
Calibration pointsUp to 5Up to 5
Calibration editingYesYes
Calibration featuresLinear point to point, non-linear selectable auto-blank, low concentration range stabilityLinear point to point, non-linear selectable auto-blank, low concentration range stability
Range (temperature)–5 to 105ºC, 23 to 221ºF–5 to 105ºC, 23 to 221ºF
Resolution (temperature)0.1ºC, 0.1ºF0.1ºC, 0.1ºF
Relative accuracy (temperature)±0.1ºC, ±0.1ºF±0.1ºC, ±0.1ºF
Temperature offset calibration1 point1 point
Number of datalogs5,000 with time and date stamp5,000 with time and date stamp
Log functionManual, ready (includes AUTOREAD), timedManual, ready, (includes AUTO-READ), timed
pH electrode or ISEBNC, reference pinBNC, reference pin
Conductivity or ATC probe8-pin mini DIN8-pin mini-DIN
DO probe9-pin mini-DIN--
OutputsRS232, USBRS232, USB
AC adapterOptional—universal, 100–240VACOptional—universal, 100-240VAC
Battery powerIncluded—4AAsIncluded—4 AAs
Battery life800 hrs800 hrs
  Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
Advanced
 Eutech ION2700 pH/Ion Bench Meter
 Economical, routine laboratory meters
pH range–2.000 to 20.000
pH resolution0.001 / 0.01 / 0.1
pH calibration pointsUp to 6
mV, RmV modesYes
Ion concentration range0.001 to 19900 ppm
Ion calibration points2 to 8
Range (temperature)0.0 to 100.0 °C, 32.0 to 212.0 °F
Number of channels1
Data log500 with date/time
Calibration logMost recent viewable on meter and stored in meter memory
Data export optionsRS232
Power supplyUniversal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401
 PremiumAdvancedBasic
 Eutech pH 450 Portable MeterEutech pH 620 Portable MeterEutech ION 6+ Portable Meter
pH range–2.00 to 16.00 pH-2.000 to  20.000 pH0.00 to 14.00 pH
pH resolution0.01 pH0.1 / 0.01 / 0.001 pH0.01 pH
pH relative accuracy±0.01 pH±0.002 pH±0.01 pH
pH calibration pointsUp to 6Up to 6Up to 3
Range (mV)±2000 mV±2000 mV±500 mV
Resolution (mV)0.1 mV ±999.9 mV; 1 mV beyond0.1 mV0.1 mV (±199.9 mV) / 1 mV beyond
Relative accuracy±0.2 mV +2 LSD or
+0.5 % of reading;
whichever is greater
±0.2 mV + 1 LSD±0.2 mV / ±2 mV + 1 LSD
pH ORP Mode---YesYes
Range (ISE)0.0 to 2000 ppm0.001 to 19900 0.01 to 0.99 / 1.0 to 199.9 / 200 to 1999 ppm
Calibration pointsFrom 2-4 or 2-5Up to 8Up to 3
Range (temperature)-10.0 to 110.0 ºC
(14.0 to 230.0 ºF)
-10.0 to 110.0 °C
(14.0 to 230.0 °F)
0.0 to 100.0 ºC
Number of datalogs500 with time and date stamp500 with time and date stamp---
Data export optionsUSB or RS232RS232C through LED, IrDA---
AC adapter 9 V DC adapter, 1.3 A
(100/240  VAC, SMPS) (optional)
9 V DC adapter (500 mA)---
Battery powerTwo AA batteries (included)Included—(4) AA4 x AAA batteries
Premium
Advanced
 Orion Versa Star Pro
pH/Ion Bench Meter
(Versa Star Pro series)
Orion Pro Star PI217 Dual Channel pH/Ion Bench Meter
(Pro Star series)
Orion Pro Star PI214
pH/Ion Bench Meter
(Pro Star series)
 Premium benchtop Ion Specific (ISE) Meters
 Versatile, high-performance laboratory metersEasy to use, dependable
laboratory meters
Easy to use, dependable
laboratory meters
Ideal applicationsPharmaceuticals,
biotechnology, industrial
Environmental, water/ wastewater, food & beverage, industrialEnvironmental, water/ wastewater, food & beverage, industrial
pH range-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000
pH resolution0.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.1 
pH calibration pointsUp to 6Up to 5Up to 5
mV, RmV, ORP modesYesYesYes
Automatic buffer setsUSA/NIST, DIN, customUSA/NIST, DIN, pure water, standard, customUSA/NIST, DIN, pure water, standard, custom
Ion concentration range0.0001 to 19990 0.0001 to 19990 0.0001 to 19990 
Incremental techniquesSingle known addition or subtraction, double known addition or subtraction, analyte (sample) addition or subtractionSingle known addition or subtraction, double known addition or subtraction, analyte (sample) addition or subtractionSingle known addition or subtraction, double known addition or subtraction, analyte (sample) addition or subtraction
Temperature range-5 to 105°C
23 to 221°F
-30 to 130°C
-22 to 266°F
-30 to 130°C
-22 to 266°F
Number of channels4, customizable with modules21
Stirrer probe inputs221
Display5.3-inch, backlit color display7-inch, high-resolution,
backlit color display
7-inch, high-resolution,
backlit color display
TouchscreenNoYesYes
Onscreen text promptsStep-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Identification optionsElectrode ID, sample ID, user IDElectrode ID, sample ID, user IDElectrode ID, sample ID, user ID
Data log2000 with date/time10000 with date/time10000 with date/time
Calibration log30 per parameter and channel20 per parameter and channel20 per parameter
Data export optionsComputer, printerComputer, printer, USB flash driveComputer, printer, USB flash drive
Power supplyUniversal, 100–240 VAC, 50-60 Hz (Cat. No. STARA-PWR)Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Premium
Advanced
 Orion Star A329 pH/ISE
(STARA3290 series)
Orion Star A324 pH/ISE
(STARA3240 series)
 Premium portable Ion specific ISE metersAdvanced portable Ion specific ISE meters
 Versatile, powerful, multiparameter meterRugged and dependable multi-parameter meter
Ideal for
applications in:
Industrial—waste water, drinking water, food & beverage; Environmental—lakes, pondsIndustrial—waste water, cooling tanks; Environmental—lakes, ponds
pH range–2.000 to 20.000–2.000 to 20.000
pH resolution0.1/0.01/0.0010.1/0.01/0.001
pH relative accuracy±0.002±0.002
pH calibration pointsUp to 5Up to 5
pH calibration editingYesYes
Range (mV)±2000.0 mV±2000.0 mV
Resolution (mV)0.1 mV0.1 mV
Relative accuracy±0.2 mV or ±0.05% of reading, whichever is greater±0.2 mV or ±0.05% of reading, whichever is greater
EH ORP modeYesYes
Range (ISE)0.0001 to 19,9000.0001 to 19,900
Resolution (ISE)0.0001 minimum, auto ranging, up to 3 significant digits (user selectable)0.0001 minimum, auto ranging, up to 3 significant digits (user selectable)
Relative accuracy (ISE)±0.2 mV or ±0.05% of reading, whichever is greater±0.2 mV or ±0.05% of reading, whichever is greater
Unitsppm, M, mg/L, %, ppb, noneppm, M, mg/L, %, ppb, none
Calibration pointsUp to 5Up to 5
Calibration editingYesYes
Calibration featuresLinear point to point, non-linear selectable auto-blank, low concentration range stabilityLinear point to point, non-linear selectable auto-blank, low concentration range stability
Range (temperature)–5 to 105ºC, 23 to 221ºF–5 to 105ºC, 23 to 221ºF
Resolution (temperature)0.1ºC, 0.1ºF0.1ºC, 0.1ºF
Relative accuracy (temperature)±0.1ºC, ±0.1ºF±0.1ºC, ±0.1ºF
Temperature offset calibration1 point1 point
Number of datalogs5,000 with time and date stamp5,000 with time and date stamp
Log functionManual, ready (includes AUTOREAD), timedManual, ready, (includes AUTO-READ), timed
pH electrode or ISEBNC, reference pinBNC, reference pin
Conductivity or ATC probe8-pin mini DIN8-pin mini-DIN
DO probe9-pin mini-DIN--
OutputsRS232, USBRS232, USB
AC adapterOptional—universal, 100–240VACOptional—universal, 100-240VAC
Battery powerIncluded—4AAsIncluded—4 AAs
Battery life800 hrs800 hrs
  Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
Advanced
 Eutech ION2700 pH/Ion Bench Meter
 Economical, routine laboratory meters
pH range–2.000 to 20.000
pH resolution0.001 / 0.01 / 0.1
pH calibration pointsUp to 6
mV, RmV modesYes
Ion concentration range0.001 to 19900 ppm
Ion calibration points2 to 8
Range (temperature)0.0 to 100.0 °C, 32.0 to 212.0 °F
Number of channels1
Data log500 with date/time
Calibration logMost recent viewable on meter and stored in meter memory
Data export optionsRS232
Power supplyUniversal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401
 PremiumAdvancedBasic
 Eutech pH 450 Portable MeterEutech pH 620 Portable MeterEutech ION 6+ Portable Meter
pH range–2.00 to 16.00 pH-2.000 to  20.000 pH0.00 to 14.00 pH
pH resolution0.01 pH0.1 / 0.01 / 0.001 pH0.01 pH
pH relative accuracy±0.01 pH±0.002 pH±0.01 pH
pH calibration pointsUp to 6Up to 6Up to 3
Range (mV)±2000 mV±2000 mV±500 mV
Resolution (mV)0.1 mV ±999.9 mV; 1 mV beyond0.1 mV0.1 mV (±199.9 mV) / 1 mV beyond
Relative accuracy±0.2 mV +2 LSD or
+0.5 % of reading;
whichever is greater
±0.2 mV + 1 LSD±0.2 mV / ±2 mV + 1 LSD
pH ORP Mode---YesYes
Range (ISE)0.0 to 2000 ppm0.001 to 19900 0.01 to 0.99 / 1.0 to 199.9 / 200 to 1999 ppm
Calibration pointsFrom 2-4 or 2-5Up to 8Up to 3
Range (temperature)-10.0 to 110.0 ºC
(14.0 to 230.0 ºF)
-10.0 to 110.0 °C
(14.0 to 230.0 °F)
0.0 to 100.0 ºC
Number of datalogs500 with time and date stamp500 with time and date stamp---
Data export optionsUSB or RS232RS232C through LED, IrDA---
AC adapter 9 V DC adapter, 1.3 A
(100/240  VAC, SMPS) (optional)
9 V DC adapter (500 mA)---
Battery powerTwo AA batteries (included)Included—(4) AA4 x AAA batteries
Browse additional ion concentration products
Meter Accessories
find the right benchtop meter for your pH and ORP measurement needs
Shop our comprehensive range of meter accessories designed to boost productivity, safeguard your investment and ensure optimal performance. Our assortment of accessories for bench and portable meters helps streamline your workflow while enhancing your user experience.
Electrodes and Probes
Electrodes, probes, and sensors for use with conductivity meters
Obtain accurate results with electrodes and probes developed to support the requirements of your sample testing. Choose from a wide variety of parameters, body materials, body styles, fill types, reference systems, junctions and more to meet the needs of your application.
Standards and Solutions
Orion standards and solutions
Ensure the reliability of your measurements with our premium standards and solutions. Our high-quality calibration and maintenance solutions are essential for precise and consistent results. Shop a variety of standards, electrode filling solutions, electrode storage solutions and more.
Explore Multiparameter Meters
Measure pH, ion concentration, conductivity, dissolved oxygen, and temperature with a single, powerful meter. Our bench and portable multiparameter meters are designed to increase your productivity in the lab and in the field.
Learn more
Request Consultation
Get answers to your top water analysis and water purification questions and see products in action. Learn about:
  • pH & Multiparameter Benchtop & Portable Meters
  • Electrodes (pH Electrodes, ISE, Conductivity, DO)
  • Potentiometric Titrators
  • Spectrophotometers
  • Water Purification Systems
Submit request
Support
Validation and qualification (IQ/OQ) services for Thermo Scientific Orion bench and portable meters are currently available through Unity Lab Services in the United States ›
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
Find your software
Style Sheet for Global Design System