Search Thermo Fisher Scientific
Measure pH, ion concentration, conductivity, dissolved oxygen, and temperature with a single, powerful meter. Our benchtop and portable multiparameter meters are designed to increase your productivity in the lab and in the field. Choosing a multiparameter meter reduces the amount of equipment on your benchtop or in your hands while in the field, eliminates the need for multiple meters as the display showcases multiple parameters on one unit, and saves time in transferring electrodes from meter to meter for measurements. Combine your parameters today for simultaneous measurements saving space, time, and money. Browse our complete portfolio or let one of our experts help you make a selection.
Premium | Advanced | ||||
---|---|---|---|---|---|
Orion Versa Star Pro pH/Ion, Conductivity & DO Bench Meter (Versa Star Pro series) | Orion Versa Star Pro pH/Conductivity Bench Meter (Versa Star Pro series) | Orion Pro Star MP218 pH/Ion, Conductivity & DO Bench Meter (Pro Star series) | Orion Pro Star PD216 pH/Ion & DO Bench Meter (Pro Star series) | Orion Pro Star PC215 pH/Ion & Conductivity Bench Meter (Pro Star series) | |
![]() | ![]() | ![]() | ![]() | ![]() | |
Multiparameter high-performance laboratory meters | Dual parameter high-performance laboratory meters | Multiparameter easy to use, dependable laboratory meters | Multiparameter easy to use, dependable laboratory meters | Multiparameter easy to use, dependable laboratory meters | |
Ideal applications | Food & beverage, industrial, water | Pharmaceutical, biotechnology, food & beverage | Environmental, water/ wastewater, food & beverage, industrial | Environmental, water/ wastewater, food & beverage, industrial | Environmental, water/ wastewater, food & beverage, industrial |
pH range | -2.000 to 20.000 | -2.000 to 20.000 | -2.000 to 20.000 | -2.000 to 20.000 | -2.000 to 20.000 |
pH resolution | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 |
pH calibration points | Up to 6 | Up to 6 | Up to 5 | Up to 5 | Up to 5 |
mV, RmV, ORP modes | Yes | Yes | Yes | Yes | Yes |
Ion concentration mode | Yes | No | Yes | Yes | Yes |
Conductivity mode | Yes | Yes | Yes | No | Yes |
Additional modes | TDS, salinity, resistivity | TDS, salinity, resistivity | TDS, salinity, resistivity | — | TDS, salinity, resistivity |
Dissolved oxygen mode | Yes | No | Yes | Yes | No |
Compatible probes | Optical, polarographic | — | Optical, polarographic | Optical, polarographic | — |
Temperature range | -5 to 105°C 23 to 221°F | -5 to 105°C 23 to 221°F | -30 to 130ºC -22 to 266ºF | -30 to 130ºC -22 to 266ºF | -30 to 130ºC -22 to 266ºF |
Number of channels | 4, customizable with modules | 4, customizable with modules | 3 | 2 | 2 |
Stirrer probe inputs | 2 | 2 | 2 | 2 | 2 |
Display | 5.3-inch, backlit color display | 5.3-inch, backlit color display | 7-inch, high- resolution, backlit color display | 7-inch, high-resolution, backlit color display | 7-inch, high-resolution, backlit color display |
Touchscreen | No | No | Yes | Yes | Yes |
Onscreen text prompts | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions |
Data log | 2000 with date/time | 2000 with date/time | 10000 with date/time | 10000 with date/time | 10000 with date/time |
Calibration log | 30 per parameter | 30 per parameter | 20 per parameter | 20 per parameter | 20 per parameter |
Identification options | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID |
Data export options | USB (2), RS232 | USB (2), RS232 | Computer, printer, USB flash drive | Computer, printer, USB flash drive | Computer, printer, USB flash drive |
Power supply | Universal, 100–240 VAC, 50–60 Hz (Cat. No. STARA-PWR) | Universal, 100–240 VAC, 50–60 Hz (Cat. No. STARA-PWR) | Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR), 9 DC adapter, 2.0A | Universal, 100-240 VAC, 50-60 Hz, (Cat. No. PSTAR-PWR), 9 DC adapter, 2.0A | Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR), 9 DC adapter, 2.0A |
Premium | Premium | Premium | |
---|---|---|---|
Orion Star A325 Multiparameter meter (STARA3250 series) | Orion Star A326 Multiparameter meter (STARA3260 series) | Orion Star A329 Multiparameter meter (STARA3290 series) | |
![]() | ![]() | ![]() | |
Rugged, dependable multiparameter meter | Rugged, versatile, and dependable multiparameter meter | Versatile, powerful, multiparameter meter | |
Ideal for applications in: | Industrial—cooling towers, wastewater; Environmental—ponds, lakes | Industrial—wastewater; Environmental—ponds, lakes | Industrial—wastewater, drinking water, food & beverage; Environmental—ponds, lakes |
pH range | –2.000 to 20.000 | –2.000 to 20.000 | –2.000 to 20.000 |
pH resolution | 0.1/0.01/0.001 | 0.1/0.01/0.001 | 0.1/0.01/0.001 |
pH relative accuracy | ±0.002 | ±0.002 | ±0.002 |
pH calibration points | Up to 5 | Up to 5 | Up to 5 |
mV, RmV range | ±2,000.0 mV | ±2,000.0 mV | ±2,000.0 mV |
EH/ORP mode | Yes | Yes | Yes |
Dissolved oxygen mode | No | Yes | Yes |
TDS relative accuracy | ±0.5% reading ±1 digit | -- | ±0.5% reading ±1 digit |
Temperature range | –5 to 105°C, 23 to 221°F | –5 to 105ºC, 23 to 221ºF | –5 to 105ºC, 23 to 221ºF |
Number of datalogs | 5000, with time and date stamp | 5000, with time and date stamp | 5000, with time and date stamp |
Data export options | RS232, USB | RS232, USB | RS232, USB |
AC adapter | Optional—universal, 100–240VAC | Optional—universal, 100–240VAC | Optional—universal, 100–240VAC |
Battery power | Included—4AAs | Included—4AAs | Included—4AAs |
Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories › |
Advanced | |
---|---|
Eutech PC2700 pH/Conductivity Bench Meter | |
![]() | |
Economical, routine laboratory meters | |
pH range | -2.000 to 20.000 |
pH resolution | 0.001 / 0.01 / 0.1 |
pH calibration points | Up to 6 |
mV, RmV modes | Yes |
Ion concentration mode | Yes |
Conductivity mode | Yes |
Additional modes | TDS, salinity, resistivity |
Range (temperature) | 0.0 to 100.0 °C, 32.0 to 212.0 °F |
Backlit display | Yes |
Number of channels | 2 |
Data log | 500 with date/time |
Calibration log | Most recent viewable on meter and stored in meter memory |
Data export options | RS232 |
Power supply | Universal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401 |
Premium | Premium | Advanced | Advanced | Advanced | Advanced | |
---|---|---|---|---|---|---|
Eutech PD 450 Portable Multiparameter Meter | Eutech PC 450 Portable Multiparameter Meter | Eutech PCD 650 Portable Multiparameter Meter | Eutech PC 650 Portable Multiparameter Meter | Eutech PD 650 Portable Multiparameter Meter | Eutech CD 650 Portable Multiparameter Meter | |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
pH range | -2.00 to 16.00 | -2.00 to 16.00 | -2.00 to 20.00 | -2.00 to 20.00 | -2.00 to 20.00 | --- |
pH resolution | 0.01 pH | 0.01 pH | 0.1 / 0.01 Ph | 0.1 / 0.01 Ph | 0.1 / 0.01 Ph | --- |
pH relative accuracy | ±0.01 pH | ±0.01 pH | ±0.01 pH | ±0.01 pH | ±0.01 pH | --- |
pH calibration points | Up to 6 | Up to 6 | Up to 6 | Up to 6 | Up to 6 | --- |
mV, RmV modes | Yes | Yes | Yes | Yes | Yes | --- |
EH/ORP mode | --- | --- | Yes | Yes | Yes | --- |
Conductivity mode | --- | Yes | Yes | Yes | --- | Yes |
Dissolved oxygen mode | Yes | --- | Yes | --- | Yes | Yes |
TDS relative accuracy | --- | ±1 % full scale | ±1 % full scale | ±1 % full scale | ±1 % full scale | ±1 % full scale + |
Ion concentration mode | --- | --- | Yes | Yes | Yes | --- |
Temperature range | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) |
Number of datalogs | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp |
Data export options | USB or RS232 | USB or RS232 | RS232C through LED, IrDA | RS232C through LED, IrDA | RS232C through LED, IrDA | RS232C through LED, IrDA |
AC adapter | 9 V DC adapter (optional) | 9 V DC adapter (optional) | 9 V DC adapter | 9 V DC adapter | 9 V DC adapter | 9 V DC adapter |
Battery power | 2 AA batteries (included) | 2 AA batteries (included) | 4 AA batteries (included) | 4 AA batteries (included) | 4 AA batteries (included) | 4 AA batteries (included) |
Premium | Advanced | ||||
---|---|---|---|---|---|
Orion Versa Star Pro pH/Ion, Conductivity & DO Bench Meter (Versa Star Pro series) | Orion Versa Star Pro pH/Conductivity Bench Meter (Versa Star Pro series) | Orion Pro Star MP218 pH/Ion, Conductivity & DO Bench Meter (Pro Star series) | Orion Pro Star PD216 pH/Ion & DO Bench Meter (Pro Star series) | Orion Pro Star PC215 pH/Ion & Conductivity Bench Meter (Pro Star series) | |
![]() | ![]() | ![]() | ![]() | ![]() | |
Multiparameter high-performance laboratory meters | Dual parameter high-performance laboratory meters | Multiparameter easy to use, dependable laboratory meters | Multiparameter easy to use, dependable laboratory meters | Multiparameter easy to use, dependable laboratory meters | |
Ideal applications | Food & beverage, industrial, water | Pharmaceutical, biotechnology, food & beverage | Environmental, water/ wastewater, food & beverage, industrial | Environmental, water/ wastewater, food & beverage, industrial | Environmental, water/ wastewater, food & beverage, industrial |
pH range | -2.000 to 20.000 | -2.000 to 20.000 | -2.000 to 20.000 | -2.000 to 20.000 | -2.000 to 20.000 |
pH resolution | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 | 0.001 / 0.01 / 0.1 |
pH calibration points | Up to 6 | Up to 6 | Up to 5 | Up to 5 | Up to 5 |
mV, RmV, ORP modes | Yes | Yes | Yes | Yes | Yes |
Ion concentration mode | Yes | No | Yes | Yes | Yes |
Conductivity mode | Yes | Yes | Yes | No | Yes |
Additional modes | TDS, salinity, resistivity | TDS, salinity, resistivity | TDS, salinity, resistivity | — | TDS, salinity, resistivity |
Dissolved oxygen mode | Yes | No | Yes | Yes | No |
Compatible probes | Optical, polarographic | — | Optical, polarographic | Optical, polarographic | — |
Temperature range | -5 to 105°C 23 to 221°F | -5 to 105°C 23 to 221°F | -30 to 130ºC -22 to 266ºF | -30 to 130ºC -22 to 266ºF | -30 to 130ºC -22 to 266ºF |
Number of channels | 4, customizable with modules | 4, customizable with modules | 3 | 2 | 2 |
Stirrer probe inputs | 2 | 2 | 2 | 2 | 2 |
Display | 5.3-inch, backlit color display | 5.3-inch, backlit color display | 7-inch, high- resolution, backlit color display | 7-inch, high-resolution, backlit color display | 7-inch, high-resolution, backlit color display |
Touchscreen | No | No | Yes | Yes | Yes |
Onscreen text prompts | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions | Step-by-step calibration and guided setup instructions |
Data log | 2000 with date/time | 2000 with date/time | 10000 with date/time | 10000 with date/time | 10000 with date/time |
Calibration log | 30 per parameter | 30 per parameter | 20 per parameter | 20 per parameter | 20 per parameter |
Identification options | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID | Electrode ID, sample ID, user ID |
Data export options | USB (2), RS232 | USB (2), RS232 | Computer, printer, USB flash drive | Computer, printer, USB flash drive | Computer, printer, USB flash drive |
Power supply | Universal, 100–240 VAC, 50–60 Hz (Cat. No. STARA-PWR) | Universal, 100–240 VAC, 50–60 Hz (Cat. No. STARA-PWR) | Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR), 9 DC adapter, 2.0A | Universal, 100-240 VAC, 50-60 Hz, (Cat. No. PSTAR-PWR), 9 DC adapter, 2.0A | Universal, 100-240 VAC, 50-60 Hz (Cat. No. PSTAR-PWR), 9 DC adapter, 2.0A |
Premium | Premium | Premium | |
---|---|---|---|
Orion Star A325 Multiparameter meter (STARA3250 series) | Orion Star A326 Multiparameter meter (STARA3260 series) | Orion Star A329 Multiparameter meter (STARA3290 series) | |
![]() | ![]() | ![]() | |
Rugged, dependable multiparameter meter | Rugged, versatile, and dependable multiparameter meter | Versatile, powerful, multiparameter meter | |
Ideal for applications in: | Industrial—cooling towers, wastewater; Environmental—ponds, lakes | Industrial—wastewater; Environmental—ponds, lakes | Industrial—wastewater, drinking water, food & beverage; Environmental—ponds, lakes |
pH range | –2.000 to 20.000 | –2.000 to 20.000 | –2.000 to 20.000 |
pH resolution | 0.1/0.01/0.001 | 0.1/0.01/0.001 | 0.1/0.01/0.001 |
pH relative accuracy | ±0.002 | ±0.002 | ±0.002 |
pH calibration points | Up to 5 | Up to 5 | Up to 5 |
mV, RmV range | ±2,000.0 mV | ±2,000.0 mV | ±2,000.0 mV |
EH/ORP mode | Yes | Yes | Yes |
Dissolved oxygen mode | No | Yes | Yes |
TDS relative accuracy | ±0.5% reading ±1 digit | -- | ±0.5% reading ±1 digit |
Temperature range | –5 to 105°C, 23 to 221°F | –5 to 105ºC, 23 to 221ºF | –5 to 105ºC, 23 to 221ºF |
Number of datalogs | 5000, with time and date stamp | 5000, with time and date stamp | 5000, with time and date stamp |
Data export options | RS232, USB | RS232, USB | RS232, USB |
AC adapter | Optional—universal, 100–240VAC | Optional—universal, 100–240VAC | Optional—universal, 100–240VAC |
Battery power | Included—4AAs | Included—4AAs | Included—4AAs |
Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories › |
Advanced | |
---|---|
Eutech PC2700 pH/Conductivity Bench Meter | |
![]() | |
Economical, routine laboratory meters | |
pH range | -2.000 to 20.000 |
pH resolution | 0.001 / 0.01 / 0.1 |
pH calibration points | Up to 6 |
mV, RmV modes | Yes |
Ion concentration mode | Yes |
Conductivity mode | Yes |
Additional modes | TDS, salinity, resistivity |
Range (temperature) | 0.0 to 100.0 °C, 32.0 to 212.0 °F |
Backlit display | Yes |
Number of channels | 2 |
Data log | 500 with date/time |
Calibration log | Most recent viewable on meter and stored in meter memory |
Data export options | RS232 |
Power supply | Universal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401 |
Premium | Premium | Advanced | Advanced | Advanced | Advanced | |
---|---|---|---|---|---|---|
Eutech PD 450 Portable Multiparameter Meter | Eutech PC 450 Portable Multiparameter Meter | Eutech PCD 650 Portable Multiparameter Meter | Eutech PC 650 Portable Multiparameter Meter | Eutech PD 650 Portable Multiparameter Meter | Eutech CD 650 Portable Multiparameter Meter | |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
pH range | -2.00 to 16.00 | -2.00 to 16.00 | -2.00 to 20.00 | -2.00 to 20.00 | -2.00 to 20.00 | --- |
pH resolution | 0.01 pH | 0.01 pH | 0.1 / 0.01 Ph | 0.1 / 0.01 Ph | 0.1 / 0.01 Ph | --- |
pH relative accuracy | ±0.01 pH | ±0.01 pH | ±0.01 pH | ±0.01 pH | ±0.01 pH | --- |
pH calibration points | Up to 6 | Up to 6 | Up to 6 | Up to 6 | Up to 6 | --- |
mV, RmV modes | Yes | Yes | Yes | Yes | Yes | --- |
EH/ORP mode | --- | --- | Yes | Yes | Yes | --- |
Conductivity mode | --- | Yes | Yes | Yes | --- | Yes |
Dissolved oxygen mode | Yes | --- | Yes | --- | Yes | Yes |
TDS relative accuracy | --- | ±1 % full scale | ±1 % full scale | ±1 % full scale | ±1 % full scale | ±1 % full scale + |
Ion concentration mode | --- | --- | Yes | Yes | Yes | --- |
Temperature range | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) | -10.0 to 110.0 °C (14.0 to 230.0 °F) |
Number of datalogs | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp | 500 with date and time stamp |
Data export options | USB or RS232 | USB or RS232 | RS232C through LED, IrDA | RS232C through LED, IrDA | RS232C through LED, IrDA | RS232C through LED, IrDA |
AC adapter | 9 V DC adapter (optional) | 9 V DC adapter (optional) | 9 V DC adapter | 9 V DC adapter | 9 V DC adapter | 9 V DC adapter |
Battery power | 2 AA batteries (included) | 2 AA batteries (included) | 4 AA batteries (included) | 4 AA batteries (included) | 4 AA batteries (included) | 4 AA batteries (included) |
Shop our comprehensive range of meter accessories designed to boost productivity, safeguard your investment and ensure optimal performance. Our assortment of accessories for bench and portable meters helps streamline your workflow while enhancing your user experience.
Obtain accurate results with electrodes and probes developed to support the requirements of your sample testing. Choose from a wide variety of parameters, body materials, body styles, fill types, reference systems, junctions and more to meet the needs of your application.
Ensure the reliability of your measurements with our premium buffers, standards and solutions. Our high-quality calibration and maintenance solutions are essential for precise and consistent results. Shop a variety of buffers, buffer kits, standards, electrode filling solutions, electrode storage solutions and more.
Discover the Orion Versa Star Pro multiparameter bench meter in an immersive 3D tour, showcasing its advanced features and intuitive design. Ideal for laboratories, this virtual experience offers a detailed look at how the meter enhances precision and efficiency. Explore why professionals worldwide trust the Orion Versa Star Pro for their multiparameter measurement needs.
Get answers to your top water analysis and water purification questions and see products in action. Learn about:
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.