Search Thermo Fisher Scientific
Phosphoramidites for Oligonucleotide Synthesis |
Thermo Scientific phosphoramidites are built to help meet varying needs of oligonucleotide synthesis from research applications to commercial manufacturing for diagnostic and therapeutic developers. For research and early development, there are over 136 ready-to-ship phosphoramidites (abbreviated as amidites below) of small pack sizes in the online catalog.
Custom solutions and modification capabilities are available to support you from the beginning at the discovery phase. Our core scale-up competencies help ensure continuity all the way to the commercial volume needed for go-to-market. With manufacturing expertise in custom nucleic acid chemistry for more than 40 years, our technical experts help ensure the consistent quality and security of supply chain for scaling up from grams to metric tons.
Amidites are the building blocks used in chemical synthesis of oligonucleotides. Optimize your oligo synthesis and manufacturing process development with our large selection of amidites offerings that are also available in two quality grades.
Common base modifications—2′-MOE, 2′-OMe, and 2′-fluoro—for the synthesis of antisense oligos and siRNAs.
For synthesis of DNA oligos for diagnostic assays and therapeutic applications.
Synthesis of RNA oligos for diagnostic assays and therapeutic applications.
With fluorescent labels for synthesis of oligos for detection in molecular assays and microarrays.
Linkers at 3′, 5′, or internal positions for NHS ester conjugation; and spacers to increase conformational support.
With modifications (e.g., 5m-dC, N6-Me-dA, N4-Ethyl-dC, 8-oxo-dG) for synthesis of primers, probes, or oligos.
DNA succinates for the derivatization of controlled pore glass, polystyrene, or other supports used in solid-phase oligo synthesis.
Processes are monitored from starting material procurement to final QC and product release. Our rigorously controlled manufacturing processes help maximize batch-to-batch reproducibility and minimize levels of reactive P(III) impurities, which could accumulate as by-products into the oligonucleotide during synthesis.
TheraPure | Standard | |
---|---|---|
Recommended application |
|
|
Animal origin free (AOF) manufacturing | Manufactured with animal origin free (AOF) materials | |
HPLC purity | 99% or higher with unknown, non-critical, and critical impurity specifications | 98% |
31P NMR purity | 99% or higher with unknown, non-critical, and critical impurity specifications | 98% |
Water content | 0.3% or lower | |
Solution clarity | 0.2 M solution in acetonitrile is free of undissolved particulates | – |
UV spectral identification | The λ maxima of the sample and standard spectra are within ±2 nm | – |
Residual solvents | 4.0% or lower | – |
Phosphoramidites are the key raw material and building blocks of oligonucleotide therapeutics. Incorporating the quality testing step into the manufacturing process development from the beginning is a best practice for developing therapeutics or diagnostics from discovery to commercialization. The comprehensive QC processes include the analysis of phosphoramidites that is crucial to monitoring the impurity levels.
Technical expertise
Over 40 years of expertise in custom chemistry and scale-up experience for innovative projects.
Security of supply
5x capacity expansion to enable emerging diagnostic assays and oligo therapeutics.
Refined quality
ISO 9001:2015–certified quality management system for nucleic acid products
Quality manufacturing and documentation
Batch-to-batch consistency, minimal level of reactive impurities, and analytical documentation support.
Service excellence
48 hours or less response to orders, >98% delivery to customer at promised date*.
Flexible packaging
Multiple customization formats of synthesizers, solvent methods, capping, fill volumes, and more.
Our scientists discuss the history of Thermo Fisher's Milwaukee facility, customer partnership, manufacturing process, quality improvement, mitigation of supply chain issues, and the recent response to global needs.
Quick tour of the Thermo Fisher Scientific facility at Milwaukee for phosphoramidites and nucleotides
In addition to our Thermo Scientific phosphoramidites, we offer custom services for oligonucleotide chemistry tailored to fit your needs in diagnostic and therapeutic applications. We are fully set up to provide customization of our existing phosphoramidites, synthesis of new compounds, and development of oligonucleotide delivery chemistries. Simply fill out this short form with your interest so that our representative can explore available options with you.
![]() | Phosphoramidite customization options A. Dimethoxytrityl (DMT) substitution with alternate protecting groups B. Base = A, C, G, T, or U protection modifications or base modifications C. Sugar modifications and protection options |
TheraPure services—access to customized materials for producing nucleic acid therapeutics
Depending on your selected phosphoramidites, they may be available in one or more of these options. Please contact us if you do not find one that meets your needs.
Base N-protections to choose depending on your deprotection methods, their sensitivity to base hydrolysis, and their reactivity with tags and labels:
| Package formats to accommodate your synthesis scales, synthesizers, and solvation methods:
| |
Speak with a technical specialist about support available for the oligonucleotide manufacturing process development of your unique projects.
Access webinars, videos, articles, and more to optimize your success in oligonucleotide synthesis for diagnostic and therapeutic manufacturing process development.
Browse the educational assets related to raw materials selection, process development using in vitro transcription, scale-up synthesis, and the latest trends in mRNA production.
For Research Use Only. Not for use in diagnostic procedures.