Search Thermo Fisher Scientific
Linker and Spacer Phosphoramidites |
Linker phosphoramidites provide options to link oligonucleotides to additional groups (e.g., chemical delivery system for therapeutics, fluorescent dyes, amines, phosphates) or attach to a solid surface like a DNA chip. On the other hand, spacer phosphoramidites can alter oligonucleotide conformation and/or reduce steric hinderance for applications such as aptamers and tagging/labeling assays.
Thermo Scientific 5′ amino linker ß-cyanoethyl phosphoramidites (CEP) are available as MMT or TFA protected.
Thermo Scientific phosphorylating CEP can be used for addition of a phosphate group to the 5′ position of an oligonucleotide (final synthesis addition), or 3′ end when used as the first addition off a solid support.
Linker phosphoramidites | |||
---|---|---|---|
5′-Aminohexyl Linker CEP | TFA-Aminolinker | Phosphorylating CEP | |
Applied Biosystems/AKTA Synthesizer | Custom orders available for 27-0035. Request quote | 27-1792 Request quote | 27-1794-04 |
Expedite Synthesizer | 27-1794-09 | ||
MerMade Synthesizer | 27-1794-10 |
Click images to enlarge
5′-Aminohexyl Linker CEP | TFA-Aminolinker CEP | Phosphorylating CEP | |
---|---|---|---|
Cat. No. | 27-0035 | 27-1792 | 27-1794 |
Chemical name | 6-(4-monomethoxytritylamino)-hexyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite | 6-(N-trifluoroacetylamino)hexyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite | 2-[2-(4,4-dimethoxytrityloxy)ethylsulfonyl]ethyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite |
Formula | C35H48N3O3P | C17H31F3N3O3P | C34H45N2O7PS |
Molecular weight | 589.75 g/mol | 413.42 g/mol | 656.66 g/mol |
CAS # | 114616-27-2 | 133975-85-6 | 108783-02-4 |
HPLC purity | ≥97% | NA | ≥95% |
31P NMR purity | NA | ≥90.0% | NA |
Shipping condition | Dry ice | Dry ice | Ambient, desiccated. Keep vial upright during storage |
Water content | NA | ≤0.3% | NA |
Storage condition | –20°C |
Linker phosphoramidites are compatible with the following oligonucleotide synthesizers:
Available catalog SKU is supplied in 30 mL amber bottle with a 20-400 finish neck style.
Order catalog product ready-to-ship in a 30 mL amber bottle with a 28-400 finish neck style.
The Thermo Scientific C3 spacer CEP can be used to incorporate a three-carbon spacer or add sequentially to create longer spacer arms. Thermo Scientific spacer 18 hexaethyleneglycol (HEG) CEP helps reduce possible steric hinderance effects and/or protection from exonucleases. Thermo Scientific reverse abasic CEP introduces abasic sites in oligonucleotides to study their biological effects.
Spacer phosphoramidites | |||
---|---|---|---|
C3-Spacer-CEP | HEG (Spacer 18) phosphoramidite | Reverse abasic CEP | |
Applied Biosystems/AKTA Synthesizer | 27-1795-04 | 27-1786-02 | 27-1998 Request quote |
Expedite Synthesizer | |||
MerMade Synthesizer | 27-1795 Request quote | 27-1786 Request quote |
C3-Spacer-CEP | HEG (Spacer 18) phosphoramidite | Reverse abasic CEP | |
---|---|---|---|
Cat. No. | 27-1795 | 27-1786 | 27-1998 |
Chemical name | 3-(4,4'-dimethoxytrityloxy)propyl-1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite | 18-O-(4,4'-dimethoxytrityl)-Hexaethyleneglycol-1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite | 3-O-(4,4’-dimethoxytrityl)-1,4-anhydro-2-deoxy-D-ribitol-5-O-(β-cyanoethyl-N,N-diisopropyl) |
Formula | C33H43N2O5P | C42H61N2O10P | C35H45N2O6P |
Molecular weight | 578.7 g/mol | 784.9 g/mol | 620.72 g/mol |
HPLC purity | ≥98% | ≥95% | ≥98% |
31P NMR purity | ≥98% | ≥98% | ≥98% |
Material source | Non animal source (no TSE/BSE risk) | ||
Shipping / storage condition | Dry ice / –20°C |
Spacer phosphoramidites are compatible with the following oligonucleotide synthesizers:
Compatible catalog product is ready-to-ship in amber bottles with serum vial neck style.
Looking for additional size options, customization, or packaging solutions?
Speak with a technical specialist about support available for the oligonucleotide manufacturing process development of your unique projects.
Access webinars, videos, articles, and more to optimize your success in oligonucleotide synthesis for diagnostic and therapeutic manufacturing process development.
Browse the educational assets related to raw materials selection, process development using in vitro transcription, scale-up synthesis, and the latest trends in mRNA production.
For Research Use Only. Not for use in diagnostic procedures.