Structural phosphoramidites are nonstandard amidites used to synthesize modified oligonucleotides. Oligos with these specialty phosphoramidites are often used as primers, probes, or in structural studies. Structural phosphoramidites can be custom manufactured with base or sugar (2′, 3′, or 5′ positions) modifications.
N4-ethyl-dC, 5-methyl-dC, dI, and dU for duplex stability studies
The stability and melting point of oligonucleotide duplexes can be altered by incorporating N4-ethyl deoxycytosine (N4-Et-dC), 5-methyl deoxycytosine (5-Me-dC), deoxyinosine (dI), or deoxyurosine (dU) into the oligonucleotide sequence.
Availability by compatible synthesizer model
Common applicationDuplex stability studies 
 N4-Et-dC 5-Me-dCdI dU 
Applied Biosystems/AKTA SynthesizerCustom orders available.
Request quote
27-1748-0427-1744-0427-1738-03
Expedite Synthesizer
Request quote27-1744-1027-1738-08
Technical specifications for N4-ethyl-dC, 5-methyl-dC, dI, and dU phosphoramidites
Thermo Scientific N4-Et-dC, 5-Me-dC, dI, and dU β-cyanoethyl phosphoramidites (CEP) are used in specialty applications such as duplex stability studies. Thermo Scientific dI CEP facilitates pairing with all four bases as a degenerate base in oligo probes and primers and can also provide the oligos nuclease resistance.
  Click images to enlarge.
 N4-Et-dC CEP5-Me-dC CEPdI CEPdU CEP
Cat. No.27-174327-174827-174427-1738
Phosphoramidite description5′-O-(4,4'-dimethoxytrityl)-N4-ethyl-2′-deoxycytidine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite5′-O-(4,4'-dimethoxytrityl)-N4-benzoyl-5-methyl-2′-deoxycytidine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite5′-O-(4,4'-dimethoxytrityl)-2′-deoxyinosine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite5′-O-(4,4'-dimethoxytrityl)-2′-deoxyuridine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
FormulaC41H52N5O7PC47H54N5O8PC40H47N6O7PC39H47N4O8P
Molecular weight757.85 g/mol847.95 g/mol754.8 g/mol730.8 g/mol
CAS #195535-80-9105931-57-5141684-35-7109389-30-2
Solution clarityFree from undissolved particulate at 0.2 M in ACNNA
HPLC purity≥98.0%≥98.0%≥98.0%≥98.0%
31P NMR purity≥98.0%≥98.0%≥98.0%≥98.0%
Water contentNA≤0.3% 
Shipping / storage conditionAmbient / –20°C
Material originNonanimal source (no TSE/BSE risk)
Compatible products by synthesizer model
Structural phosphoramidites for duplex stability are compatible with the following oligonucleotide synthesizers:
Applied Biosystems and AKTA synthesizer
Compatible catalog product is ready-to-ship in amber bottles with serum vial neck style.
Expedite synthesizer
Available catalog SKU is supplied in 30 mL amber bottle with a 20-400 finish neck style.
N6-methyl-dA, 8-oxo-dG, and reverse abasic phosphoramidites for mutagenesis studies
Mutagenic effects from methylation of exocyclic amine can be studied by incorporation of N6-methyl deoxyadenosine (N6-Me-dA) into the oligonucleotide sequence. Similar studies may be performed with 8-oxo-2'-deoxyguanosine (8-oxo-dG), which mimics an oxidized guanine, and reverse abasic, which introduces abasic sites.
Availability by compatible synthesizer model
Common applicationMutagenesis studies
 N6-Me-dA CEP8-oxo-dG CEPReverse abasic CEP
Applied Biosystems/AKTA SynthesizerCustom orders available
Request quote
27-1424-04Custom orders are available
Request quote
Expedite Synthesizer
Request quote
Technical specifications for N6-methyl-dA, 8-oxo-dG, and reverse abasic phosphoramidites
Thermo Scientific N6-Me-dA, 8-oxo-dG, and reverse abasic phosphoramidites are used to synthesize oligonucleotides with base modifications for mutagenesis studies.
 N6-Me-dA CEP8-oxo-dG CEPReverse abasic CEP
Cat. No.27-174627-142427-1998
Amidite description5′-O-(4,4'-dimethoxytrityl)‑N6-methyl-2′-deoxyadenosine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite5′-O-(4,4'-dimethoxytrityl)‑N2-isobutyryl-8-oxo-deoxyguanosine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite3-O-(4,4’-dimethoxytrityl)-1,4-anhydro-2-deoxy-D-ribitol-5-O-(β-cyanoethyl-N,N-diisopropyl)
FormulaC41H50N7O6PC44H54N7O9P
C35H45N2O6P
Molecular weight767.9 g/mol855.9 g/mol620.72 g/mol
CAS #105931-58-6143060-53-1NA
Solution clarityFree from undissolved particulate at 0.1 M in ACNFree from undissolved particulate at 0.2 M in ACN
HPLC purity≥98.0%≥98.0%
≥98.0%
31P NMR purity≥98.0%≥98.0%≥98.0%
Water content NA≤0.3%≤0.5%
Shipping / storage conditionAmbient / –20°CDry ice / –20°C
Material originNonanimal source (no TSE/BSE risk)
5′-O-(4,4'-Dimethoxytrityl)
Compatible products by synthesizer model
Structural phosphoramidites for duplex stability are compatible with the following oligonucleotide synthesizers:
Applied Biosystems and AKTA synthesizer
Compatible catalog product is ready-to-ship in amber bottles with serum vial neck style.
Catalog # Name Size Price (USD)
27-1424-04 8-oxo-dG Phosphoramidite, standard grade, serum vial bottle Each
445.00
Looking for additional size options, customization, or packaging solutions?
Connect with us
Resources
Phosphoramidite Learning Resource
Access webinars, videos, articles, and more to optimize your success in oligonucleotide synthesis for diagnostic and therapeutic manufacturing process development.
mRNA Production Learning Library
Browse the educational assets related to raw materials selection, process development using in vitro transcription, scale-up synthesis, and the latest trends in mRNA production.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments