Harness the power of dPCR for precise and sensitive absolute quantification
Digital PCR (dPCR) is a leading technology for detection and quantification of rare mutations and target sequences. Overcome challenges with other ddPCR and dPCR platforms and leverage the full power of dPCR with the Applied Biosystems QuantStudio Absolute Q Digital PCR System, featuring:
  • Broad application support backed by out TaqMan Assay portfolio
  • Proprietary technology with industry-leading accuracy and consistency
  • Super-simple workflow with fast time to answer
  • Modular automation setup for flexible high-thoughput workflows
QuantStudio Absolute Q  Digital PCR System
QuantStudio Absolute Q Digital PCR System
Discover powerfully simple dPCR technology
QuantStudio Absolute Q AutoRun Suite
QuantStudio Absolute Q AutoRun Suite
Explore high-throughput automation for dPCR
digital-pcr-absolute-accelerators
Discover how researchers across the globe are using dPCR to help elevate their research
dPCR vs qPCR—What is the difference?
Video Player is loading.
Current Time 0:00
Duration 1:34
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:34
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Both digital PCR (dPCR) and real-time PCR (qPCR) can be used to detect and quantify nucleic acids. While these technologies share similarities, key differences in quantification methods confer each tool with application-dependent benefits or limitations.
    Prefer to read it? Download the infographic ›
    Learn more about dPCR basics ›
    Applications for dPCR
    With our Absolute Q Universal DNA dPCR Master Mix, explore a vast array of ready to test dPCR applications backed by an extensive portfolio of TaqMan probe based assays. Ready to get started? See if we have the assay you are looking for.
    Search pre-designed TaqMan Assays ›
    Explore dPCR assays ›
    Copy number variation
    Cell and gene therapy
    Wastewater & environmental surveillance
    Gene editing and more
    Rare mutation detection
    Liquid biopsy monitoring
    NGS library quantification & verification
    Explore dPCR applications
    The new dPCR: microfluidic array plate technology
    Not all dPCR is created equal. Our microfluidic array plate (MAP) technology offers significant benefits over droplet digital PCR (ddPCR) technologies, including:
    • Superior consistency and accuracy
    • Reduced dead volume (<5%)
    • Simpler workflow
    • Faster time to answer
    3-panel graphic showing different scale images of the microfluidic array plate for the QuantStudio Absolute Q Digital PCR System
    Microfluidic array plate for the QuantStudio Absolute Q Digital PCR System
    A simplified dPCR workflow to get answers faster
    Video Player is loading.
    Current Time 0:00
    Duration 1:26
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 1:26
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Getting started with the QuantStudio Absolute Q dPCR System is simple. It takes under five minutes to prepare a MAP16 dPCR plate and about two hours or less to get your results.
      Learn more about the QuantStudio Absolute Q system ›
      Absolute Q Digital PCR systems, assays, and reagents
      Image of the QuanStudio Absolute Q Digital PCR System over graphic background
      QuantStudio Absolute Q Digital PCR System
      Explore our powerfully simple dPCR platform powered by proprietary microfluidic array plate technology.
      Learn more ›
       Image of the automated QuanStudio Absolute Q AutoRun Suite for dPCR over graphic background
      QuanStudio Absolute Q AutoRun Suite
      Discover our flexible solution for automated, high throughput dPCR with a walk-away workflow.
      Learn more ›
      Graphic depicting the components of a TaqMan probe-based assay for digital PCR
      Digital PCR assays & kits
      We offer TaqMan probe-based assays and kits supporting a broad range of applications. Choose from wet-lab verified Absolute Q assays, custom assays, kitted solutions, and more.
      Viral titer assays
      Liquid biopsy assays
      Wastewater surveillance assays
      See all assays & kits ›
      ay plates, and other consumables for the QuantStudio Absolute Q Digital PCR system
      Digital PCR reagents & consumables
      Find master mixes and microfludic array plates and acessories for use with the QuantStudio Absolute Q dPCR System.
      Absolute Q Universal DNA dPCR Master Mix
      Absolute 1-step RT dPCR Master Mix
      QuantStudio Absolute Q MAP16 Plate Kit
      See all products ›
      Resources
      Absolute Gene-ius
      Digital PCR Content Library  
      Explore webinars & events, podcasts, publications, technical literature, and more.
      Browse library ›
      Illustration depicting a gloved hand holding a microcentrifuge tube in from of a qPCR and dPCR graphic
      Digital PCR Education Center  
      Review dPCR basics and learn more about the technology and its applications.
      Learn more ›
      Scientist in lab preparing dPCR automated run
      Talk to a specialist
      Have questions about implementing dPCR in your lab? Let us help you get started!
      Contact us ›
      For Research Use Only. Not for use in diagnostic procedures.
      Style Sheet for Global Design System
      Style Sheet for Komodo Tabs