blue-yellow-showcase
A46113-showcase
Easy-to-use and flexible gene expression master mix for real-time pcr
Features of PowerTrack SYBR Green Master Mixes include:
  • Two-color tracking dye system indicates where pipetting has occurred
  • Broad primer Tm and primer concentration compatibility allows flexibility in qPCR reaction setup with minimal optimization
  • Superior specificity and tight reproducibility in Ct values over a broad dynamic range improve data quality
  • Compatible with SuperScript IV VILO master mix reverse transcription for fast, reproducible results
  • Formulated with UNG/dUTP to prevent contamination of carryover PCR products
  • Broad instrument compatibility
“It is a wonderful product and so easy to use. Everybody was excited in the lab and feels that it will make life easier.”
Tariq M. Haqqi, M.Phil., Ph.D., Professor
Department of Anatomy & Neurobiology
Northeast Ohio Medical University
Tariq-Haqqi-scientist
Tracking dye helps reduce pipetting errors
See how it works
Video Player is loading.
Current Time 0:00
Duration 0:15
Loaded: 65.33%
Stream Type LIVE
Remaining Time 0:15
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    PowerTrack SYBR Green Master Mix includes a two-color dye tracking system to indicate where pipetting has occurred. When the yellow sample is pipetted into the well containing the blue master mix, the well turns green. Extensive testing was done to verify the tracking dye doesn’t impact data quality.
    See how PowerTrack SYBR Green Master Mix performs versus other leading suppliers ›
    Formulated for superior specificity and reproducibility
    PowerTrack SYBR Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR. It contains antibody-mediated Taq DNA polymerase with a hot-start mechanism that provides tight control over Taq enzyme activation and helps prevent undesirable early activity of the polymerase at low temperatures that can lead to nonspecific amplification. Building on over 20 years of innovation and product excellence in qPCR, PowerTrack SYBR Green Master Mix is designed for exceptional performance in your most challenging real-time PCR applications.
    In an evaluation of 24 different primer sets used with PowerTrack SYBR Green Master Mix, a single melt curve was obtained in 100% of reactions. In contrast, nonspecific amplification was observed for some of the same targets with several master mixes from other suppliers, as shown by multiple peaks in the melt curves (Figure 1). Validation of primer specificity in SYBR Green reactions is essential to data quality and validity [1]. The high specificity enabled by PowerTrack SYBR Green Master Mix allows you to spend less time optimizing and redesigning primers to get high quality data.
    Figure 1. Target specificity. Melt curve analyses for PGK1 (phosphoglyerate kinase 1) , ARL1 (ADP-ribosylation factor-like protein 1), and SNF8 (vacuolar-sorting protein) targets were run on the QuantStudio 5 Flex Real-Time PCR System with nine different master mixes. Each master mix manufacturer’s recommended cycling conditions were used with each mix on a 6-log dilution series of human universal reference cDNA. UHR cDNA was run with assays PGK1, ARL1, and SNF8. 10-µL reactions were run in quadruplicate on QuantStudio 5 384-well block real-time PCR instruments. Several competitors show amplification of non-specific product attributed to second peak in melt curve analysis.
    Competitor Q1: Qiagen QuantiNova SYBR® Green PCR Kit; Competitor B: Bio-Rad SsoAdvanced™ Universal SYBR® Green Supermix; Competitor S: BioLine (Meridian Bioscience) SensiFAST™ SYBR® Lo-ROX Kit; Competitor Q2: Qiagen QuantiFast SYBR Green PCR Kit; Competitor QB: Quanta Biosciences PerfeCTa® SYBR® Green FastMix® Low ROX Reaction Mix; Competitor K: KAPA SYBR FAST qPCR kits, Low ROX; Competitor T: Takara TB Green® Premix Ex Taq™ II; Competitor P: ProMega GoTaq® qPCR Master Mix.
    PowerTrack SYBR Green Master Mix powers through traditionally difficult targets
    Amplification curves were obtained for the PGK1 gene over a 5-log dilution series of UHR cDNA.  PowerTrack SYBR Green Master Mix delivers accurate results over a wide dynamic range of concentrations as shown by tight curves between replicates and superior PCR efficiency (Figure 2).
    Figure 2.  Reactions were run in quadruplicate on the QuantStudio 5 Real-Time PCR System with the 384-well block according to protocol.  10-µL reactions were run in quadruplicate using 60ºC annealing primer Tm. PGK1 = high expressor; ARL1, IGF2, and TCF25 = medium expressor.
    Excellent reproducibility
    Reproducibility is another important measure of data quality in real-time PCR, and reproducibility often declines at low template concentrations, where the impacts of variability are exacerbated. However, PowerTrack SYBR Green Master Mix demonstrated excellent reproducibility over a wide dynamic range with a variety of targets and RT kits tested (Figure 3). Tighter reproducibility allows for greater statistical significance when analyzing low-abundance transcripts and smaller fold changes.
    std-dev-vs-mean-chart
    Figure 3. Reproducibility of data. Six assays (PGK1, ARL1, SNF8, DF, GAPDH, and Corf1 19) were run in quadruplicate with UHR cDNA generated from four different reverse transcription (RT) kits (SuperScript IV VILO, High Capacity RNA to cDNA, iScript cDNA synthesis, and Quantitect RT kits) run with 6-fold dilution series with 400 nM primer concentration on the QuantStudio 5 Flex Real-Time PCR System according to each master mix manufacturer’s recommended protocol.
     View the complete data set
    Broad instrument compatibility
    PowerTrack SYBR Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems real-time PCR instruments. It is also compatible with the Bio-Rad CFX96, CFX384, and iQ5; Roche LightCycler 480; and Agilent Mx3005P systems.  For optimal results, primer concentrations should be in the range of 300–800 nM.
    ROX dye for improved data precision
    ROX dye is an inert reference dye used in RT-qPCR that is included in our master mixes.  It is effective in normalizing fluorescence across all samples.  It removes fluorescence variations, such as those caused by bubbles, from the data.  Applied Biosystems master mixes contain a proprietary ROX dye specifically formulated for a wide range of real-time PCR instruments and compatible with a wide range of instrument light sources and filter sets.
    UNG for carryover contamination control
    Contamination in labs that routinely run PCR is a major concern for many researchers and is the source of most false positives. The inclusion of UNG and dUTP in the PowerTrack SYBR Green Master Mix allows any previously amplified PCR products to be degraded and helps prevent contamination of subsequent qPCR reactions. Utilization of a heat-labile UNG enzyme allows for effective contamination control.
    Ordering information
    Catalog # Name Size Price (USD) Qty
    A46012 PowerTrack™ SYBR Green Master Mix for qPCR, 1 mL Each
    119.65
    Online Exclusive
    127.00
    Save 7.35 (6%)
    A46109 PowerTrack™ SYBR Green Master Mix for qPCR, 5 mL Each
    509.65
    Online Exclusive
    546.00
    Save 36.35 (7%)
    A46110 PowerTrack™ SYBR Green Master Mix for qPCR, 2 x 5 mL Each
    935.65
    Online Exclusive
    1,002.00
    Save 66.35 (7%)
    A46111 PowerTrack™ SYBR Green Master Mix for qPCR, 5 x 5 mL Each
    2,251.65
    Online Exclusive
    2,400.00
    Save 148.35 (6%)
    A46112 PowerTrack™ SYBR Green Master Mix for qPCR, 10 x 5 mL Each
    4,310.65
    Online Exclusive
    4,615.00
    Save 304.35 (7%)
    A46113 PowerTrack™ SYBR Green Master Mix for qPCR, 50 mL Each
    4,296.65
    Online Exclusive
    4,600.00
    Save 303.35 (7%)
    Reference
    1. Bustin SA, Benes V, Garson JA, Hellemans J, Huggett J, Kubista M, Mueller R, Nolan T, Pfaffl MW, Shipley GL, Vandesompele J, Wittwer CT. The MIQE guidelines: minimum information for publication of quantitative real-time PCR experiments. Clin Chem 2009 55(4):611-22.
    For Research Use only. Not for diagnostic purposes.
    This offer is available to customers in the US (excluding Puerto Rico) and Canada (excluding Quebec) who purchase at least one 5 ML bottle or above of PowerTrack SYBR Green Master Mix (Cat No. A46109, A46110, A46111, A46112, A46113). Offer valid until November 30, 2021, or until promotional t-shirts run out, whichever comes first. Limit one t-shirt per customer. Please note, it can take up to two weeks to receive your t-shirt. Applied Biosystems will fulfill t-shirt size requests whenever possible, but reserves the right to substitute sizes if necessary. Health care professionals may not participate in this promotion. Orders placed through fishersci.com or Fisher Scientific channel are not eligible. By submitting a promotional item request, you warrant that you are not prohibited by employment, contract, or law from receiving a promotional item from Applied Biosystems. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.