SYBR™ Select Master Mix offers advanced performance at an affordable price.
  • Specific - minimize primer-dimer and nonspecific amplification with a highly purified DNA polymerase with a proprietary hot start mechanism
  • Reproducible and sensitive - consistent amplification across a wide dynamic range
  • Bright - contains SYBR™ GreenER™ dye for maximum brightness
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility - compatible on most Real-Time PCR instruments
Ordering Information
Catalog # Name Size Price (USD) Qty
4472903 SYBR™ Select Master Mix, 1 x 1 mL Each
126.65
Online Exclusive
133.00
Save 6.35 (5%)
4472908 SYBR™ Select Master Mix, 1 x 5 mL Each
532.65
Online Exclusive
556.00
Save 23.35 (4%)
4472918 SYBR™ Select Master Mix, 2 x 5 mL Each
1,025.65
Online Exclusive
1,070.00
Save 44.35 (4%)
4472919 SYBR™ Select Master Mix, 5 x 5 mL Each
2,366.65
Online Exclusive
2,470.00
Save 103.35 (4%)
4472920 SYBR™ Select Master Mix, 10 x 5 mL 10 x 5 mL
4,516.65
Online Exclusive
4,715.00
Save 198.35 (4%)
4472913 SYBR™ Select Master Mix, 1 x 50 mL Each
4,483.65
Online Exclusive
4,680.00
Save 196.35 (4%)
SYBR Select Master Mix outperforms the competition with respect to:
  • Specificity - specific amplification for 100% of assays
  • Brightness - optimum brightness for high performance on most Real-Time PCR instruments
  • Dynamic range - more tolerant to high input cDNA
  • Sensitivity - true single copy detection as demonstrated with digital PCR
Exceptional specificity
Out of 44 assays and 3 independent replicate reactions only SYBR Select Master Mix had a single peak in all the melt curves (100% specificity).
Melt Curve
Click image to enlarge
Figure 1: Percentage of assays with Single Peak in Melt Curve
Spectacular dynamic range and linearity
SYBR Select produces no NTC signal and is more tolerant to high-input DNA.
NTC signal
Click image to enlarge
Figure 3: Dynamic range and linearity shown for a 7-log dilution for PGK1 assay
Brightness that is just right
SYBR Select is brighter than the majority of competitor master mixes, but not too bright to upset some Real-Time PCR instruments.
Average delta Rn
Click image to enlarge
Figure 2: Average delta Rn (relative fluorescence) across 4 dilution points for PGK1 assay
Single copy detection
SYBR Select allows single copy detection using dPCR and gDNA with RNaseP target.
single copy detection
Click image to enlarge
Figure 4: The single copy RNase P gene was amplified using genomic DNA diluted to <1 copy per reaction. Then, a normal distribution of the CT values was obtained