Surface Analysis Using X-Ray Photoelectron Spectroscopy
What is surface analysis?
Increasing demand for high-performance materials has made surface engineering crucial. The surface of a material is its interface with the environment and other materials, affecting factors such as corrosion rates, catalytic activity, adhesive properties, wettability, contact potential, and appearance. These characteristics can be tailored for desired capabilities through surface modification.
The surface layer has physical and chemical properties distinct from the bulk material, or remainder of the solid. An atom in the surface layer is not surrounded by atoms on all sides, resulting in higher reactivity and bonding potential than bulk atoms.
Surface analysis is used to quantify the surface chemistry and through that, understand the surface properties. Surface modification can alter or enhance these characteristics, with efficacy checked through surface analysis.
surface analysis
Customer Testimonial: University of Versailles
In this in-depth video, the University of Versailles shares their passion for materials science. Arnaud Etcheberry, Emeritus Director at CNRS, delves into his expertise in merging chemistry, physics of materials, and surface analysis. Mathieu Fregnaux, a research engineer at CNRS, specializes in photoemission, particularly XPS. They boast almost the entire XPS catalog from Thermo Fisher Scientific and were the pioneers in France to adopt Thermo Scientific XPS instruments. They have witnessed the complete development and evolution of our tools.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    How can X-ray photoelectron spectroscopy support surface analysis?
    X-ray photoelectron spectroscopy (XPS), or electron spectroscopy for chemical analysis (ESCA), is a common surface characterization tool used for a huge range of applications, from the every day (e.g. waterproofing fabrics or non-stick cookware coatings), to advanced R&D (e.g. clean energy materials, organic thin-film electronics, and bio-active surfaces). XPS measures the elemental composition, chemical, and electronic state of atoms on a material's surface to analyze its surface chemistry.
    XPS offers a significant advantage in detecting subtle changes in the position of peaks that reflect the chemical state of surface elements, such as metallic or oxidized states, and diverse bonding states in polymers. The other key advantage to XPS is that the analysis depth is limited to a few nanometers due to the strong electron-matter interactions. Electrons lose energy quickly as they interact with matter, preventing their detection as part of a peak. This limits the depth from which the signal is detected to around 10 nm, depending on the material, which makes XPS extremely surface sensitive.
    Schematic of an XPS instrument. X-rays are created and monochromated to ensure good energy resolution before irradiating the sample. The created photoelectrons are focused into an analyzer and counted by a detector to create a spectrum. Schematic of an XPS instrument. X-rays are created and monochromated to ensure good energy resolution before irradiating the sample. The created photoelectrons are focused into an analyzer and counted by a detector to create a spectrum.
    X-Ray photoelectron spectroscopy instruments
    Thermo Scientific K-Alpha X-ray Photoelectron Spectrometer System
    K-Alpha X-ray Photoelectron Spectrometer System
    Thermo Scientific Nexsa G2 Surface Analysis System
    Nexsa G2 Surface Analysis System
    Thermo Scientific ESCALAB QXi X-ray Photoelectron Spectrometer Microprobe
    ESCALAB QXi X-ray Photoelectron Spectrometer Microprobe