Nuclease Grade Source Cleavage Specificity MW
(kD)
pH
Optimum
Required
Cofactors
Comments
RNase A Affinity pure
RPA
RNA
Bovine 3' of ss C's and U's 13.77.0-7.5NoneInactivated by 0.1% SDS (1)
RNase I MB*
E. coli
(recombinant)
3' of ss NTPs of RNA 27.2  NoneResponsible for 98% of the EDTA-resistant RNase activity in E. coliextracts (2)
RNase T1 MB*
RNA
A. oryzae
(recombinant)
3' of ss G's 11.17.5NoneSpecificity for G is 106X greater than A (3)
RNase V1 RNA
Cobra venom dsRNA 15.97.5Na+
(100 mM)
Mg2+
(0.3 mM)
Cleaves ssRNA at 10% the rate of dsRNA (1)
RNase H MB*
E. coli
(recombinant)
RNA of RNA:DNA hybrids to give 5'-P oligonucleotides17.67.5-9.1Mg2+
(2–4 mM)
EDTA abolishes activity, but enzyme can still bind RNA:DNA hybrids; Not inhibited by vanadate (3)
DNase I RNase-free
(see also
DNA-free)
Bovine dsDNA to yield 5'-P dinucleotides and 5'-P oligonucleotides 29.1 7.0-8.0Mg2+
(2–5 mM)
Ca2+
(0.1–1 mM)
Cleaves ssDNA 500X more slowly than dsDNA (3,4)
*MB = Molecular Biology Grade
References
  1. White, J.S. & White, D.C. (1997) Source Book of Enzymes, CRC Press, Boca Raton, FL.
  2. Zaniewski, R. et al. (1984) J Biol Chem 259: 11651-11653.
  3. Eun, H-M. (1996) Enzymology Primer for Recombinant DNA Technology, Academic Press, Inc., San Diego, CA.
  4. G. Latham, unpublished data.
Ordering Information
Catalog # Name Size Price (USD) Qty
AM1906 DNA-free™ DNA Removal Kit Each
187.00
AM2222 Ambion™ DNase I (RNase-free), 2000 U Each
291.65
Online Exclusive
316.00
Save 24.35 (8%)
AM2224 Ambion™ DNase I (RNase-free), 5 x 2000 U Each
693.65
Online Exclusive
752.00
Save 58.35 (8%)
AM2270 Ambion™ RNase A, affinity purified, 1 mg/mL, 200 μg Each
122.65
Online Exclusive
135.00
Save 12.35 (9%)
AM2271 Ambion™ RNase A, affinity purified, 1 mg/mL, 1 mg Each
381.65
Online Exclusive
419.00
Save 37.35 (9%)
AM2283 RNase T1 (1 U/μL) Each
226.00
AM2286 RNase Cocktail™ Enzyme Mix, 1 mL Each
138.65
Online Exclusive
147.00
Save 8.35 (6%)
AM2288 RNase Cocktail™ Enzyme Mix, 5 x 1 mL Each
380.65
Online Exclusive
435.00
Save 54.35 (12%)
AM2292 Ribonuclease H, from E. coli (cloned) 10 U/μL, 200 U Each
145.65
Online Exclusive
155.00
Save 9.35 (6%)
AM2293 Ambion™ RNase H, from E. coli, 10 U/μL, 1000 U Each
379.65
Online Exclusive
408.00
Save 28.35 (7%)
AM2294 Ambion™ RNase I, cloned, 100 U/μL, 10,000 U Each
196.65
Online Exclusive
216.00
Save 19.35 (9%)
AM2295 Ambion™ RNase I, cloned, 100 U/μL, 25,000 U Each
395.65
Online Exclusive
437.00
Save 41.35 (9%)