Multicolor fluorescence image of mouse testis cross section.
Multicolor fluorescence image of mouse testis cross section. Spermatozoa were detected using the green-fluorescent Alexa Fluor® 488 conjugate of lectin PNA (Cat. no. L21409). Actin filaments were visualized using red-fluorescent Alexa Fluor® 568 phalloidin (Cat. no. A12380). Nuclei were stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490).
Multicolor fluorescence image of mouse testis cross section. Spermatozoa were detected using the green-fluorescent Alexa Fluor® 488 conjugate of lectin PNA (Cat. no. L21409). Actin filaments were visualized using red-fluorescent Alexa Fluor® 568 phalloidin (Cat. no. A12380). Nuclei were stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490).
Related Products
Catalog # Name Size Price (USD) Qty
A12380 Alexa Fluor™ 568 Phalloidin, Orange-red, 578/600 nm Each
628.65
Online Exclusive
660.00
Save 31.35 (5%)
L21409 Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 488 Conjugate Each
222.00
Related Images