A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin (Cat. no. A22281), an anti–a-tubulin antibody (Cat. no. A11126) and an anti–cdc6 peptide antibody (Cat. no. A21286). The anti–a-tubulin antibody was prelabeled with the Zenon® Alexa Fluor® 488 Mouse IgG1 Labeling Kit (Cat. no. Z25002) and the anti–cdc6 peptide antibody was prelabeled with the Zenon® Alexa Fluor® 647 Mouse IgG1 Labeling Kit (Cat. no. Z25008).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin (Cat. no. A22281), an anti–a-tubulin antibody (Cat. no. A11126) and an anti–cdc6 peptide antibody (Cat. no. A21286). The anti–a-tubulin antibody was prelabeled with the Zenon® Alexa Fluor® 488 Mouse IgG<sub>1</sub> Labeling Kit (Cat. no. Z25002) and the anti–cdc6 peptide antibody was prelabeled with the Zenon® Alexa Fluor® 647 Mouse IgG<sub>1</sub> Labeling Kit (Cat. no. Z25008).
Related Products
Catalog # Name Size Price (USD) Qty
A11126 alpha Tubulin Monoclonal Antibody (236-10501) 50 µg
424.00
A22281 Alexa Fluor™ 350 Phalloidin, Blue, 346/442 nm Each
640.65
Online Exclusive
660.00
Save 19.35 (3%)
Related Images