Sandmeyer reaction—a free radical reaction
In 1884, Swiss chemist Traugott Sandmeyer attempted to synthesize phenylacetylene from benzenediazonium chloride and copper(I) acetylide. However, the main product he isolated was chlorobenzene, and no trace of the desired product was found. On careful examination of the reaction, it was discovered that copper(I) chloride was formed in situ, which catalyzed the replacement of the diazonium group with a chlorine atom. Following this discovery, the substitution of aryldiazonium salts with halides or pseudo halides became known as the Sandmeyer reaction.
The required aryldiazonium halides are generally prepared from the corresponding arylamines via diazotization using either sodium nitrite and aqueous hydrochloric acid, or alkyl nitrites such as tert-butyl nitrite under anhydrous conditions. These aryldiazonium compounds are prepared and reacted in situ with copper(I) chloride, bromide, or cyanide to obtain the corresponding aryl halide or nitrile.
The Sandmeyer reaction has been used in many successful synthesis campaigns, including the preparation of the anti-psychotic drug flupentixol as well as neoamphimedine, a compound that has anti-cancer properties.
Review available Thermo Scientific products for the Sandmeyer reaction:
Catalog # Name Size Price (USD) Qty
A18668.0B Sodium nitrite, 98% Each
66.65
Special offer
Online exclusive
Ends: 30-Jun-2025
78.00
Save 11.35 (15%)
A18668.0I Sodium nitrite, 98% Each
187.65
Special offer
Online exclusive
Ends: 26-Sep-2025
220.00
Save 32.35 (15%)
11871.36 -
41837.22 -
14644.06 -
14644.14 -
A12704.18 Potassium iodide, 99% Each
27.65
Special offer
Online exclusive
Ends: 26-Sep-2025
32.60
Save 4.95 (15%)
A12704.30 Potassium iodide, 99% Each
77.65
Online Exclusive
86.00
Save 8.35 (10%)
A12704.0B Potassium iodide, 99% Each
227.65
Special offer
Online exclusive
Ends: 26-Sep-2025
268.00
Save 40.35 (15%)
A12704.0I Potassium Iodide, 99% Each
940.65
Online Exclusive
1,045.00
Save 104.35 (10%)
11601.22 -
42857.14 -
42857.22 -
Vote for your favorite name reaction
Learn more about the most popular named reactions in chemistry based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now