Innovative, robust, and easy-to-use cryo-TEM
Our full cryo-TEM portfolio features state-of-the-art technology with a range of automation features designed to extend accessibility, reduce the need for user intervention, and enable easy organization, viewing, and sharing data. We offer integrated solutions and support from sample preparation to data analysis for multiple cryo-TEM methods, including single particle analysis, MicroED, and cryo-tomography. Rely on our long-term ongoing support, as well as flexible financing options to help you bring the right cryo-TEM solution into your lab.
Tundra Cryo TEM
Epitope mapping
Accessible and smart
  • Easy, iterative loading and imaging for rapid determination of sample viability
  • AI-guided automation with results that are progressively displayed
  • Cost-effective platform for labs that are new to cryo-EM.
  • Intermediate-resolution single particle analysis: 100 kV, <3.5 Å* 
  • Medium throughput: Dataset in 24 hours 
  • Sample type: Proteins 
  • Applications: Single particle analysis
Glacios 2 Cryo-TEM
H₂O molecules and model building
Capable and versatile
  • Automated sample assessment and acquisition of large data sets for higher throughput
  • Improved detector and AI-enabled software work together to provide rapid, high-quality results
  • High-resolution single particle analysis: 200 kV, <2.5 Å* 
  • High throughput: Dataset in 30 minutes 
  • Sample type: Proteins, crystals, cells 
  • Applications: Single particle analysis, MicroED, cryo-tomography
Krios 5 Cryo-TEM
H atoms, ligand identification, interaction
Powerful and productive
  • Ultimate productivity with fast, high-quality data collection
  • High workflow efficiency for cryo-tomography and single particle analysis
  • Ultra high-resolution single particle analysis: 300 kV, <1.5 Å* 
  • Highest throughput: Dataset in minutes 
  • Sample type: Proteins, crystals, cells 
  • Applications: Single particle analysis, MicroED, cryo-tomography
* Based on best published performance, actual results will depend on non-microscope factors such as sample and user experience. Not a promise of biological resolution performance.
Electron microscopy service and support
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Cryo-EM software solutions
Simplify your cryo-TEM acquisition and analyses with innovative software solutions. Each solution features user-friendly interfaces that enable intelligent and automated cryo-TEM data acquisition and analysis to streamline workflows for multiple cryo-TEM methods, including single particle analysis, cryo-tomography, and MicroED.
CryoFlow Software
Thermo Scientific CryoFlow Software, our paperless cryo-EM data management solution, optimizes cryo-EM workflows by helping you generate protein structures using single particle analysis (SPA) and perform cellular structural analysis using cryo-electron tomography (cryo-ET). Its modular organization provides a single convenient location to track experiments through individual steps in SPA and cryo-ET workflows.
Smart EPU Software
Thermo Scientific Smart EPU Software is an embedded software for single particle acquisition. Its simplified user interface, advanced automation, and screening capabilities allow for straightforward planning and execution of experiments.
Tomography 5 Software
Thermo Scientific Tomography 5 Software enables acquisition of 3D data, even on your most challenging vitrified samples. Its user-friendly interface enables both experienced and new users to jump-start their tomography data acquisition. Tomography Software and Tomo Live Software are supported on the Krios, and Glacios cryo-TEMs.
EPU-D Software
Thermo Scientific EPU-D Software is electron diffraction acquisition software. It enables quick switching between imaging and diffraction, guides optimal low-dose setup, and drives the continuous tilt needed for data collection.
Embedded CryoSPARC Live
Embedded CryoSPARC Live significantly shortens time to results by providing real-time, multi-dimensional feedback on sample quality and TEM settings for single particle analysis.
Amira Software
Thermo Scientific Amira Software is a 2D–5D solution for visualizing and analyzing data from many image modalities, including Optical and Electron Microscopy, CT, MRI and other imaging techniques.
Cryo-TEM sample preparation solutions
Vitrobot Mark IV System
The Thermo Scientific Vitrobot Mark IV System automates the vitrification process to provide fast, easy, reproducible sample preparation—the first step in obtaining high-quality images and repeatable experimental results.
Cryo-FIB sample preparation instruments
Cryo-focused ion beam milling microscopes for the preparation of high-quality cryo-lamellae and volume imaging.
Cryo-TEM accessories
Selectris and Selectris X Imaging Filters
Designed for stability and speed, Thermo Scientific Selectris and Selectris X Imaging Filters are post-column imaging filters that improve the contrast of TEM images, resulting in high-resolution structures up to atomic resolution. Paired with the latest generation Thermo Scientific Falcon 4i Direct Electron Detector, Selectris Filters enable you to obtain high-resolution structures quickly, revealing new biological insights. To further increase the productivity and ease of use of your cryo-EM research, these filters are fully integrated into Thermo Scientific EPU SoftwareThermo Scientific Tomography Software, and instrument operation software.
Falcon 4i Direct Electron Detector
The Thermo Scientific Falcon 4i Direct Electron Detector, with optical and throughput enhancement modes in combination with a more stable stage, accelerates automated data acquisition. Aberration-free image shift (AFIS) functionality uses image beam shifts, eliminating the need to perform mechanical stage movements. Fringe-free imaging (FFI) ensures the beam is only collecting data on usable parts of the sample, improving the quality of the results. When configured with the Selectris X Imaging Filter, the capability to use a very stable narrow slit width (<10 eV) provides an additional boost in contrast for single particle cryo-EM and cryo-ET, improving resolution and obtaining results faster with less data to maximize your resolution efforts.
Ceta-D Camera
The Thermo Scientific Ceta-D Camera is a scintillator-based camera optimized for low-dose diffraction data collection. It is ideal for working with dose-sensitive materials, such as proteins, pharmaceutical molecules, and organic materials. The camera’s high sensitivity allows for reliable detection of high-resolution, low-intensity diffraction peaks. Its high signal-to-noise ratio allows for accurate measurement of the integrated peak intensity. Both of these elements are prerequisites for obtaining high-resolution structural information.
Ceta-M Camera
The Thermo Scientific Ceta-M Camera delivers sensitive and dynamic imaging with a large field of view. This unique combination follows the natural workflow in TEM imaging: from fast navigation to find the area of interest, to easy optimization of the image quality via optical adjustments, to the final result, a 4k × 4k image unrivaled in quality and detail.
Heat decontamination for cryo-TEM
Our solution for 60°C heat decontamination allows installation of the Thermo Scientific Krios Cryo-Transmission Electron Microscope (Cryo-TEM) in higher biosafety-level containment facilities (e.g. BSL-3). For these installations, we offer a 60°C heat decontamination solution consisting of both hardware and software to heat the interior of the microscope enclosure.
Looking to find a cryo-EM facility near you?
There are nearly 300 world-class cryo-EM facilities where you can advance your research by obtaining cryo-EM structures and proof-of-concept data using your own samples. Most facilities have an application form that will guide you through proposal requirements. Although these vary by facility, generally you can expect to submit a research proposal for independent scientific and technical evaluation based on:
  • Scientific background and project significance
  • Technical suitability and feasibility, including descriptions of the sample and sample quality
Individual facilities also offer different levels of service, which may include sample preparation, grid screening, and cryo-EM training.
For Research Use Only. Not for use in diagnostic procedures.