The Thermo Scientific Ceta-D Camera is a scintillator-based camera optimized for low-dose diffraction data collection. It is ideal for working with dose-sensitive materials, such as proteins, pharmaceutical molecules, and organic materials.
The camera’s high sensitivity allows for reliable detection of high-resolution, low-intensity diffraction peaks, and its high signal-to-noise ratio enables accurate measurement of the integrated peak intensity. Both of these properties are prerequisites for obtaining high-resolution structural information. After acquisition, the diffraction patterns and metadata are readily available for processing (similar to standard X-ray diffraction data), using software packages like DIALS or XDS. Resolutions better than 1 Å have been reported.
MicroED package
Coupled with the Ceta-D Camera, the MicroED Package provides all the necessary elements to perform micro electron diffraction, either on a new instrument or as a retrofit onto an existing tool. It includes:
  • Thermo Scientific EPU-D Software for screening and automated data acquisition. EPU-D Software now expands the intuitive capabilities and unified user experience of Thermo Scientific EPU Software to the electron crystallography domain. 
  • Optimized smaller beam stop to mask the central beam without blocking low-resolution diffraction spots. 
  • Optimized aperture set to maximally suppress unwanted background signals from the area surrounding the crystal. 
  • Dedicated electron-optical settings to enable optimal camera utilization while benefiting from best sample-beam geometry.
The MicroED Package is a perfect match with the CETA-D Camera but may also be combined with other Thermo Scientific cameras for special application requirements.
Proteinase K structured determined by micro electron diffraction.
Proteinase K structured determined by micro electron diffraction. Data provided by the Gonen Laboratory HHMI/UCLA.
Key Features
Ceta-D camera
  • Optimized for low-dose diffraction data collection
  • High sensitivity for reliable detection of high-resolution, low-intensity diffraction peaks 
  • High signal-to-noise ratio for accurate integrated peak intensity
MicroED package
  • Dedicated software for crystal/diffraction screening as well as automated data acquisition
  • Modified beam stop, optimized for micro electron diffraction
  • Optical presets provide optimized camera exposure for MicroED
  • Optimized C2/SA aperture set and 90° rotation projection system (200 kV systems only)
  • Compatible with Thermo Scientific Krios, Glacios, Talos Arctica and Talos F200 Cryo-TEMs. For compatibility with other systems, please consult your sales representative
Specifications
Style Sheet for Products Table Specifications
Sensor4,096 x 4,096, 14 μm pixel CMOS
Camera architecture
Fiber optic coupled scintillator (1:1) frame rate
Standard: 4k × 4k, 2 fps; 2k × 2k, 8 fps; 1k × 1k, 18 fps
Noise reduction: 4k × 4k, 2 fps; 2k × 2k, 6 fps; 1k × 1k, 6 fps
Imaging performanceIn 4k × 4k mode: DQE at 0.1 Nyquist: > 26% at 300 kV, > 40% at 200 kV
Duty cycle in movie mode100% in rolling shutter mode
Conversion efficiency
>26 counts/primary electron at 200 kV
>22 counts/primary electron at 300 kV
Mounting position
On-axis, bottom mounted, retractable
Style Sheet to change H2 style to p with em-h2-header class
Applications
Infectious disease research
Infectious Disease Research
Cryo-EM techniques enable multiscale observations of 3D biological structures in their near-native states, informing faster, more efficient development of therapeutics.
Drug discovery
Drug Discovery
Learn how to take advantage of rational drug design for many major drug target classes, leading to best-in-class drugs.
Style Sheet for Komodo Tabs
Techniques
MicroED
MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
Learn more ›
MicroED
MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
Learn more ›
Contact us
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards