c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
NanoBuilder rapid prototyping software
Thermo Scientific NanoBuilder Software is a tool for the systematic planning and construction of multi-layer nanodevices. This is done by dividing CAD files into ordered projects, facilitating the generation of complex structures that previously were tedious or even impossible to build. NanoBuilder Software allows for accurate patterning of large and complex nanostructures on multiple sites, supporting all DualBeam (focused ion beam scanning electron microscopy, FIB-SEM) patterning processes: FIB milling, gas-assisted FIB milling, FIB induced deposition, and electron-induced deposition.
Key features of NanoBuilder rapid prototyping software
Fully automated
Fully automated job execution, stage navigation, milling, and deposition.
FIB and GIS
Focused ion beam (FIB) and gas injection systems (GIS) optimized.
CAD
CAD-based prototyping.
Alignment
Automated alignment and drift control.
Patterning sequences
Plan patterning sequences on your desktop PC using Thermo Scientific NanoArchitect Software.
Specifications
Style Sheet for Products Table Specifications
GDSTODB
NanoBuilder
Import GDSII files
GDSII viewer
Convert to Thermo Scientific stream files
Automated alignment and drift control
Automated job execution
Multi-site support
Integrated editing of microscope properties
Alignment without exposing the site of interest
Style Sheet for Komodo Tabs
Resources
Applications
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Fundamental Materials Research
Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
Techniques
In Situ experimentation
Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
Learn more ›
Nanoscale Prototyping
As technology continues to miniaturize, the demand for nanoscale devices and structures is ever increasing. 3D nanoprototyping with DualBeam instruments helps you to quickly design, create, and inspect micro- and nanoscale functional prototypes.
Learn more ›
In Situ experimentation
Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
Learn more ›
Nanoscale Prototyping
As technology continues to miniaturize, the demand for nanoscale devices and structures is ever increasing. 3D nanoprototyping with DualBeam instruments helps you to quickly design, create, and inspect micro- and nanoscale functional prototypes.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards