All elements in one measurement
The high light throughput optics combined with the charge injection device detector, the CID821, ensure the shortest possible analysis times. When using the iFR analysis mode, the complete spectrum is captured in one measurement, making the analysis time even shorter.
Advanced performance
For the most demanding applications that require the highest sensitivity in the UV region of the spectrum to determine elements such as mercury, antinomy, and lead, the eUV analysis mode enhances the sensitivity, ensuring regulations for toxic elements are easily met.
Shortest analysis time
Experience high-speed analysis of your trace elemental samples with the Thermo Scientific iCAP PRO XPS ICP-OES. It meets your specific regulatory requirements with unmatched throughput and versatility.
Perform like a PRO with the iCAP PRO Series ICP-OES
The Thermo Scientific iCAP PRO Series ICP-OES provides analysis in the fastest possible time with the assurance that your trace analyses will always be of the highest caliber for accuracy and productivity. Download the iCAP PRO Series ICP-OES information pack to learn how.
Download information pack ›
Semi-quantitative analysis for the iCAP PRO Series ICP-OES
Do you need to screen samples for their elemental composition as part of method development or to inform further analysis? The new semi-quantitative analysis tool within the Qtegra ISDS Software leverages artificial intelligence to enable you to do this in the simplest way. Find out how by reading this product spotlight.
Read now
 iCAP PRO XP ICP-OESCAP PRO XPS ICP-OES
Plasma viewVertical Radial or Vertical Duo
Start-up time5 minutes (from standby)
Wavelength rangeiFR mode 167.021 nm - 852.145 nm
eUV mode 167.021 nm - 240.063 nm
Resolution at 200 nm7 pm
RF source radial and duo750 to 1600 W
Gas flow controlsVariable MFC
Plasma gas0 to 20 L/min
Auxiliary gas0 to 2 L/min
Nebulizer gas0.0 to 1.5 L/min
Additional gas 0 - 0.25 L/min
Sample throughput Medium to highVery high
iSC-65 Autosampler
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how the iSC-65 Autosampler offers exceptional performance in streamlining workflows and sample throughput.
    The Thermo Scientific iSC-65 Autosampler is a workhorse partner of the iCAP PRO Series ICP-OES. It has been designed with reliability from the ground up to ensure all samples are analyzed the first time.
    • Efficiency and productivity: The iSC-65 Autosampler allows sample-specific probe depths, avoiding unnecessary reruns due to blocked sample introduction systems. With the advanced Step Ahead feature, the autosampler probe is sent to rinse while data is still being acquired, using sample in the probe line to complete the analysis. 
    • Durable and robust design: Acid-resistant construction ensures confident sampling as well as longevity and robust operation in laboratory environments.
    • Visual feedback: Through LED lighting, provides clear, easy-to-see visual feedback at a distance on autosampler and analysis run status for technicians in crowded laboratories.
    • Software integration: Native Qtegra ISDS Software plugin allows user to control all automation parameters, including alignment, individual depth locations, pump speed, XYZ movement speeds, and direction of sampling sequences.
    • Unattended analyses: With the advanced quality control capabilities in Qtegra ISDS Software, the iSC-65 Autosampler instills confidence in high throughout, unattended ICP-OES and ICP-MS analyses.
    • Low contamination sample introduction: Utilize the iSC-65 Autosampler with the Thermo Scientific iCAP Qnova Series ICP-MS to eliminate manual sample introduction and the potential of contamination when analyzing ultra-pure high sensitivity samples.
    Keep up with the speed of science
    The Lab-Forward Trade-In Program
    The world needs science to move faster. Now more than ever. Which is why when more scientists have access to the latest scientific instrumentation, we all benefit. That is the sole purpose of our Lab-Forward Trade-In Program. For a limited time, trade in your chromatography and mass spectrometry instruments and save!
    Learn more
    * Required field
    *
    *
    *
    *
    *
    *
    *
    *
    *
     Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
    This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
    Style Sheet for Global Design System