Search Thermo Fisher Scientific
Meet your lab’s needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific Multidrop reagent dispensers. Designed to support the varied requirements for liquid dispensing in pharmaceutical and biotechnology laboratories. Compact Thermo Scientific Multidrop dispensers fit easily on the bench, and the intuitive user interface makes setup and programming a breeze. Fill plates in seconds with excellent precision and reproducibility, helping you run tests faster and increase productivity.
Multidrop Combi Automated Reagent Dispensers |
![]() Multidrop Combi+ | ![]() Multidrop Combi nL | ||
User interface | On-board interface or FILLit PC software | On-board interface or FILLit PC software | On-board interface or FILLit PC software |
---|---|---|---|
Volume | 0.5-2500 µl | 0.5-2500 µl | 50 nl-50 µl |
Volume increments | Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments Standard tube dispensing cassette: 5- 2,000 µL in 5 µLuncrements | Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments Standard tube dispensing cassette: 5- 2,000 µL in 5 µLuncrements | 1nL increments 50-999nL, 10nL increments 1.00-9.99µL, 100nL increments 10.0-50.0µL, FILLit Software 1nL increments |
Dispense volume adjustability | Per column | Per column | Per well |
Dead volume | < 1 mL volume range: 0.5 - 50 µL <7 mL volume range: >50 µL (it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent) | < 1 mL volume range: 0.5 - 50 µL <7 mL volume range: >50 µL (it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent) | <1.2 mL |
Plate formats | 9- to 1,536-well plates: including 96- ad 384-deep well and tubes in rack format (plate height: 5 - 50 mm) | 9- to 1,536-well plates: including 96- ad 384-deep well and tubes in rack format (plate height: 5 - 50 mm) | 96-1536 well plates; including 96, 384- tubes in rack format |
Plate height adjustment | Automatic | Automatic | Automatic |
Automation/robot compatible | Yes | Yes | Yes |
Dispensing Technology | Peristaltic pump | Peristaltic pump | Pressurized valve system |
Dimensions | 14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) | 14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) | 14 x 8.6 x 14.8 in 35.5 x 22 x 37.5 cm |
Weight | 20.1 lb/9.1K | 20.1 lb/9.1K | 21.2 lb/9.6 kg |
*Multidrop Combi SMART+ uses RFID to track cassette usage
Increase your control of Thermo Scientific Multidrop reagent dispensers. Thermo Scientific FILLit Software is an easy-to-use tool that allows rapid development of protocols that can be run directly from the PC or transferred into instrument memory.
Features:
The comprehensive line of Thermo Scientific Multidrop dispensing cassettes offers optimal solutions for a wide volume range and a variety of reagents for the best performance and results. Choose between standard and small tube cassettes and SMART+ versions of these, which store dispensing lifetime data.
![]() Multidrop Combi+ | ![]() Multidrop Combi nL | ||
User interface | On-board interface or FILLit PC software | On-board interface or FILLit PC software | On-board interface or FILLit PC software |
---|---|---|---|
Volume | 0.5-2500 µl | 0.5-2500 µl | 50 nl-50 µl |
Volume increments | Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments Standard tube dispensing cassette: 5- 2,000 µL in 5 µLuncrements | Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments Standard tube dispensing cassette: 5- 2,000 µL in 5 µLuncrements | 1nL increments 50-999nL, 10nL increments 1.00-9.99µL, 100nL increments 10.0-50.0µL, FILLit Software 1nL increments |
Dispense volume adjustability | Per column | Per column | Per well |
Dead volume | < 1 mL volume range: 0.5 - 50 µL <7 mL volume range: >50 µL (it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent) | < 1 mL volume range: 0.5 - 50 µL <7 mL volume range: >50 µL (it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent) | <1.2 mL |
Plate formats | 9- to 1,536-well plates: including 96- ad 384-deep well and tubes in rack format (plate height: 5 - 50 mm) | 9- to 1,536-well plates: including 96- ad 384-deep well and tubes in rack format (plate height: 5 - 50 mm) | 96-1536 well plates; including 96, 384- tubes in rack format |
Plate height adjustment | Automatic | Automatic | Automatic |
Automation/robot compatible | Yes | Yes | Yes |
Dispensing Technology | Peristaltic pump | Peristaltic pump | Pressurized valve system |
Dimensions | 14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) | 14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) | 14 x 8.6 x 14.8 in 35.5 x 22 x 37.5 cm |
Weight | 20.1 lb/9.1K | 20.1 lb/9.1K | 21.2 lb/9.6 kg |
*Multidrop Combi SMART+ uses RFID to track cassette usage
Increase your control of Thermo Scientific Multidrop reagent dispensers. Thermo Scientific FILLit Software is an easy-to-use tool that allows rapid development of protocols that can be run directly from the PC or transferred into instrument memory.
Features:
The comprehensive line of Thermo Scientific Multidrop dispensing cassettes offers optimal solutions for a wide volume range and a variety of reagents for the best performance and results. Choose between standard and small tube cassettes and SMART+ versions of these, which store dispensing lifetime data.
Get a look inside the Multidrop Combi+ and Combi SMART+
Note: This tool is best viewed in Chrome, Firefox or Safari Browsers. Learn more ›
Multidrop Pico |
User interface | PicoIT PC software | PicoIT PC software |
---|---|---|
Volumes | 11 pL - 20 µl or 1 nL - 200 µl | 11 pL - 20 µl or 1 nL - 200 µl |
Plate formats | 12, 24, 48, 96, 384, 1536 well plate | 12, 24, 48, 96, 384 well plate |
Plate height adjustment | Automatic | Manual |
Automation/robot compatible | Yes | No |
Dispensing Technology | Non-contact digital dispensing | Non-contact digital dispensing |
Application | Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting | |
Dimensions | 47 x 38 x 32 cm | 31 x 28 x 20 cm |
Weight | 14.1 kg | 5.9 kg |
An easy-to-use Multidrop PicoIT software platform with application-focused wizards and a highlighted loading guide allows effortless and quick plate setup and operation. Available in one- and eight-well formats to support different throughputs and amounts of reagents and samples.
The pre-programmed protocols provide:
![]() |
|
User interface | PicoIT PC software | PicoIT PC software |
---|---|---|
Volumes | 11 pL - 20 µl or 1 nL - 200 µl | 11 pL - 20 µl or 1 nL - 200 µl |
Plate formats | 12, 24, 48, 96, 384, 1536 well plate | 12, 24, 48, 96, 384 well plate |
Plate height adjustment | Automatic | Manual |
Automation/robot compatible | Yes | No |
Dispensing Technology | Non-contact digital dispensing | Non-contact digital dispensing |
Application | Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting | |
Dimensions | 47 x 38 x 32 cm | 31 x 28 x 20 cm |
Weight | 14.1 kg | 5.9 kg |
An easy-to-use Multidrop PicoIT software platform with application-focused wizards and a highlighted loading guide allows effortless and quick plate setup and operation. Available in one- and eight-well formats to support different throughputs and amounts of reagents and samples.
The pre-programmed protocols provide:
![]() |
|
Get a look inside the Pico
Note: This tool is best viewed in Chrome, Firefox or Safari Browsers. Learn more ›