Search Thermo Fisher Scientific
Organism Type | Mouse | a subline derived from the BALB/c mouse MOPC-104E plasmacytoma cell line. KD83 cells are dependent on exogenous IL6 (see also: Factor-dependent cell lines ) and proliferate in response to murine and human IL6 . The cells also respond to murine IL5 but not to human IL5 . KDR83 cells do not respond to IL1 , IL2 , IL3 , IL4 , IFN-alpha , IFN-beta , IFN-gamma , G-CSF , GM-CSF . The cells have been used in bioassays of IL6 . |
Tissues | Blood | |
Phenotype | Suspension | |
Primary | no | |
Application | Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||