Clicktainer
Sample Safety with a Multi-Click
Introducing the newest innovation to sample collection. Designed with unique leakproof technology, the new Thermo Scientific Samco  Clicktainer Vials provide ultimate sample protection and user safety with an audible multi-click seal. 
Buy now
Wide range of cap colors and packaging configurations
clicktainer caps
Different Cap Colors
  • Cap colors to differentiate and manage samples
  • Available pre-assembled, sterile and non-sterile, for convenient usage
  • Separate closure and vial bulk packs  for fixative pre-filling applications
  • Individual peel pouches for operating rooms, cleanrooms, and other sterile environments
  • Dispenser boxes ideal for neat storage in smaller labs and doctors’ offices
Clearly Designed Labels
  • One label multiple languages – English, Spanish and French
  • Fields to capture all FDA required information
  • Optional temperature strip
  • Also available unlabeled
Clicktainer label
Variety of Applications
  • Collection and transport of fluids, solids and powders
  • Collection of valuable and often hazardous patient samples, such as urine, feces and sputum
  • Transportation through hospital pneumatic/vacuum transport systems
  • Suitability for use with formalin or fixatives for histology applications
Superior Leak Tight Performance
The Clicktainer Vials are proven leak tight in accordance with BS EN 14254, validated as 95kPa compliant and rigorously tested for suitability in pneumatic tubes. 
See the competitive difference
 ClicktainerBrand ABrand BBrand C
Passive Leak Test≥95%≥94%Not tested≥89%
95 kPa Vacuum≥95%≥94%≥92%0%
Pneumatic Transport System≥95%≥77%Not tested≥71%
Clicktainer vial offering
Select the product code of interest and it will appear on the top of the ordering page.
120mL/53mm vials
Catalog # Name Size Price (USD) Qty
120ORG53-1000 Samco 120mL/53mm CLK SPC LBL OR CAP ST Case of 300
162.00
Online Exclusive
180.00
Save 18.00 (10%)
120ORG53-4000 Samco 120mL/53mm CLK O/R POUCH OR CAP ST, Case of 100 Case of 100 -
120WHT53-1000 Samco 120mL/53mm CLK SPC LBL WT CAP ST Case of 300
165.60
Online Exclusive
184.00
Save 18.40 (10%)
120WHT53-2000 Samco 120mL/53mm CLK SPC N/L WT CAP NS Case of 300
123.43
Online Exclusive
165.00
Save 41.57 (25%)
120WHT53-3000 Samco 120mL/53mm CLK MSU NO FUNNEL ST, Case of 100 Case of 100 -
120WHT53-4000 Samco 120mL/53mm CLK POUCH WT CAP ST, Case of 100 Case of 100 -
90mL/53mm vials
Catalog # Name Size Price (USD) Qty
90GRN53-1000 Samco 90mL/53mm CLK SPC LBL GN CAP ST, Case of 400 Case of 400 -
90ORG53-1000 Samco 90mL/53mm CLK SPC LBL OR CAP ST Case of 400
103.40
Online Exclusive
188.00
Save 84.60 (45%)
90ORG53-1001 Samco 90mL/53mm CLK SPC LBL OR CAP ST, Case of 320 Case of 320 -
90WHT53-1000 Samco 90mL/53mm CLK SPC LBL WT CAP ST Case of 300
124.20
Online Exclusive
138.00
Save 13.80 (10%)
90WHT53-1001 Samco 90mL/53mm CLK SPC TSLBL WT CAP ST Case of 300
123.27
Online Exclusive
292.00
Save 168.73 (58%)
90WHT53-1002 Samco 90mL/53mm CLK SPC TS WT CAP ST Case of 400
140.27
Online Exclusive
332.00
Save 191.73 (58%)
90WHT53-2000 Samco 90mL/53mm CLK SPC LBL WT CAP NS Case of 300
88.08
Online Exclusive
133.00
Save 44.92 (34%)
90WHT53-2001 Samco 90mL/53mm CLK SPC TSLBL WT CAP NS Case of 300
216.90
Online Exclusive
241.00
Save 24.10 (10%)
90WHT53-2002 Samco 90mL/53mm CLK SPC N/LBL WT CAP NS Case of 300
111.60
Online Exclusive
124.00
Save 12.40 (10%)
90WHT53-2003 Samco 90mL/53mm CLK SPC N/LBL WT CAP NS Case of 300
117.00
Online Exclusive
130.00
Save 13.00 (10%)
90WHT53-2004 Samco 90mL/53mm CLK SPC TS WT CAP NS Case of 400
233.10
Online Exclusive
259.00
Save 25.90 (10%)
Caps and uncapped vials
120mL/53mm vials
Catalog # Name Size Price (USD) Qty
120ORG53-1000 Samco 120mL/53mm CLK SPC LBL OR CAP ST Case of 300
162.00
Online Exclusive
180.00
Save 18.00 (10%)
120ORG53-4000 Samco 120mL/53mm CLK O/R POUCH OR CAP ST, Case of 100 Case of 100 -
120WHT53-1000 Samco 120mL/53mm CLK SPC LBL WT CAP ST Case of 300
165.60
Online Exclusive
184.00
Save 18.40 (10%)
120WHT53-2000 Samco 120mL/53mm CLK SPC N/L WT CAP NS Case of 300
123.43
Online Exclusive
165.00
Save 41.57 (25%)
120WHT53-3000 Samco 120mL/53mm CLK MSU NO FUNNEL ST, Case of 100 Case of 100 -
120WHT53-4000 Samco 120mL/53mm CLK POUCH WT CAP ST, Case of 100 Case of 100 -
90mL/53mm vials
Catalog # Name Size Price (USD) Qty
90GRN53-1000 Samco 90mL/53mm CLK SPC LBL GN CAP ST, Case of 400 Case of 400 -
90ORG53-1000 Samco 90mL/53mm CLK SPC LBL OR CAP ST Case of 400
103.40
Online Exclusive
188.00
Save 84.60 (45%)
90ORG53-1001 Samco 90mL/53mm CLK SPC LBL OR CAP ST, Case of 320 Case of 320 -
90WHT53-1000 Samco 90mL/53mm CLK SPC LBL WT CAP ST Case of 300
124.20
Online Exclusive
138.00
Save 13.80 (10%)
90WHT53-1001 Samco 90mL/53mm CLK SPC TSLBL WT CAP ST Case of 300
123.27
Online Exclusive
292.00
Save 168.73 (58%)
90WHT53-1002 Samco 90mL/53mm CLK SPC TS WT CAP ST Case of 400
140.27
Online Exclusive
332.00
Save 191.73 (58%)
90WHT53-2000 Samco 90mL/53mm CLK SPC LBL WT CAP NS Case of 300
88.08
Online Exclusive
133.00
Save 44.92 (34%)
90WHT53-2001 Samco 90mL/53mm CLK SPC TSLBL WT CAP NS Case of 300
216.90
Online Exclusive
241.00
Save 24.10 (10%)
90WHT53-2002 Samco 90mL/53mm CLK SPC N/LBL WT CAP NS Case of 300
111.60
Online Exclusive
124.00
Save 12.40 (10%)
90WHT53-2003 Samco 90mL/53mm CLK SPC N/LBL WT CAP NS Case of 300
117.00
Online Exclusive
130.00
Save 13.00 (10%)
90WHT53-2004 Samco 90mL/53mm CLK SPC TS WT CAP NS Case of 400
233.10
Online Exclusive
259.00
Save 25.90 (10%)
Caps and uncapped vials
*The term “leakproof” applies to Thermo Scientific Samco Clicktainer products, 90mL/53 mm and 120mL/53 mm, that meet the following criteria: a) a vacuum test that draws 95kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with BS EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by 1 hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco Clicktainer Vials under conditions of their planned applications. Thermo Scientific Samco Clicktainer Vials are leakproof at ambient temperature and pressure when used with their Clicktainer closures.
Style Sheet for Global Design System