Overview
Performance, robustness, and confidence at the throughput you need to support your oligonucleotide analyses.
When purity and characterization are critical to your work, you can rely on our anion-exchange and reversed phase column chemistries. For over 30 years, the Thermo Scientific DNAPac family of columns has been the go-to for high resolution U/HPLC separation of oligonucleotides. These columns have the performance and robustness to meet your oligonucleotide analysis needs so you can be confident in your data while meeting the throughput demands of your laboratory.
Oligonucleotide HPLC & UHPLC Columns
DNAPac RP HPLC ColumnsDNAPac PA200 & DNAPac PA200RS HPLC ColumnsDNASwift HPLC Columns
✓ Compatible with LC-UV and LC-MS/MS
✓ Offer ion-pair reversed phase separations of nucleic acid mixtures
✓ Oligonucleotide samples from siRNA to mRNA easily resolve on this polymer chemistry
✓ Deliver outstanding separations
✓ Compatible with LC-UV
✓ Strong anion-exchange columns for n, n-1, and n+1 separations of oligonucleotide samples
✓ Offer orthogonal separation to reversed phase columns
✓ Separate oligonucleotide samples by size and charge
✓ A monolithic column designed for strong anion-exchange purification of oligonucleotide samples using analytical HPLC
✓ High loadability, with slightly less resolution than our analytical columns
Order now
Order now
Order now
3 ways to search
  1. Find an equivalent column to the one you’re using now
  1. Find the best column for your USP method
  1. Find the right column for your new method
Find your HPLC column with ease ›
Tips to improve your charge variant analysis by ion exchange
Learn about the common mistakes users make with ion exchange columns and ways to avoid those mistakes.
Related resources
* Required field
*
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code