Related products to optimize your cell culture workflow
Thermo Scientific CO2 resistant shakers, biological safety cabinets, centrifuges and Thermo Scientific Nunc cell culture ware are essential products for your cell culture lab.
CO₂ resistant shakers
Get reliable around-the-clock operation ideally suited for flexible use inside a CO2 incubator or on the benchtop with CO2 resistant shakers. Orbital shaking through magnetic power eliminates the need to replace belts and the small footprint is ideal for use in a CO2 incubator. Mechanical components are specially treated to protect in conditions up to 20% CO2 and 95% humidity. An external control box with LED display allows for easy adjustments without opening the chamber door. 
Biological safety cabinets
Thermo Scientific biological safety cabinets (BSCs) offer certified performance and protection in an ergonomic, energy efficient design. Our BSCs include advanced features like dual DC motors to automatically balance the cabinet inflow and downflow air velocities in real time, better protecting you and your precious cultures. The Digital Airflow Verification (DAVe) alarm signals any out-of-spec conditions for added assurance.
Centrifuges
Because harvesting and separating are critical steps in your cell culture process, it’s important to consider the lab centrifuge requirements and technical specifications for your applications.  From selecting the appropriate speed and g-force to exploring the latest trends in centrifugation, we offer a wide array of Thermo Scientific centrifuges and innovative rotors for all your processing needs.
Cell culture flasks and dishes
Thermo Scientific Nunc cell culture vessels are designed for consistency, reproducibility, and proper cell health, with the surfaces and ancillary options you're looking for in a tissue culture flask. Choose from a variety of sizes, with culture areas ranging from 25cm2 to 500cm2 and a selection of surface treatments to address your specific applications and cell types.
Smart-Vue Pro Remote Monitoring System
Thermo Scientific wireless monitoring solutions are designed to help put your mind at ease. Knowing a state-of-the-art, wireless monitoring system is watching, safeguarding, and data logging critical equipment parameters, and knowing that in the event of a power or mechanical failure, you’ll be notified right away provides peace of mind. Thermo Scientific™ Smart-Vue™ Pro system is a remote monitoring solution for temperature, relative humidity, and CO₂ in real time.
Cell culture workflow
From sample preparation to storage, explore the steps and all products we can support you with for your cell culture application.
1x1 image pixel for data collection