Search Thermo Fisher Scientific
Use your existing CO2 incubator to investigate suspension culture and scale-up. Save space and achieve reliable around-the-clock operation with CO2 resistant shakers ideally suited to maximize cell health within your existing CO2 incubator.
Sealed electronics and proven mechanism technologies provide minimal heat dissipation to ensure conditions within the CO2 chamber are not affected, and cells can thrive.
CO2 resistant shakers can be easily removed from the incubator to facilitate cleaning and disinfection, protecting your cultures from unwanted contamination, providing ease of use for multi-tasking cell culture labs.
Flexibility allows it to go from the CO2 incubator to the benchtop to the microbiological incubator for culturing mammalian suspension cells, insect cells, bacteria, fungi or other cell types for maximum value and versatility.
CO2 Resistant Shaker 3mm Orbit | CO2 Resistant Shaker 19mm Orbit | MaxQ 416 Shaker | |
![]() | ![]() | ![]() | |
Learn more | Learn more | Learn more | |
Maximum vessel size | 5L Erlenmeyer flask | 5L Erlenmeyer flask | 6L Erlenmeyer flask |
Available platforms | 14.1 x 11.8 in. 18 x 18 in. | 14.1 x 11.8 in. 18 x 18 in. | 18 x 18 in. 18 x 24 in. |
Speed | 30 to 600 rpm | 30 to 300 rpm | 25 to 525 rpm |
Environmental temperature range | 4° to 60°C | 4° to 60°C | 4° to 40°C |
Humidity limits | 20% to 95% | 20% to 95% | 50% to 80% |
Exterior dimensions (H x W x D) | 3.8 x 14.2 x 11.8 in. (9.7 x 36 x 30 cm) | 3.8 x 14.2 x 11.8 in. (9.7 x 36 x 30 cm) | 7.8 x 30.5 x 26 in. (19.8 x 77.5 x 66 cm) |
Orbit | .118 (0.3 cm) | .75 in. (1.9 cm) | 1 in. (2.5 cm) |
Weight capacity | 22 lbs (10 kg)* | 13 lb. (6 kg) | 71 lb. (32 kg) |
Data output | 4-20 mA | 4-20 mA | NA |
Accessories | Platforms & Clamps › Test Tube Racks › Plugs & Accessories › | Platforms & Clamps › Test Tube Racks › Plugs & Accessories › | Platforms & Clamps › Test Tube Racks › Accessories › |
*Maximum load when <150 rpm
Want some help making a decision? Talk with one of our expert sales representatives. Contact us
Maximize efficiency and productivity by mixing and matching labware on various platform and holder types and truly customize your shaker to fit your unique needs. Go ›
Racks and trays will outfit your shaker to accommodate a wide variety of applications and objectives, and to keep conditions clean and safe. Go ›
Plugs and miscellaneous accessories ensure you have the correct configuration for your geographic region plus parts to keep your shaker performing at its best. Go ›
Choose a universal platform for maximum flexibility, using a mix of different size labware on a single platform, or choose dedicated platforms that come with pre-installed clamps.
Must purchase desired test tube rack in addition to holder.
Thermo Scientific CO2 incubators and environmental chambers are ideal to pair with your Thermo Scientific CO2 resistant shakers for the flexibility to scale up suspension cultures while continuing stationary cultures in one system.
CO2 Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
---|---|---|---|---|
Forma 310 Direct Heat CO2 Incubator | 1 | 9 | 470 x 470mm | 16kg slide in and out |
Forma Steri-Cycle CO2 Incubator | 1 | 9 | 470 x 470mm | 16kg slide in and out |
Forma Series II 3110 Water-Jacketed CO2 Incubator | 1 | 9 | 470 x 470mm | 13.6kg slide in and out |
Large-Capacity Reach-In CO2 Incubator | 8 units, 4 on each shelf | 72 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
Heracell 150i CO2 Incubators -Stainless | 1 | 9 | 423 x 465mm | 10kg |
Heracell 150i CO2 Incubators -Copper | 1 | 9 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Stainless | 1 | 9 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Copper | 1 | 9 | 423 x 465m | 10kg |
Heracell 240i CO2incubator-Stainless | 1 | 18 | 560 x 500mm | 10kg |
Heracell 240i CO2incubator-Copper | 1 | 18 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Stainless | 1 | 18 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Copper | 1 | 18 | 560 x 500mm | 10kg |
Thermo Scientific Forma 3940 Series Environmental Chambers | 3 | 48 | 656 x 777mm | 90kg fixed reinforced shelves* (1900005) |
CO2 Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
---|---|---|---|---|
Forma 310 Direct Heat CO2 Incubator | 0 | 0 | 470 x 470mm | 16kg slide in and out |
Forma Steri-Cycle CO2 Incubator | 0 | 0 | 470 x 470mm | 16kg slide in and out |
Forma Series II 3110 Water-Jacketed CO2 Incubator | 0 | 0 | 470 x 470mm | 13.6kg slide in and out |
Large-Capacity Reach-In CO2 Incubator | 8 units, 4 on each shelf | 80 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
Heracell 150i CO2 Incubators -Stainless | 0 | 0 | 423 x 465mm | 10kg |
Heracell 150i CO2 Incubators -Copper | 0 | 0 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Stainless | 0 | 0 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Copper | 0 | 0 | 423 x 465m | 10kg |
Heracell 240i CO2incubator-Stainless | 1 | 10 | 560 x 500mm | 10kg |
Heracell 240i CO2incubator-Copper | 1 | 10 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Stainless | 1 | 10 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Copper | 1 | 10 | 560 x 500mm | 10kg |
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
CO2 Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
---|---|---|---|---|
Thermo Scientific Large-Capacity Reach-In CO2 Incubator | 2 | 48 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
Thermo Scientific Forma 3960 Series Environmental Chambers | 3 | 48 | 656 x 777mm | 90kg fixed reinforced shelves* (1900005) |
CO2 Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
---|---|---|---|---|
Forma 310 Direct Heat CO2 Incubator | 1 | 9 | 470 x 470mm | 16kg slide in and out |
Forma Steri-Cycle CO2 Incubator | 1 | 9 | 470 x 470mm | 16kg slide in and out |
Forma Series II 3110 Water-Jacketed CO2 Incubator | 1 | 9 | 470 x 470mm | 13.6kg slide in and out |
Large-Capacity Reach-In CO2 Incubator | 8 units, 4 on each shelf | 72 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
Heracell 150i CO2 Incubators -Stainless | 1 | 9 | 423 x 465mm | 10kg |
Heracell 150i CO2 Incubators -Copper | 1 | 9 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Stainless | 1 | 9 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Copper | 1 | 9 | 423 x 465m | 10kg |
Heracell 240i CO2incubator-Stainless | 1 | 18 | 560 x 500mm | 10kg |
Heracell 240i CO2incubator-Copper | 1 | 18 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Stainless | 1 | 18 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Copper | 1 | 18 | 560 x 500mm | 10kg |
Thermo Scientific Forma 3940 Series Environmental Chambers | 3 | 48 | 656 x 777mm | 90kg fixed reinforced shelves* (1900005) |
CO2 Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
---|---|---|---|---|
Forma 310 Direct Heat CO2 Incubator | 0 | 0 | 470 x 470mm | 16kg slide in and out |
Forma Steri-Cycle CO2 Incubator | 0 | 0 | 470 x 470mm | 16kg slide in and out |
Forma Series II 3110 Water-Jacketed CO2 Incubator | 0 | 0 | 470 x 470mm | 13.6kg slide in and out |
Large-Capacity Reach-In CO2 Incubator | 8 units, 4 on each shelf | 80 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
Heracell 150i CO2 Incubators -Stainless | 0 | 0 | 423 x 465mm | 10kg |
Heracell 150i CO2 Incubators -Copper | 0 | 0 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Stainless | 0 | 0 | 423 x 465mm | 10kg |
Heracell VIOS 160i CO2incubator-Copper | 0 | 0 | 423 x 465m | 10kg |
Heracell 240i CO2incubator-Stainless | 1 | 10 | 560 x 500mm | 10kg |
Heracell 240i CO2incubator-Copper | 1 | 10 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Stainless | 1 | 10 | 560 x 500mm | 10kg |
Heracell VIOS 250i CO2incubator-Copper | 1 | 10 | 560 x 500mm | 10kg |
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
CO2 Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
---|---|---|---|---|
Thermo Scientific Large-Capacity Reach-In CO2 Incubator | 2 | 48 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
Thermo Scientific Forma 3960 Series Environmental Chambers | 3 | 48 | 656 x 777mm | 90kg fixed reinforced shelves* (1900005) |
Our CO2 resistant orbital shakers provide reliable around-the-clock operation ideally suited to keep your cells flourishing within your working environment.
Compared to a dedicated CO2 shaking incubator, our CO2 resistant shaker offers culturing flexibility, reliability, and ease of use for multi-tasking cell culture labs. Unlike a combined shaking incubator, a CO2 resistant shaker can be easily removed from the incubator to facilitate cleaning and disinfection, protecting your cultures from unwanted contamination.