Mini Gel Tank specifications
CapacityUp to 2 mini gels
Gel compatibilityGel size: 8 x 8cm
Gel cassette: 10 x 10cm
Thickness: 1.0mm or 1.5mm
Compatible gelsAll Invitrogen precast mini gels, SureCast handcast gels, and Invitrogen empty mini gel cassettes
Buffer chamber requirement400 mL for each mini gel
Optional transfer apparatusMini Blot Module
Transfer capacityUp to 2 mini gels
Compatible power suppliesPowerEase Touch 120W and PowerEase Touch 350W, Owl systems, for other systems such as Hoefer or Bio-Rad use with Novex Power Supply Adapters (Cat. No. ZA10001)
Unit dimensions32 x 11.5 x 16 cm (height with lid)
MaterialPolycarbonate
Electrode wirePlatinum
Electrode limits500 VDC or 100 Watts
Chemical resistanceThe Mini Gel Tank is impervious to most alcohols but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene), or acetone.
Catalog numberA25977
Description
The Mini Gel Tank has a unique tank design that enables convenient side-by-side gel loading and enhanced viewing during use. Each tank can accommodate up to two gels per run where each gel is run in its own separate chamber- making it easy to run just one gel. This advantage allows for lower buffer requirements- only use the buffer needed to run one gel.
The Mini Gel Tank can be used for wet tank transfers using the Mini Blot Module. Two Mini Blot Modules fit into the Mini Gel Tank to allow transfer of two gels at a time. The unique design of the blot module allows for lower transfer buffer- only 220 mL per gel. This lower requirement keeps the amount of methanol needed to a minimum. Learn more about transferring in the Mini Gel Tank here.
Learn how to separate proteins using the Invitrogen Mini Gel Tank.
Video Player is loading.
Current Time 0:00
Duration 5:24
Loaded: 3.04%
Stream Type LIVE
Remaining Time 5:24
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how to perform a western wet tank transfer using the Invitrogen Mini Blot Module in the Mini Gel Tank.
    Video Player is loading.
    Current Time 0:00
    Duration 4:05
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 4:05
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Recommended electrophoresis run conditions
      Gel TypeVoltage (V)Current (mA)Ending CurrentRun Time
      Bolt Bis-Tris Plus 4-12% (MES)2001607020
      Bolt Bis-Tris Plus 4-12% (MOPS)2001605035
      NuPAGE 4-12% BisTris (MES)2001609030
      NuPAGE 4-12% BisTris (MOPS)2001405042
      Novex 4-20% TrisGlycine (denatured)1254010100
      Novex 4-20% TrisGlycine (native)125301090
      NuPAGE 3-8% TrisAcetate (denatured)150602050
      NuPAGE 3-8% TrisAcetate (native)1504010100
      Novex 10-20% Tricine1251104065
      NativePAGE 3-12%15010<1080
      pH 3-10 IEFprogram85150
      10% Zymogram (Gelatin)125401090
      10% TBE-Urea (DNA)180101050
      Catalog # Name Size Price (USD) Qty
      A25977 Mini Gel Tank Each
      735.65
      Online Exclusive
      760.00
      Save 24.35 (3%)
      A25944 Mini Gel Tank Lid Each
      211.65
      Online Exclusive
      218.00
      Save 6.35 (3%)
      A25945 Cassette Clamp, right Each
      173.65
      Online Exclusive
      179.00
      Save 5.35 (3%)
      A25946 Cassette Clamp, left Each
      170.65
      Online Exclusive
      176.00
      Save 5.35 (3%)
      A25950 Mini Gel Tank Base Each
      43.65
      Online Exclusive
      44.64
      NW2000 Mini Gel Tank and Blot Module Set Each
      1,314.65
      Online Exclusive
      1,368.00
      Save 53.35 (4%)
      B4478641 Gel Runner Tank Each
      197.00
      Mini Gel Tank specifications
      CapacityUp to 2 mini gels
      Gel compatibilityGel size: 8 x 8cm
      Gel cassette: 10 x 10cm
      Thickness: 1.0mm or 1.5mm
      Compatible gelsAll Invitrogen precast mini gels, SureCast handcast gels, and Invitrogen empty mini gel cassettes
      Buffer chamber requirement400 mL for each mini gel
      Optional transfer apparatusMini Blot Module
      Transfer capacityUp to 2 mini gels
      Compatible power suppliesPowerEase Touch 120W and PowerEase Touch 350W, Owl systems, for other systems such as Hoefer or Bio-Rad use with Novex Power Supply Adapters (Cat. No. ZA10001)
      Unit dimensions32 x 11.5 x 16 cm (height with lid)
      MaterialPolycarbonate
      Electrode wirePlatinum
      Electrode limits500 VDC or 100 Watts
      Chemical resistanceThe Mini Gel Tank is impervious to most alcohols but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene), or acetone.
      Catalog numberA25977
      Description
      The Mini Gel Tank has a unique tank design that enables convenient side-by-side gel loading and enhanced viewing during use. Each tank can accommodate up to two gels per run where each gel is run in its own separate chamber- making it easy to run just one gel. This advantage allows for lower buffer requirements- only use the buffer needed to run one gel.
      The Mini Gel Tank can be used for wet tank transfers using the Mini Blot Module. Two Mini Blot Modules fit into the Mini Gel Tank to allow transfer of two gels at a time. The unique design of the blot module allows for lower transfer buffer- only 220 mL per gel. This lower requirement keeps the amount of methanol needed to a minimum. Learn more about transferring in the Mini Gel Tank here.
      Learn how to separate proteins using the Invitrogen Mini Gel Tank.
      Video Player is loading.
      Current Time 0:00
      Duration 5:24
      Loaded: 3.04%
      Stream Type LIVE
      Remaining Time 5:24
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Learn how to perform a western wet tank transfer using the Invitrogen Mini Blot Module in the Mini Gel Tank.
        Video Player is loading.
        Current Time 0:00
        Duration 4:05
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 4:05
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Recommended electrophoresis run conditions
          Gel TypeVoltage (V)Current (mA)Ending CurrentRun Time
          Bolt Bis-Tris Plus 4-12% (MES)2001607020
          Bolt Bis-Tris Plus 4-12% (MOPS)2001605035
          NuPAGE 4-12% BisTris (MES)2001609030
          NuPAGE 4-12% BisTris (MOPS)2001405042
          Novex 4-20% TrisGlycine (denatured)1254010100
          Novex 4-20% TrisGlycine (native)125301090
          NuPAGE 3-8% TrisAcetate (denatured)150602050
          NuPAGE 3-8% TrisAcetate (native)1504010100
          Novex 10-20% Tricine1251104065
          NativePAGE 3-12%15010<1080
          pH 3-10 IEFprogram85150
          10% Zymogram (Gelatin)125401090
          10% TBE-Urea (DNA)180101050
          Catalog # Name Size Price (USD) Qty
          A25977 Mini Gel Tank Each
          735.65
          Online Exclusive
          760.00
          Save 24.35 (3%)
          A25944 Mini Gel Tank Lid Each
          211.65
          Online Exclusive
          218.00
          Save 6.35 (3%)
          A25945 Cassette Clamp, right Each
          173.65
          Online Exclusive
          179.00
          Save 5.35 (3%)
          A25946 Cassette Clamp, left Each
          170.65
          Online Exclusive
          176.00
          Save 5.35 (3%)
          A25950 Mini Gel Tank Base Each
          43.65
          Online Exclusive
          44.64
          NW2000 Mini Gel Tank and Blot Module Set Each
          1,314.65
          Online Exclusive
          1,368.00
          Save 53.35 (4%)
          B4478641 Gel Runner Tank Each
          197.00