Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Arcturus® RNA Amplification Kits for research use enable gene expression analysis of millions of previously inaccessible formalin-fixed, paraffin-embedded (FFPE) research tissue samples and frozen samples. Amplify RNA from as little as 1 ng of total RNA or 5 ng of fixed RNA.
The RiboAmp® PLUS Frozen RNA Amplification Kits enable researchers to perform molecular analysis on RNA samples that, until now, have been too small for microarray analysis. Using a proprietary process for high-efficiency, high-fidelity linear amplification, messenger RNA is amplified up to 1,000-fold in one synthesis round and up to 1,000,000-fold in two rounds. The RiboAmp® PLUS Kits produce amplified antisense RNA (aRNA), ready for labeling and microarray hybridization or real-time PCR, and can be used to generate biologically relevant results with cDNA and oligonucleotide array platforms.
Figure 1. Schematic from Arcturus® RiboAmp® HS PLUS Kits. |
RNA Amplification Kit | Method | Applications | Linear Amplification Range (Lower Limit) | Cat. No. |
---|---|---|---|---|
Arcturus® RiboAmp® PLUS Amplification Kits | Proprietary | Labeling and microarray hybridization, real-time PCR | 1 ng of total RNA | KIT0526 (1.5-Round-6 samples) KIT0501 (Bulk-24 samples) KIT0521 (6 samples) KIT0527 (0.5 Round-6 samples) |
Arcturus® RiboAmp® HS PLUS cDNA Kit | Random primers | qRT-PCR | 100 pg of total RNA | KIT0529 |
Arcturus® RiboAmp® HS PLUS Amplification Kits (High Sensitivity) | Proprietary | Labeling and microarray hybridization, real-time PCR | 100 pg of total RNA | KIT0525 (6 samples) KIT0505 (Bulk-24 samples) KIT0528 (1.5 Round-6 samples) KIT0508 (1.5 Round-Bulk) |