Tyramide signal amplification is a technique that improves the detection of low abundance targets in imaging by covalently depositing fluorescent or chromogenic reporter molecules onto the target. Invitrogen Alexa Fluor Tyramide SuperBoost kits provide exceptional signal amplification and clarity, while SuperBoost colorimetric kits with EverRed and EverBlue allow for permanent staining in colorimetric immunohistochemistry (IHC) experiments.
What is tyramide signal amplification?
Tyramide signal amplification is a technique used to enhance the detection of low-abundance targets in imaging applications based on the enzymatic deposition of fluorescent or chromogenic reporter molecules onto the target of interest. Alexa Fluor Tyramide SuperBoost kits and reagents are specifically designed for exceptional signal amplification, offering additional signal definition and clarity required for fluorescent imaging of low-abundance targets. SuperBoost EverRed and EverBlue colorimetric kits and reagents can also be used for permanent staining in colorimetric IHC experiments for multiplexing with DAB substrate or as an alternative colorimetric stain.
How does tyramide signal amplification work?
Tyramide signal amplification used in the SuperBoost kits utilizes the catalytic activity of horseradish peroxidase (HRP) for high-density labeling of a target protein or nucleic acid sequence in situ. Typical immunocytochemistry (ICC), immunohistochemistry (IHC) and in situ hybridization (ISH) experiments using the SuperBoost tyramide reagents require 10–5000 times less primary antibody than standard ICC/IHC/ISH experiments to achieve the same signal intensity. Since the SuperBoost kits greatly enhance specific signal intensity over background, they can be easily optimized to detect specific signal in samples where high endogenous autofluorescence is observed.
The tyramide signal amplification process includes the use of horseradish peroxidase (HRP) to enzymatically convert fluorophore or chromogen tyramides to covalently bind tyrosine residues on and surrounding the protein epitope targeted by the primary antibody. As a controlled enzymatic reaction, tyramide signal amplification does not diffuse from the site of enzyme activity and therefore, provides better spatial resolution as compared to HRP or alkaline phosphatase-based methods.
Benefits of tyramide signal amplification and deposition
  • Enable excellent detection of low-abundance targets due to poly HRP secondary antibody
  • Ability to label with antibodies from the same species due to covalent attachment of the fluorophore, allowing antibody stripping in between rounds of tyramide signal amplification
  • High sensitivity allows the use of significantly lower concentrations of primary antibody
Table 1. Invitrogen tyramide signal amplification and deposition reagent options
 Alexa Fluor Tyramide SuperBoost ReagentsSuperBoost EverRed and EverBlue Tyramide Reagents
DetectionFluorescenceColorimetric and fluorescence
UseSingle or 2-plex staining for fluorescence ICC, IHC, and ISHSingle or multiplex permanent colorimetric staining for IHC
BrightnessHigherHigh
Sample typesCells or TissuesTissues
Alexa Fluor Tyramide SuperBoost kits for signal amplification for ICC/IHC
Combining the brightness of Invitrogen Alexa Fluor dyes with poly-HRP–mediated tyramide signal amplification, SuperBoost reagents yield precision and sensitivity 10–200 times greater than standard ICC/ IHC/ISH and 2–10 times that of other tyramide amplification techniques.
Order Alexa Fluor SuperBoost Kits
Benefits of the SuperBoost tyramide signal amplification kits include:
  • A highly sensitive fluorescent imaging detection method for low-abundance, hard-to-detect targets
  • Easy-to-use kits that produce signals compatible with standard filters and microscopes
  • High-resolution images and multiplex compatibility with fluorescent proteins, DAPI, secondary antibodies, and other SuperBoost kits
Figure 2. SuperBoost Tyramide kits and reagents with poly-HRP show brighter signal for longer duration. HeLa cells were incubated with various concentrations of anti-prohibitin antibody (manufacturer recommends a 1:150 dilution, or 5 µg/mL final), then labeled with the reagents in (1) the Invitrogen Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–rabbit IgG and Alexa Fluor 488 Tyramide); (2) our original Invitrogen TSA Kit #12 (goat anti–rabbit IgG and Alexa Fluor 488 Tyramide); or (3) an Invitrogen F(ab′)2 rabbit anti–goat IgG (H+L) secondary antibody. Cell images were captured from each treatment (using the same exposure and gain) with an Invitrogen EVOS FL Auto Imaging System (see information about EVOS imaging systems). These images indicate that the Alexa Fluor 488 Tyramide SuperBoost Kit offers higher-sensitivity detection than either our original TSA kits or directly labeled secondary antibodies.
How does SuperBoost tyramide signal amplification work?
Excellent for 1 or 2 plex imaging, Alexa Fluor Tyramide SuperBoost kits are simple to use and incorporate by itself or with other reagents and fluorophores. In this workflow, the fluorophore-conjugated secondary antibodies are replaced with SuperBoost tyramide signal amplification including secondary antibodies conjugated with poly-HRP. The only additional steps are incubation with conjugated tyramides for 2–10 minutes and addition of stop solution to halt HRP activity once the specific signal is detected. The stop solution additionally helps maintain the specificity and resolution of fluorescent signal. SuperBoost tyramide signal amplification kits offer a simple workflow like those used in standard ICC, IHC, and FISH.
Workflow for SuperBoost tyramide signal kits. This is a six- or seven-step process that can be optimized to provide clear and bright signal. This workflow indicates where to add other fluorescent reagents and secondary antibody fluorophores.
Examples of SuperBoost tyramide based amplification in IF, IHC, and ISH
Alexa Fluor Tyramide SuperBoost kits are compatible with a range of other marker detection and cell staining techniques, enabling multiplex experiments and fluorescence colocalization studies. These kits work with cell types and fluorescence imaging systems commonly used in standard ICC, IHC, and ISH methods. We have tested the performance of SuperBoost tyramide signal kits using formaldehyde-fixed cell lines in 2D and 3D cultures, FFPE tissues, and cryosectioned tissues.
SuperBoost tyramide signal reagent multiplexing can be achieved with:
  • Fluorescent markers for counterstaining, such as DAPI
  • Fluorescent proteins (i.e., GFP & RFP)
  • Standard ICC/IHC
  • Additional Alexa Fluor Tyramide SuperBoost kits
Table 2. Alexa Fluor Tyramide SuperBoost reagent selection guide
 Alexa Fluor 350 Tyramide ReagentAlexa Fluor 488 Tyramide ReagentAlexa Fluor 546 Tyramide ReagentAlexa Fluor 555 Tyramide ReagentAlexa Fluor 568 Tyramide ReagentAlexa Fluor 594 Tyramide ReagentAlexa Fluor 647 Tyramide ReagentAlexa Fluor Plus 750 Tyramide ReagentBiotin-XX
DetectionFluorescence
Ex/Em347/442495/519556/573555/565579/604591/617650/668750/790N/A
Channel430488514555594594647750N/A
EVOS filterCFPGFPYFPRFPTexas RedTexas RedCy5Cy7N/A
ApplicationSingle or 2-plex staining for fluorescence ICC, IHC, and ISH
Key benefits
  • Compatible with standard filters and microscopes
  • Amplify signal 10X, meaning less primary antibody is used
Tyramide ReagentB40952B40953B40954B40955B40956B40957B40958B56131B40951
SuperBoost Kits: goat anti-mouse B40912
B40941 (50 coverslips)
 B40913 B40915
B40942 (50 coverslips)
B40916 B40911
SuperBoost Kits: goat anti-rabbit B40922
B40943 (50 coverslips)
 B40923 B40925
B40944 (50 coverslips)
B40926 B40921
SuperBoost kits: Streptavidin B40932 B40933 B40935B40921 B40931
Table 3. Ordering information for stand-alone poly HRP antibodies
ProductDescriptionCat. No.
SuperBoost Goat anti-Mouse Poly HRPPoly HRP secondary antibody for mouse primariesB40961
SuperBoost Goat anti-Rabbit Poly HRPPoly HRP secondary antibody for rabbit primariesB40962
Alexa Fluor Tyramide SuperBoost experimental results
Cells displaying signal in red, blue, and some co-localized green.
With IHC and FFPE samples
Sample type: Rat intestinal section (FFPE).
Antibodies: Immunolabeled sequentially with three primary antibodies against H2B, actin and Ki-67.
Method: In between each antibody labeling, samples were microwaved in citrate buffer pH6 on high until boiling (~2 min), then microwaved for 15 minutes at 20% power and then allowed to cool to room temperature before labeling with the next rabbit antibody. Samples were labeled with 3 different primary antibodies: rabbit anti-H2B detected with Alexa Fluor 647 Tyramide SuperBoost Kit (green), rabbit anti–smooth muscle actin antibody detected with Alexa Fluor 488 Tyramide SuperBoost Kit (red), and rabbit anti-Ki67 antibody detected with Alexa Fluor 594 Tyramide SuperBoost Kit (blue).
Cells displaying signal in red and green, and some co-localized yellow.
With ICC in cell culture
Sample type: Fixed and permeabilized HeLa cells.
Antibodies: Immunolabeled sequentially with primary antibodies against anti–ATP synthase antibody and anti–β-catenin antibody.
Method: Cells were labeled with anti–ATP synthase antibody and an Alexa Fluor 594–conjugated secondary antibody. Additionally, the cells were incubated with an anti–β-catenin antibody and labeled with the reagents in the Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 488 tyramide). Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired on a confocal microscope.
Cells displaying signal in blue  and green.
With FISH and cell culture
Sample type: Fixed and permeabilized U2OS cells.
Antibodies: Immunolabled with anti-Cas9 antibody and probed with an oligo targeting the hprt gene.
Method: U2OS cells were fixed and permeabilized and then incubated with an hprt gene probes plus inactive Cas9 protein. Hprt probes were designed for Cas9 recognition, containing sg-RNA. To detect Cas9 protein and hprt probe complex assembled at hprt loci, anti-Cas9 antibody was used. This primary antibody was detected by Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 488 tyramide) detecting hprt loci specifically. Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired and analyzed on an EVOS FL Auto Imaging System (see information about EVOS imaging systems).
SuperBoost multiplexed with standard ICC/IHC
With standard ICC/IHC
Sample type: Cultured HeLa cells, fixed and permeabilized.
Antibodies: Cells were immunolabeled with an anti–tubulin and anti–ATP synthase subunit IF1 antibody.
Method: Fixed and permeabilized HeLa cells, treated using the reagents in the Image-iT Fixation/Permeabilization Kit, were incubated with an anti-tubulin primary antibody and an Alexa Fluor 488 goat anti–mouse IgG (H+L) secondary antibody. Cells were then incubated with an anti–ATP synthase subunit IF1 antibody and labeled with the reagents in the Alexa Fluor 594 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 594 tyramide). Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired on a confocal microscope.
SuperBoost multiplexed with another SuperBoost kit
With another SuperBoost Kit
Sample type: Cultured HeLa cells, fixed and permeabilized.
Antibodies: HeLa cells were immunolabeled with an anti-prohibitin antibody and an anti–β-catenin antibody.
Method: Fixed and permeabilized HeLa cells, treated using the reagents in the Image-iT Fixation/Permeabilization Kit, were incubated with an anti-prohibitin antibody and labeled with the reagents in the Alexa Fluor 647 Tyramide SuperBoost Kit (goat anti–rabbit IgG and Alexa Fluor 647 tyramide). Additionally, cells were incubated with anti–β-catenin and labeled with the reagents in the Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 488 tyramide). Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired on a confocal microscope.
SuperBoost EverRed and EverBlue kits for permanent colorimetric IHC
SuperBoost colorimetric signal anchoring, achieved with SuperBoost EverRed and EverBlue Colorimetric HRP Kits, enables highly sensitive and permanent target detection in multiplexable colorimetric IHC experiments. It combines poly-HRP-mediated tyramide signal deposition with SuperBoost EverRed and EverBlue for precise and sensitive colorimetric tissue staining. These SuperBoost kits use HRP catalytic activity for high-density labeling, resulting in wash-resistant, mountable, and archivable staining. SuperBoost EverRed and EverBlue provide sharp and intense signals without the need for air-drying or unconventional protocols. These user-friendly kits work with various tissue types and can be imaged with any brightfield microscope, yielding high-resolution images.
Order SuperBoost EverRed and EverBlue Kits
Benefits of the SuperBoost EverRed and EverBlue HRP Kits include:
  • Permanent colorimetric staining that is also fluorescent, allowing confirmation of the staining pattern with both brightfield and fluorescence imaging
  • Compatibility with other common colorimetric counterstains
  • Multiplexable between EverRed, EverBlue, and DAB
How does SuperBoost color signal anchoring work?
SuperBoost EverRed and EverBlue HRP Kits offer a simplified workflow similar to standard IHC staining methods. After detecting the antigen with a primary antibody, a secondary antibody or streptavidin (for biotinylated primary antibodies) conjugated with poly-HRP is used for staining. The HRP enzyme activates a chromogen tyramide, leading to localized covalent deposition of the tyramide derivative at tyrosine residues surrounding the targeted protein epitope. Treatment with the EverRed or EverBlue reagent then generates a permanent colorimetric signal at the site of the covalent tyramide deposition.
Table 4. SuperBoost EverRed and EverBlue reagent selection guide
 SuperBoost EverRed HRP Kit SuperBoost EverBlue HRP Kit
DetectionColorimetric and fluorescent
Colorimetric readoutRed colorBlue color
Fluorescence readoutTRITC/RFPCy5
Sample typeTissue
ApplicationSingle or multiplex permanent colorimetric staining for IHC
Key benefits
  • Permanent colorimetric staining
  • Can multiplex, compatible with other colorimetric counterstains
HRP Substrate Kits*E40963
E40971
E40964
E40972
SuperBoost Kits: goat anti-mouseE40965E40966
SuperBoost Kits: goat anti rabbitE40967E40968
SuperBoost kits: StreptavidinE40969E40970
*The HRP substate kits include the tyramide HRP substrate and SuperBoost EverRed or EverBlue reagent for permanent red or blue colorimetric detection in IHC using your own HRP conjugate (not provided).
Table 5. Ordering information for stand-alone poly HRP antibodies
ProductDescriptionCat. No.
SuperBoost Goat anti-Mouse Poly HRPPoly HRP secondary antibody for mouse primariesB40961
SuperBoost Goat anti-Rabbit Poly HRPPoly HRP secondary antibody for rabbit primariesB40962
SuperBoost EverRed and EverBlue experimental results
What is tyramide signal amplification?
Tyramide signal amplification is a technique used to enhance the detection of low-abundance targets in imaging applications based on the enzymatic deposition of fluorescent or chromogenic reporter molecules onto the target of interest. Alexa Fluor Tyramide SuperBoost kits and reagents are specifically designed for exceptional signal amplification, offering additional signal definition and clarity required for fluorescent imaging of low-abundance targets. SuperBoost EverRed and EverBlue colorimetric kits and reagents can also be used for permanent staining in colorimetric IHC experiments for multiplexing with DAB substrate or as an alternative colorimetric stain.
How does tyramide signal amplification work?
Tyramide signal amplification used in the SuperBoost kits utilizes the catalytic activity of horseradish peroxidase (HRP) for high-density labeling of a target protein or nucleic acid sequence in situ. Typical immunocytochemistry (ICC), immunohistochemistry (IHC) and in situ hybridization (ISH) experiments using the SuperBoost tyramide reagents require 10–5000 times less primary antibody than standard ICC/IHC/ISH experiments to achieve the same signal intensity. Since the SuperBoost kits greatly enhance specific signal intensity over background, they can be easily optimized to detect specific signal in samples where high endogenous autofluorescence is observed.
The tyramide signal amplification process includes the use of horseradish peroxidase (HRP) to enzymatically convert fluorophore or chromogen tyramides to covalently bind tyrosine residues on and surrounding the protein epitope targeted by the primary antibody. As a controlled enzymatic reaction, tyramide signal amplification does not diffuse from the site of enzyme activity and therefore, provides better spatial resolution as compared to HRP or alkaline phosphatase-based methods.
Benefits of tyramide signal amplification and deposition
  • Enable excellent detection of low-abundance targets due to poly HRP secondary antibody
  • Ability to label with antibodies from the same species due to covalent attachment of the fluorophore, allowing antibody stripping in between rounds of tyramide signal amplification
  • High sensitivity allows the use of significantly lower concentrations of primary antibody
Table 1. Invitrogen tyramide signal amplification and deposition reagent options
 Alexa Fluor Tyramide SuperBoost ReagentsSuperBoost EverRed and EverBlue Tyramide Reagents
DetectionFluorescenceColorimetric and fluorescence
UseSingle or 2-plex staining for fluorescence ICC, IHC, and ISHSingle or multiplex permanent colorimetric staining for IHC
BrightnessHigherHigh
Sample typesCells or TissuesTissues
Alexa Fluor Tyramide SuperBoost kits for signal amplification for ICC/IHC
Combining the brightness of Invitrogen Alexa Fluor dyes with poly-HRP–mediated tyramide signal amplification, SuperBoost reagents yield precision and sensitivity 10–200 times greater than standard ICC/ IHC/ISH and 2–10 times that of other tyramide amplification techniques.
Order Alexa Fluor SuperBoost Kits
Benefits of the SuperBoost tyramide signal amplification kits include:
  • A highly sensitive fluorescent imaging detection method for low-abundance, hard-to-detect targets
  • Easy-to-use kits that produce signals compatible with standard filters and microscopes
  • High-resolution images and multiplex compatibility with fluorescent proteins, DAPI, secondary antibodies, and other SuperBoost kits
Figure 2. SuperBoost Tyramide kits and reagents with poly-HRP show brighter signal for longer duration. HeLa cells were incubated with various concentrations of anti-prohibitin antibody (manufacturer recommends a 1:150 dilution, or 5 µg/mL final), then labeled with the reagents in (1) the Invitrogen Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–rabbit IgG and Alexa Fluor 488 Tyramide); (2) our original Invitrogen TSA Kit #12 (goat anti–rabbit IgG and Alexa Fluor 488 Tyramide); or (3) an Invitrogen F(ab′)2 rabbit anti–goat IgG (H+L) secondary antibody. Cell images were captured from each treatment (using the same exposure and gain) with an Invitrogen EVOS FL Auto Imaging System (see information about EVOS imaging systems). These images indicate that the Alexa Fluor 488 Tyramide SuperBoost Kit offers higher-sensitivity detection than either our original TSA kits or directly labeled secondary antibodies.
How does SuperBoost tyramide signal amplification work?
Excellent for 1 or 2 plex imaging, Alexa Fluor Tyramide SuperBoost kits are simple to use and incorporate by itself or with other reagents and fluorophores. In this workflow, the fluorophore-conjugated secondary antibodies are replaced with SuperBoost tyramide signal amplification including secondary antibodies conjugated with poly-HRP. The only additional steps are incubation with conjugated tyramides for 2–10 minutes and addition of stop solution to halt HRP activity once the specific signal is detected. The stop solution additionally helps maintain the specificity and resolution of fluorescent signal. SuperBoost tyramide signal amplification kits offer a simple workflow like those used in standard ICC, IHC, and FISH.
Workflow for SuperBoost tyramide signal kits. This is a six- or seven-step process that can be optimized to provide clear and bright signal. This workflow indicates where to add other fluorescent reagents and secondary antibody fluorophores.
Examples of SuperBoost tyramide based amplification in IF, IHC, and ISH
Alexa Fluor Tyramide SuperBoost kits are compatible with a range of other marker detection and cell staining techniques, enabling multiplex experiments and fluorescence colocalization studies. These kits work with cell types and fluorescence imaging systems commonly used in standard ICC, IHC, and ISH methods. We have tested the performance of SuperBoost tyramide signal kits using formaldehyde-fixed cell lines in 2D and 3D cultures, FFPE tissues, and cryosectioned tissues.
SuperBoost tyramide signal reagent multiplexing can be achieved with:
  • Fluorescent markers for counterstaining, such as DAPI
  • Fluorescent proteins (i.e., GFP & RFP)
  • Standard ICC/IHC
  • Additional Alexa Fluor Tyramide SuperBoost kits
Table 2. Alexa Fluor Tyramide SuperBoost reagent selection guide
 Alexa Fluor 350 Tyramide ReagentAlexa Fluor 488 Tyramide ReagentAlexa Fluor 546 Tyramide ReagentAlexa Fluor 555 Tyramide ReagentAlexa Fluor 568 Tyramide ReagentAlexa Fluor 594 Tyramide ReagentAlexa Fluor 647 Tyramide ReagentAlexa Fluor Plus 750 Tyramide ReagentBiotin-XX
DetectionFluorescence
Ex/Em347/442495/519556/573555/565579/604591/617650/668750/790N/A
Channel430488514555594594647750N/A
EVOS filterCFPGFPYFPRFPTexas RedTexas RedCy5Cy7N/A
ApplicationSingle or 2-plex staining for fluorescence ICC, IHC, and ISH
Key benefits
  • Compatible with standard filters and microscopes
  • Amplify signal 10X, meaning less primary antibody is used
Tyramide ReagentB40952B40953B40954B40955B40956B40957B40958B56131B40951
SuperBoost Kits: goat anti-mouse B40912
B40941 (50 coverslips)
 B40913 B40915
B40942 (50 coverslips)
B40916 B40911
SuperBoost Kits: goat anti-rabbit B40922
B40943 (50 coverslips)
 B40923 B40925
B40944 (50 coverslips)
B40926 B40921
SuperBoost kits: Streptavidin B40932 B40933 B40935B40921 B40931
Table 3. Ordering information for stand-alone poly HRP antibodies
ProductDescriptionCat. No.
SuperBoost Goat anti-Mouse Poly HRPPoly HRP secondary antibody for mouse primariesB40961
SuperBoost Goat anti-Rabbit Poly HRPPoly HRP secondary antibody for rabbit primariesB40962
Alexa Fluor Tyramide SuperBoost experimental results
Cells displaying signal in red, blue, and some co-localized green.
With IHC and FFPE samples
Sample type: Rat intestinal section (FFPE).
Antibodies: Immunolabeled sequentially with three primary antibodies against H2B, actin and Ki-67.
Method: In between each antibody labeling, samples were microwaved in citrate buffer pH6 on high until boiling (~2 min), then microwaved for 15 minutes at 20% power and then allowed to cool to room temperature before labeling with the next rabbit antibody. Samples were labeled with 3 different primary antibodies: rabbit anti-H2B detected with Alexa Fluor 647 Tyramide SuperBoost Kit (green), rabbit anti–smooth muscle actin antibody detected with Alexa Fluor 488 Tyramide SuperBoost Kit (red), and rabbit anti-Ki67 antibody detected with Alexa Fluor 594 Tyramide SuperBoost Kit (blue).
Cells displaying signal in red and green, and some co-localized yellow.
With ICC in cell culture
Sample type: Fixed and permeabilized HeLa cells.
Antibodies: Immunolabeled sequentially with primary antibodies against anti–ATP synthase antibody and anti–β-catenin antibody.
Method: Cells were labeled with anti–ATP synthase antibody and an Alexa Fluor 594–conjugated secondary antibody. Additionally, the cells were incubated with an anti–β-catenin antibody and labeled with the reagents in the Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 488 tyramide). Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired on a confocal microscope.
Cells displaying signal in blue  and green.
With FISH and cell culture
Sample type: Fixed and permeabilized U2OS cells.
Antibodies: Immunolabled with anti-Cas9 antibody and probed with an oligo targeting the hprt gene.
Method: U2OS cells were fixed and permeabilized and then incubated with an hprt gene probes plus inactive Cas9 protein. Hprt probes were designed for Cas9 recognition, containing sg-RNA. To detect Cas9 protein and hprt probe complex assembled at hprt loci, anti-Cas9 antibody was used. This primary antibody was detected by Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 488 tyramide) detecting hprt loci specifically. Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired and analyzed on an EVOS FL Auto Imaging System (see information about EVOS imaging systems).
SuperBoost multiplexed with standard ICC/IHC
With standard ICC/IHC
Sample type: Cultured HeLa cells, fixed and permeabilized.
Antibodies: Cells were immunolabeled with an anti–tubulin and anti–ATP synthase subunit IF1 antibody.
Method: Fixed and permeabilized HeLa cells, treated using the reagents in the Image-iT Fixation/Permeabilization Kit, were incubated with an anti-tubulin primary antibody and an Alexa Fluor 488 goat anti–mouse IgG (H+L) secondary antibody. Cells were then incubated with an anti–ATP synthase subunit IF1 antibody and labeled with the reagents in the Alexa Fluor 594 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 594 tyramide). Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired on a confocal microscope.
SuperBoost multiplexed with another SuperBoost kit
With another SuperBoost Kit
Sample type: Cultured HeLa cells, fixed and permeabilized.
Antibodies: HeLa cells were immunolabeled with an anti-prohibitin antibody and an anti–β-catenin antibody.
Method: Fixed and permeabilized HeLa cells, treated using the reagents in the Image-iT Fixation/Permeabilization Kit, were incubated with an anti-prohibitin antibody and labeled with the reagents in the Alexa Fluor 647 Tyramide SuperBoost Kit (goat anti–rabbit IgG and Alexa Fluor 647 tyramide). Additionally, cells were incubated with anti–β-catenin and labeled with the reagents in the Alexa Fluor 488 Tyramide SuperBoost Kit (goat anti–mouse IgG and Alexa Fluor 488 tyramide). Nuclei were labeled with NucBlue Fixed Cell ReadyProbes Reagent. Images were acquired on a confocal microscope.
SuperBoost EverRed and EverBlue kits for permanent colorimetric IHC
SuperBoost colorimetric signal anchoring, achieved with SuperBoost EverRed and EverBlue Colorimetric HRP Kits, enables highly sensitive and permanent target detection in multiplexable colorimetric IHC experiments. It combines poly-HRP-mediated tyramide signal deposition with SuperBoost EverRed and EverBlue for precise and sensitive colorimetric tissue staining. These SuperBoost kits use HRP catalytic activity for high-density labeling, resulting in wash-resistant, mountable, and archivable staining. SuperBoost EverRed and EverBlue provide sharp and intense signals without the need for air-drying or unconventional protocols. These user-friendly kits work with various tissue types and can be imaged with any brightfield microscope, yielding high-resolution images.
Order SuperBoost EverRed and EverBlue Kits
Benefits of the SuperBoost EverRed and EverBlue HRP Kits include:
  • Permanent colorimetric staining that is also fluorescent, allowing confirmation of the staining pattern with both brightfield and fluorescence imaging
  • Compatibility with other common colorimetric counterstains
  • Multiplexable between EverRed, EverBlue, and DAB
How does SuperBoost color signal anchoring work?
SuperBoost EverRed and EverBlue HRP Kits offer a simplified workflow similar to standard IHC staining methods. After detecting the antigen with a primary antibody, a secondary antibody or streptavidin (for biotinylated primary antibodies) conjugated with poly-HRP is used for staining. The HRP enzyme activates a chromogen tyramide, leading to localized covalent deposition of the tyramide derivative at tyrosine residues surrounding the targeted protein epitope. Treatment with the EverRed or EverBlue reagent then generates a permanent colorimetric signal at the site of the covalent tyramide deposition.
Table 4. SuperBoost EverRed and EverBlue reagent selection guide
 SuperBoost EverRed HRP Kit SuperBoost EverBlue HRP Kit
DetectionColorimetric and fluorescent
Colorimetric readoutRed colorBlue color
Fluorescence readoutTRITC/RFPCy5
Sample typeTissue
ApplicationSingle or multiplex permanent colorimetric staining for IHC
Key benefits
  • Permanent colorimetric staining
  • Can multiplex, compatible with other colorimetric counterstains
HRP Substrate Kits*E40963
E40971
E40964
E40972
SuperBoost Kits: goat anti-mouseE40965E40966
SuperBoost Kits: goat anti rabbitE40967E40968
SuperBoost kits: StreptavidinE40969E40970
*The HRP substate kits include the tyramide HRP substrate and SuperBoost EverRed or EverBlue reagent for permanent red or blue colorimetric detection in IHC using your own HRP conjugate (not provided).
Table 5. Ordering information for stand-alone poly HRP antibodies
ProductDescriptionCat. No.
SuperBoost Goat anti-Mouse Poly HRPPoly HRP secondary antibody for mouse primariesB40961
SuperBoost Goat anti-Rabbit Poly HRPPoly HRP secondary antibody for rabbit primariesB40962
SuperBoost EverRed and EverBlue experimental results
Featured resources
BioProbes article
Image of the SuperBoost BioProbes article
SuperBoost kit protocol
Image of SuperBoost tyramide protocol
Selected publications with detailed use
  1. Avens HJ, Berron BJ, May AM, Voigt KR, Seedorf GJ, Balasubramaniam V, Bowman CN. Sensitive immunofluorescent staining of cells via generation of fluorescent nanoscale polymer films in response to biorecognition. J Histochem Cytochem. 2011 Jan;59(1):76-87. PMID: 21339175 .
  2. Kosmac K, Peck BD, Walton RG, Mula J, Kern PA, Bamman MM, Dennis RA, Jacobs CA, Lattermann C, Johnson DL, Peterson CA. Immunohistochemical Identification of Human Skeletal Muscle Macrophages. Bio Protoc. 2018 Jun 20;8(12):e2883. PMID: 30148186 .
  3. Tóth ZE, Mezey E. Simultaneous visualization of multiple antigens with tyramide signal amplification using antibodies from the same species. J Histochem Cytochem. 2007 Jun;55(6):545-54. PMID: 17242468 .
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments