Stability chambers meet ICH guidelines with precise temperature and humidity control
These stability chambers provide precise control over a wide range of temperature and humidity settings. They are ideal for drug stability studies, shelf-life testing for packaged products, and insect hatching and other large-scale biological research.
Key features
Temperature
Control range of 0°C to 60°C (32°F to 140°F) in 0.1°C increments.
Humidity
Control range of above ambient to 95% @ 37°C (98.6°F)
Extensive cabinet options
Doors, shelves, access ports, sockets, shelving, chart recorders and shaker systems.
Extensive humidity system options
Designed to accommodate labs with a variety of water access and drainage needs.
Achieves regulatory compliance
Meets International Conference on Harmonization (ICH) guidelines for drug substance and product storage testing.
Controlling relative humidity
This chart illustrates the maximum and minimum attainable humidity within the cabinet at a given set temperature under the following nominal conditions: refrigeration on, defrost off, no wet bulb recorder, no air exchange, set for 22.2°C (72°F) and 50% RH ambient. Optional heatless dryer is available for lower humidity levels.
Humidity range of the Forma Environmental Chamber
Specifications
Large Stability Chamber
Small Stability Chamber
Size
29 cu. ft.
11 cu. ft.
Exterior (W x H x D)
965 x 2248 x 813mm
38.00 x 88.50 x 32.00 in.
965 x 1308 x 813mm
38.00 x 51.50 x 32.00 in.
Interior (W x H x D)
787 x 1524 x 686mm
31.00 x 60.00 x 27.00 in
787 x 609 x 686mm
31.00 x 24.00 x 27.00 in.
Net weight
347kg / 765 lbs
261kg / 575 lbs
Shipping weight
472kg / 1040 lbs
325kg / 715 lbs
Temperature
Temperature range
0°C to 60°C (32°F to 140°F)
Controller, sensor
Watlow Microprocessor - PID, with RTD sensor
Display
Digital dual LED, 0.1°C steps readability & setability
Programmability
Single set point, ramp & soak (to 40 steps)
Temp. uniformity @ 20 to 37°C
≤ ±0.3°C
Temp. stability @ 20 to 37°C
≤ ±0.1°C 
Humidity
Water consumption / specification
Max. 12L per day / 3 gal per day;
demineralized water, resistance: 0.5 - 1 MΩ cm
Relative humidity range
Above ambient to 95% @ 37°C (98.6°F) ±5.0% RH
Water connection*
Fill port: 1⁄8" female NPT, drain port: 3⁄8" FPT & 3⁄8" O.D. P-trap right rear, initial fill approx. 1L / 1qt.
Electrical
Voltage
208-220V, 60Hz
Plug
NEMA 14-20P
Power switch
2-Pole
Dry Contacts
Common, NO, NC
Data Output
4-20 milliamp, temperature and RH
Access port
54mm / 2.1in diameter; left side; with exterior and interior silicone stopper
Options
Cabinet Options Available
Choice of access ports
Duplex sockets
Choice of chart recorders
Infrared CO2 Control Package
Choice of door types and styles, including Lexan(tm) inner doors
Choice of shaker support systems
Temperature mapping and calibration certificate
Humidity Systems Options Available
Heatless dryer
Condensate pump kit
Condensate evaporator
Carboy kit
Order Number
3940
3911