Search Thermo Fisher Scientific
With an expansive range of automation tips, Thermo Scientific provide solutions for your liquid handling needs. Our products help increase efficiency through accurate and precise performance. We have more the 200 Automation tips designed to fit a variety of liquid handling workstation and pipetting head variations.
Every production lot of our automation tips undergoes a through quality inspection focusing on four specific areas: Visual, dimensional, functional and purity*
Inspection focus | Details |
Visual | Identify distortion, short shots, bubbles, cloudiness, knit lines, and blemish |
Dimensional | Measure physical attributes such as tip length, straightness, and inner/outer diameter |
Functional | Tests the interaction of the tip with the acutal workstation. Tip fit/seal and pipetting performance (%CV) and proper tip ejection |
Purity | Certifies sterility, products free of RNA, DNA, RNase, DNase, pyrogens and ATP. |
Tecan |
Beckman | Revvity |
Qiagen | Eppendorf |
---|---|---|---|---|
![]() |
![]() |
![]() |
![]() |
![]() |
Tecan Freedom Evo Tecan Te-MO Tecan Genesis |
Beckman Biomek FX Beckman Biomek NX Beckman Biomek 3000 |
JANUS MultiPROBE II PerkinElmer SciClone PerkinElmer Zephyr |
BioRobot 3000/8000/9600/9604 | epMotion |
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.