Engineered with the value to transfer your samples
Thermo Scientific pipette tips are developed through robust engineering, innovative design, extensive performance testing, and only exceptional materials. We understand how valuable your samples are and develop pipette tips that meet or exceed your expectations.
Universal fit on pipettes
Thermo Scientific ART universal pipette tips enhance pipetting efficiency across various pipette brands. Engineered to fit seamlessly, these pipette tips provide optional performance and compatibility to help ensure a hassle-free and efficient pipetting experience. We understand the importance of precision and accuracy in scientific research, which is why ART tips are manufactured with high-quality, ultra-clear polypropylene to enable optimal clarity for easy sample visualization and monitoring liquid levels. ART pipette tips are available in a wide range of volume capacities, catering to the diverse needs of scientific researchers.
Lower attachment and ejection pipette tips
We understand the importance of efficient and hassle-free pipetting in the laboratory workflow.  That’s why our pipette tips are designed with innovative lower attachment and ejection features, helping ensure a seamless and reliable pipetting experience. The lower attachment feature is engineered to securely and firmly attach to the pipette, helping prevent leakage and dislodgement during pipetting procedures.  An easy ejection mechanism allows for effortless and quick removal of the pipette after use, helping reduce pipetting strain and improving pipetting efficiency.
Lower retention for optimal transference
Our low retention pipette tips are engineered to minimize sample loss and enable the maximum transfer of valuable sample.  One of the key features is the use of hydrophobic material to minimize the adhesion of liquid to the inner walls of the tips. The reduced surface tension helps prevent liquid droplets from forming and clinging, helping improve accuracy and reproducibility in pipetting tasks. 
Sustainable packaging and tip options
We are committed to minimizing our environmental impact and promoting sustainability in all aspects of our business.  We strive to minimize single-use packaging as well as the size, weight, and transportation cost of our packaging.  We have implemented the ACT label to signify our dedication to responsible practices.  We understand the importance of recycling in the laboratory setting and the need to reduce waste. 
Tips certified for quality and fit for your application
We prioritize the quality and performance of our pipette tips. Our pipette tips are manufactured in accordance with certified quality standards including ISO 9001, ISO 14001, and ISO 13485 to enable optimal performance and reliable results.  We designate pipette tips that are sterile, PCR-ready, and forensic ready. 
Sterile and PCR badge 250
Color-coded volume identification
Our pipette tips are designed with color-coded volume identification, allowing researchers to select the correct pipette tip size and volume capacity for their specific application. Furthermore, our color-coded volume identification system is standardized across our entire range of pipette tips, helping ensure consistency and compatibility with various pipette branches.  Experience the convenience and reliability of our color-coded volume identification system.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.