Search Thermo Fisher Scientific
Organism Type | Rabbit | Rabbit kidney (RK13) cells (provided by Dr. Dennis O’Callaghan, Department of Microbiology and Immunology, Louisiana State University Health Sciences Center, Shreveport, LA) were maintained and cultured in DMEM supplemented with 10% bovine calf serum and antibiotics/antimycotics. |
Tissues | Kidney | |
Phenotype | Adherent | |
Primary | no | |
Application | RNAi |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||