Cryo-electron tomography sample preparation workflow
Cryo-electron tomography (cryo-ET) is a technique that allows the study of the 3D structure of cells and tissues at near-native conditions. Sample preparation is a critical step in cryo-ET, which can greatly affect the quality and resolution of the results. Cryo-ET has been applied to many different sample types, from single molecules to protein complexes, viruses, bacteria, cells, to tissue cells and large tissue samples. Cells are either vitrified through plunge-freezing (as in single particle analysis) or high pressure freezing (HPF). Plunge-freezing is suitable for samples that are relatively thin or have a limited amount of materials. For larger specimens like organoids or bulk tissue, high pressure freezing is commonly used. After the vitrification step, depending on the size of the sample, a thinning step is included in the sample preparation using focused ion beam (FIB) or plasma ion beam milling to produce cryo-lamella that can be penetrated by electron beams.  
Cryo-ET sample preparation using plunge freezing
Sample Prep Cryo Tomo Workflow
Cells seeded and grown on a TEM grid support
Cells seeded and grown on a TEM grid support
To prepare samples for cellular cryo-ET, routine culture methods are used to prepare cells in bulk, once your culture is ready the cells are prepared into a suspension and seeded onto carbon-coated gold (for biocompatibility) electron microscopy grids. These grids can be additionally functionalized for better cellular adhesion or distribution. Depends on the cell type, there are two different workflows can be used. For cells naturally in suspension they can be applied in the Thermo Scientific Vitrobot System directly to the grid. For adherent or large cells, they require incubation for 1-7 days to allow adhesion and flattening of the cells. When the cells are evenly spaced on the grid, they are taken for plunge freezing.  
Locating the structure of interest can be difficult in the vast complexity of the natural cellular environment. However, through correlative light and electron microscopy, (CLEM), various florescent labels can be used to tag cells. For the cellular cryo-tomography workflow, Thermo Scientific iFLM (Integrated Fluorescence Light Microscope) Correlative System combines light and electron microscopy into one system, eliminating extra sample transfer steps and enabling a streamlined cryo-correlative solution for cryo-tomography. 
Cryo ET lamella prepared with the Aquilos 2 Cryo FIB with overlay of SEM image and fluorescence image
Cell culture media
Gibco FBS Performance Media
Cell collection and culture plastics
Cell culture plastics
Fluorescence microscopy reagents
Chinese Hamster Ovary cells on a grid with overlaid fluorescence, Hoechst Blue and Mitotracker
Cryo-ET sample preparation using high pressure freezing
Cryo-tomography imaging of bulk samples like organoids or larger models allows imaging at more life-like conditions for the sample. The cryofixation of samples with thickness of several hundred micrometers such as tissue biopsies, organoids, plant tissues, etc., is only possible by high-pressure freezing. High-pressure freezing is a reliable method for cryoimmobilization of versatile cellular specimens. Samples are subjected to high pressure during freezing with nitrogen, to prevent formation of ordered crystalline ice which expands in volume during freezing.
For imaging within the TEM, the most applied method is the sandwich or waffle method. Here a TEM grid is loaded with tens of microns of sample and processes similarly to the in-situ workflow. The second method is cryo-lift out, a method most like laser dissection. Here a piece of tissue is extracted from site-specific regions with the high-precision Thermo Scientific EasyLift NanoManipulator. These lamellas can subsequently be placed inside Autogrids for cryo-TEM and further imaging of protein structures and protein networks.
For bulk samples like biopsies, which might not have any fluorescence, another exciting workflow is volume electron microscopy (vEM).  Here, an HPF sample is serially sliced with the ion beam, each time imaging the freshly cut surface. This information can be reconstructed into a 3D volume. This suits biological questions which require resolution in nanometers rather than angstroms. These 2 methods can be combined to provide ways to find regions of interest which are not fluorescently labelled.
High-pressure frozen mouse brain sample for cryo-ET analysis.
Contextual volume imaging of high-pressure frozen mouse brain with no added stains or fixatives viewed with Thermo Scientific Auto Slice and View Software. Sample courtesy of MRC-LMB.
Cryo-lift-out lamella preparation
Prepare lamellas from specific targeted regions with nanometer-position accuracy. The high-precision EasyLift NanoManipulator option allows for extraction of site-specific regions, such as fluorescently labeled proteins, even from high-pressure frozen samples. These lamellas can subsequently be placed inside Autogrids for cryo-TEM and further imaging of protein structures and protein networks.
Organoids, Spheroids, and 3D Cell Culture
3D Cell Culture
Aquilos 2 Cryo-FIB
Thermo Scientific Aquilos 2 Cryo-FIB